Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 91217114-91217657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUH
Entrez Gene ID
549
Gene Name
AU RNA binding protein/enoyl-CoA hydratase
Gene Aliases
-
UniGene
Hs.175905
Gene Chromosome Location
Chr.9: 91213815-91361969 on Build GRCh38
Transcripts
NM_001306190.1 NM_001698.2 XM_005252066.3 XM_005252067.4 XM_005252069.4 XM_006717150.3 XM_011518802.2 XM_017014849.1 XM_017014850.1 XM_017014851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4664744 c.879T>G p.I293M Chr.9: 91217292-91217292 on Build GRCh38
6184455 c.893A>G p.E298G Chr.9: 91217278-91217278 on Build GRCh38
309396 c.888delG p.M297fs*5 Chr.9: 91217283-91217283 on Build GRCh38
3908803 c.865G>A p.A289T Chr.9: 91217306-91217306 on Build GRCh38
5958212 c.856A>G p.M286V Chr.9: 91217315-91217315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747814597 Chr.9: 91217214-91217214 on Build GRCh38
rs1000454412 Chr.9: 91217198-91217198 on Build GRCh38
rs973223235 Chr.9: 91217485-91217485 on Build GRCh38
rs758771631 Chr.9: 91217288-91217288 on Build GRCh38
rs780581499 Chr.9: 91217291-91217291 on Build GRCh38
rs763784488 Chr.9: 91217377-91217377 on Build GRCh38
rs863223913 Chr.9: 91217305-91217305 on Build GRCh38
rs770380014 Chr.9: 91217620-91217620 on Build GRCh38
rs781702997 Chr.9: 91217364-91217364 on Build GRCh38
rs962131124 Chr.9: 91217472-91217472 on Build GRCh38
rs759318765 Chr.9: 91217240-91217240 on Build GRCh38
rs531787520 Chr.9: 91217139-91217139 on Build GRCh38
rs767247333 Chr.9: 91217370-91217370 on Build GRCh38
rs577813233 Chr.9: 91217476-91217476 on Build GRCh38
rs925706050 Chr.9: 91217607-91217607 on Build GRCh38
rs767302239 Chr.9: 91217249-91217249 on Build GRCh38
rs747389638 Chr.9: 91217149-91217149 on Build GRCh38
rs571717648 Chr.9: 91217376-91217376 on Build GRCh38
rs891900371 Chr.9: 91217361-91217361 on Build GRCh38
rs758834249 Chr.9: 91217367-91217368 on Build GRCh38
rs565432767 Chr.9: 91217161-91217161 on Build GRCh38
rs932435545 Chr.9: 91217171-91217171 on Build GRCh38
rs553714523 Chr.9: 91217575-91217575 on Build GRCh38
rs756770155 Chr.9: 91217272-91217272 on Build GRCh38
rs532853132 Chr.9: 91217187-91217187 on Build GRCh38
rs551351122 Chr.9: 91217269-91217269 on Build GRCh38
rs940511726 Chr.9: 91217222-91217222 on Build GRCh38
rs1049752683 Chr.9: 91217290-91217290 on Build GRCh38
rs748245565 Chr.9: 91217321-91217321 on Build GRCh38
rs769924401 Chr.9: 91217347-91217347 on Build GRCh38
rs774202555 Chr.9: 91217362-91217362 on Build GRCh38
rs141731177 Chr.9: 91217625-91217625 on Build GRCh38
rs905934967 Chr.9: 91217412-91217412 on Build GRCh38
rs1014354818 Chr.9: 91217447-91217447 on Build GRCh38
rs746444416 Chr.9: 91217282-91217282 on Build GRCh38
rs112394234 Chr.9: 91217560-91217560 on Build GRCh38
rs1049535037 Chr.9: 91217172-91217172 on Build GRCh38
rs976377524 Chr.9: 91217559-91217559 on Build GRCh38
rs753190979 Chr.9: 91217361-91217362 on Build GRCh38
rs1035023643 Chr.9: 91217415-91217415 on Build GRCh38
rs554400107 Chr.9: 91217536-91217537 on Build GRCh38
rs984035405 Chr.9: 91217506-91217506 on Build GRCh38
rs561040345 Chr.9: 91217626-91217626 on Build GRCh38
rs189293998 Chr.9: 91217314-91217314 on Build GRCh38
rs771770937 Chr.9: 91217424-91217424 on Build GRCh38
rs780228199 Chr.9: 91217140-91217140 on Build GRCh38
rs199569104 Chr.9: 91217307-91217307 on Build GRCh38
rs573455764 Chr.9: 91217582-91217582 on Build GRCh38
rs111299534 Chr.9: 91217382-91217382 on Build GRCh38
rs550394259 Chr.9: 91217158-91217158 on Build GRCh38
rs373678498 Chr.9: 91217294-91217294 on Build GRCh38
rs920463911 Chr.9: 91217517-91217517 on Build GRCh38
rs377735590 Chr.9: 91217258-91217258 on Build GRCh38
rs567397989 Chr.9: 91217372-91217372 on Build GRCh38
rs931917327 Chr.9: 91217274-91217274 on Build GRCh38
rs867583124 Chr.9: 91217557-91217557 on Build GRCh38
rs776767438 Chr.9: 91217315-91217315 on Build GRCh38
rs180921881 Chr.9: 91217409-91217409 on Build GRCh38
rs765291575 Chr.9: 91217342-91217345 on Build GRCh38
rs146338941 Chr.9: 91217435-91217435 on Build GRCh38
rs1014658164 Chr.9: 91217458-91217458 on Build GRCh38
rs549084783 Chr.9: 91217369-91217369 on Build GRCh38
rs959703060 Chr.9: 91217416-91217416 on Build GRCh38
rs779550477 Chr.9: 91217275-91217275 on Build GRCh38
rs986010367 Chr.9: 91217574-91217574 on Build GRCh38

View Full Product Details