Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92466190-92466722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPN
Entrez Gene ID
54829
Gene Name
asporin
Gene Aliases
OS3, PLAP-1, PLAP1, SLRR1C
UniGene
Hs.435655
Gene Chromosome Location
Chr.9: 92456205-92482562 on Build GRCh38
Transcripts
NM_001193335.1 NM_017680.4 NM_001012267.2 NM_001286969.1 XM_011518689.1

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001193335.1 NM_017680.4 NM_001012267.2 NM_001286969.1 XM_011518689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5120146 c.601+4A>C p.? Chr.9: 92466354-92466354 on Build GRCh38
3982969 c.465G>C p.L155L Chr.9: 92466494-92466494 on Build GRCh38
6232074 c.510C>A p.P170P Chr.9: 92466449-92466449 on Build GRCh38
6205706 c.388-3A>C p.? Chr.9: 92466574-92466574 on Build GRCh38
3908886 c.457C>T p.R153* Chr.9: 92466502-92466502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531268036 Chr.9: 92466244-92466244 on Build GRCh38
rs371768627 Chr.9: 92466501-92466501 on Build GRCh38
rs750143544 Chr.9: 92466476-92466476 on Build GRCh38
rs931618671 Chr.9: 92466379-92466379 on Build GRCh38
rs774799220 Chr.9: 92466312-92466315 on Build GRCh38
rs776917242 Chr.9: 92466478-92466478 on Build GRCh38
rs537347263 Chr.9: 92466607-92466607 on Build GRCh38
rs952024088 Chr.9: 92466286-92466286 on Build GRCh38
rs778643481 Chr.9: 92466445-92466445 on Build GRCh38
rs757923286 Chr.9: 92466454-92466454 on Build GRCh38
rs746176926 Chr.9: 92466444-92466444 on Build GRCh38
rs575015780 Chr.9: 92466639-92466639 on Build GRCh38
rs143358512 Chr.9: 92466502-92466502 on Build GRCh38
rs138209327 Chr.9: 92466571-92466571 on Build GRCh38
rs541260748 Chr.9: 92466623-92466623 on Build GRCh38
rs753528217 Chr.9: 92466433-92466433 on Build GRCh38
rs371856281 Chr.9: 92466589-92466589 on Build GRCh38
rs368329214 Chr.9: 92466469-92466469 on Build GRCh38
rs576973569 Chr.9: 92466599-92466599 on Build GRCh38
rs1047938581 Chr.9: 92466296-92466296 on Build GRCh38
rs765345253 Chr.9: 92466372-92466372 on Build GRCh38
rs200280971 Chr.9: 92466543-92466543 on Build GRCh38
rs929488866 Chr.9: 92466290-92466290 on Build GRCh38
rs1006311203 Chr.9: 92466353-92466353 on Build GRCh38
rs770217740 Chr.9: 92466610-92466610 on Build GRCh38
rs866116107 Chr.9: 92466519-92466519 on Build GRCh38
rs780513757 Chr.9: 92466458-92466458 on Build GRCh38
rs374932025 Chr.9: 92466587-92466587 on Build GRCh38
rs41278695 Chr.9: 92466381-92466381 on Build GRCh38
rs41278697 Chr.9: 92466418-92466418 on Build GRCh38
rs757082299 Chr.9: 92466551-92466551 on Build GRCh38
rs775538414 Chr.9: 92466534-92466534 on Build GRCh38
rs565810963 Chr.9: 92466491-92466491 on Build GRCh38
rs758355889 Chr.9: 92466691-92466691 on Build GRCh38
rs369326728 Chr.9: 92466453-92466453 on Build GRCh38
rs747550942 Chr.9: 92466461-92466461 on Build GRCh38
rs569274142 Chr.9: 92466515-92466515 on Build GRCh38
rs921706454 Chr.9: 92466366-92466366 on Build GRCh38
rs762136374 Chr.9: 92466314-92466315 on Build GRCh38
rs41278699 Chr.9: 92466494-92466494 on Build GRCh38
rs146775001 Chr.9: 92466394-92466394 on Build GRCh38
rs776877419 Chr.9: 92466342-92466342 on Build GRCh38
rs966554052 Chr.9: 92466450-92466450 on Build GRCh38
rs769055070 Chr.9: 92466330-92466330 on Build GRCh38
rs187543149 Chr.9: 92466321-92466321 on Build GRCh38
rs1013763853 Chr.9: 92466457-92466457 on Build GRCh38
rs756921551 Chr.9: 92466439-92466439 on Build GRCh38
rs747321863 Chr.9: 92466329-92466329 on Build GRCh38
rs542327992 Chr.9: 92466646-92466646 on Build GRCh38
rs763940804 Chr.9: 92466542-92466542 on Build GRCh38
rs772512883 Chr.9: 92466312-92466312 on Build GRCh38
rs755951081 Chr.9: 92466414-92466414 on Build GRCh38
rs760165232 Chr.9: 92466549-92466551 on Build GRCh38
rs767126722 Chr.9: 92466436-92466438 on Build GRCh38
rs760762922 Chr.9: 92466538-92466538 on Build GRCh38
rs144954352 Chr.9: 92466365-92466365 on Build GRCh38
rs748558453 Chr.9: 92466480-92466480 on Build GRCh38
rs1027954605 Chr.9: 92466274-92466279 on Build GRCh38
rs928817053 Chr.9: 92466289-92466289 on Build GRCh38
rs781724741 Chr.9: 92466604-92466604 on Build GRCh38
rs769057452 Chr.9: 92466470-92466470 on Build GRCh38
rs778120761 Chr.9: 92466613-92466613 on Build GRCh38
rs371262202 Chr.9: 92466403-92466403 on Build GRCh38
rs367736538 Chr.9: 92466602-92466602 on Build GRCh38
rs769292219 Chr.9: 92466650-92466650 on Build GRCh38
rs148376603 Chr.9: 92466598-92466598 on Build GRCh38
rs763128075 Chr.9: 92466386-92466386 on Build GRCh38
rs41278701 Chr.9: 92466634-92466634 on Build GRCh38
rs1014804612 Chr.9: 92466590-92466590 on Build GRCh38
rs374381931 Chr.9: 92466452-92466452 on Build GRCh38

View Full Product Details