Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92712443-92712956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICD2
Entrez Gene ID
23299
Gene Name
BICD cargo adaptor 2
Gene Aliases
SMALED2, bA526D8.1
UniGene
Hs.436939
Gene Chromosome Location
Chr.9: 92711363-92764841 on Build GRCh38
Transcripts
NM_001003800.1 NM_015250.3 XM_017014551.1

SNP Detail

SNP ID Chromosome Location
rs190219056 Chr.9: 92712741-92712741 on Build GRCh38
rs190790523 Chr.9: 92712869-92712869 on Build GRCh38
rs931037835 Chr.9: 92712527-92712527 on Build GRCh38
rs185358007 Chr.9: 92712715-92712715 on Build GRCh38
rs558839697 Chr.9: 92712898-92712898 on Build GRCh38
rs939840197 Chr.9: 92712495-92712495 on Build GRCh38
rs968543070 Chr.9: 92712776-92712776 on Build GRCh38
rs778478109 Chr.9: 92712562-92712562 on Build GRCh38
rs779869172 Chr.9: 92712897-92712897 on Build GRCh38
rs750260288 Chr.9: 92712734-92712734 on Build GRCh38
rs961242639 Chr.9: 92712880-92712880 on Build GRCh38
rs181590675 Chr.9: 92712607-92712607 on Build GRCh38
rs1031096048 Chr.9: 92712629-92712629 on Build GRCh38
rs992729968 Chr.9: 92712678-92712678 on Build GRCh38
rs968056066 Chr.9: 92712544-92712544 on Build GRCh38
rs41273372 Chr.9: 92712825-92712825 on Build GRCh38
rs902050065 Chr.9: 92712728-92712728 on Build GRCh38
rs889427484 Chr.9: 92712656-92712656 on Build GRCh38
rs919677147 Chr.9: 92712509-92712509 on Build GRCh38
rs1049878672 Chr.9: 92712577-92712577 on Build GRCh38
rs542396951 Chr.9: 92712905-92712905 on Build GRCh38
rs1011518558 Chr.9: 92712513-92712513 on Build GRCh38
rs1007840820 Chr.9: 92712671-92712671 on Build GRCh38
rs1000937025 Chr.9: 92712814-92712814 on Build GRCh38
rs766494601 Chr.9: 92712915-92712915 on Build GRCh38
rs528732083 Chr.9: 92712658-92712658 on Build GRCh38
rs753371349 Chr.9: 92712933-92712933 on Build GRCh38
rs182153690 Chr.9: 92712774-92712774 on Build GRCh38
rs771978389 Chr.9: 92712890-92712890 on Build GRCh38
rs1036781638 Chr.9: 92712500-92712500 on Build GRCh38
rs746766435 Chr.9: 92712916-92712916 on Build GRCh38
rs546867625 Chr.9: 92712664-92712664 on Build GRCh38
rs186267521 Chr.9: 92712862-92712862 on Build GRCh38
rs1009732650 Chr.9: 92712769-92712769 on Build GRCh38
rs899738003 Chr.9: 92712476-92712476 on Build GRCh38
rs999578619 Chr.9: 92712612-92712612 on Build GRCh38
rs919564083 Chr.9: 92712810-92712810 on Build GRCh38
rs1049476272 Chr.9: 92712911-92712911 on Build GRCh38
rs959389516 Chr.9: 92712852-92712852 on Build GRCh38
rs960822043 Chr.9: 92712676-92712678 on Build GRCh38
rs974370817 Chr.9: 92712777-92712777 on Build GRCh38
rs1042989933 Chr.9: 92712900-92712900 on Build GRCh38
rs899686198 Chr.9: 92712891-92712891 on Build GRCh38

View Full Product Details