Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92720210-92720711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICD2
Entrez Gene ID
23299
Gene Name
BICD cargo adaptor 2
Gene Aliases
SMALED2, bA526D8.1
UniGene
Hs.436939
Gene Chromosome Location
Chr.9: 92711363-92764841 on Build GRCh38
Transcripts
NM_001003800.1 NM_015250.3 XM_017014551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6611569 c.754G>A p.E252K Chr.9: 92720608-92720608 on Build GRCh38
6611564 c.746C>T p.T249M Chr.9: 92720616-92720616 on Build GRCh38
369031 c.778G>T p.E260* Chr.9: 92720584-92720584 on Build GRCh38
4505499 c.694C>T p.L232F Chr.9: 92720668-92720668 on Build GRCh38
1111168 c.915C>T p.H305H Chr.9: 92720447-92720447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369921309 Chr.9: 92720252-92720252 on Build GRCh38
rs780098431 Chr.9: 92720435-92720435 on Build GRCh38
rs753891194 Chr.9: 92720406-92720406 on Build GRCh38
rs145083783 Chr.9: 92720615-92720615 on Build GRCh38
rs566510315 Chr.9: 92720559-92720559 on Build GRCh38
rs747084103 Chr.9: 92720598-92720600 on Build GRCh38
rs766675228 Chr.9: 92720293-92720293 on Build GRCh38
rs375710311 Chr.9: 92720553-92720553 on Build GRCh38
rs781147760 Chr.9: 92720544-92720544 on Build GRCh38
rs772599458 Chr.9: 92720457-92720457 on Build GRCh38
rs772316990 Chr.9: 92720366-92720368 on Build GRCh38
rs200288349 Chr.9: 92720381-92720381 on Build GRCh38
rs111250580 Chr.9: 92720273-92720273 on Build GRCh38
rs748997644 Chr.9: 92720634-92720634 on Build GRCh38
rs775974479 Chr.9: 92720580-92720580 on Build GRCh38
rs745452446 Chr.9: 92720648-92720648 on Build GRCh38
rs373858530 Chr.9: 92720447-92720447 on Build GRCh38
rs756806772 Chr.9: 92720636-92720636 on Build GRCh38
rs761087136 Chr.9: 92720603-92720603 on Build GRCh38
rs754659441 Chr.9: 92720671-92720671 on Build GRCh38
rs748866068 Chr.9: 92720557-92720557 on Build GRCh38
rs768612898 Chr.9: 92720443-92720443 on Build GRCh38
rs757409146 Chr.9: 92720513-92720513 on Build GRCh38
rs371897802 Chr.9: 92720629-92720629 on Build GRCh38
rs761526161 Chr.9: 92720376-92720376 on Build GRCh38
rs768173580 Chr.9: 92720501-92720501 on Build GRCh38
rs762107122 Chr.9: 92720678-92720678 on Build GRCh38
rs760001632 Chr.9: 92720480-92720480 on Build GRCh38
rs777795062 Chr.9: 92720338-92720338 on Build GRCh38
rs201389004 Chr.9: 92720649-92720649 on Build GRCh38
rs562617547 Chr.9: 92720573-92720573 on Build GRCh38
rs533852054 Chr.9: 92720609-92720609 on Build GRCh38
rs1007045018 Chr.9: 92720470-92720470 on Build GRCh38
rs752952319 Chr.9: 92720393-92720393 on Build GRCh38
rs777900476 Chr.9: 92720480-92720482 on Build GRCh38
rs77530912 Chr.9: 92720318-92720318 on Build GRCh38
rs767895612 Chr.9: 92720484-92720484 on Build GRCh38
rs372821254 Chr.9: 92720494-92720494 on Build GRCh38
rs367698188 Chr.9: 92720430-92720431 on Build GRCh38
rs147814238 Chr.9: 92720383-92720383 on Build GRCh38
rs754243885 Chr.9: 92720281-92720281 on Build GRCh38
rs769856791 Chr.9: 92720249-92720249 on Build GRCh38
rs533199924 Chr.9: 92720610-92720610 on Build GRCh38
rs769512205 Chr.9: 92720452-92720452 on Build GRCh38
rs878999998 Chr.9: 92720432-92720432 on Build GRCh38
rs750471177 Chr.9: 92720612-92720612 on Build GRCh38
rs368131334 Chr.9: 92720387-92720387 on Build GRCh38
rs778828848 Chr.9: 92720517-92720517 on Build GRCh38
rs762587567 Chr.9: 92720464-92720464 on Build GRCh38
rs200066450 Chr.9: 92720633-92720633 on Build GRCh38
rs779331320 Chr.9: 92720569-92720569 on Build GRCh38
rs368860496 Chr.9: 92720312-92720312 on Build GRCh38
rs775155214 Chr.9: 92720371-92720371 on Build GRCh38
rs768559794 Chr.9: 92720374-92720374 on Build GRCh38
rs761467367 Chr.9: 92720277-92720277 on Build GRCh38
rs772504613 Chr.9: 92720576-92720576 on Build GRCh38
rs201922946 Chr.9: 92720272-92720272 on Build GRCh38
rs770331418 Chr.9: 92720473-92720473 on Build GRCh38
rs115787125 Chr.9: 92720391-92720391 on Build GRCh38
rs369847588 Chr.9: 92720614-92720614 on Build GRCh38
rs758897472 Chr.9: 92720288-92720288 on Build GRCh38
rs774968696 Chr.9: 92720474-92720474 on Build GRCh38
rs41273374 Chr.9: 92720254-92720254 on Build GRCh38
rs776953074 Chr.9: 92720608-92720608 on Build GRCh38
rs754913360 Chr.9: 92720537-92720537 on Build GRCh38
rs905682709 Chr.9: 92720574-92720574 on Build GRCh38
rs771197200 Chr.9: 92720628-92720629 on Build GRCh38
rs747485547 Chr.9: 92720666-92720666 on Build GRCh38
rs201926954 Chr.9: 92720405-92720405 on Build GRCh38
rs776420956 Chr.9: 92720275-92720275 on Build GRCh38
rs777536867 Chr.9: 92720556-92720556 on Build GRCh38
rs757281343 Chr.9: 92720409-92720409 on Build GRCh38
rs771991551 Chr.9: 92720350-92720350 on Build GRCh38
rs79542696 Chr.9: 92720390-92720390 on Build GRCh38
rs1002664605 Chr.9: 92720590-92720590 on Build GRCh38
rs141444039 Chr.9: 92720525-92720525 on Build GRCh38
rs753994970 Chr.9: 92720510-92720510 on Build GRCh38
rs148937530 Chr.9: 92720616-92720616 on Build GRCh38
rs950357888 Chr.9: 92720354-92720354 on Build GRCh38
rs745818814 Chr.9: 92720345-92720345 on Build GRCh38
rs769096602 Chr.9: 92720670-92720670 on Build GRCh38
rs746899633 Chr.9: 92720436-92720436 on Build GRCh38
rs772728967 Chr.9: 92720382-92720382 on Build GRCh38
rs753005623 Chr.9: 92720485-92720485 on Build GRCh38
rs760894899 Chr.9: 92720492-92720492 on Build GRCh38
rs764345086 Chr.9: 92720607-92720607 on Build GRCh38
rs771797323 Chr.9: 92720260-92720260 on Build GRCh38
rs369298186 Chr.9: 92720637-92720637 on Build GRCh38
rs775310481 Chr.9: 92720269-92720269 on Build GRCh38
rs755156975 Chr.9: 92720297-92720297 on Build GRCh38
rs1051614965 Chr.9: 92720418-92720418 on Build GRCh38
rs571839806 Chr.9: 92720386-92720386 on Build GRCh38
rs1051176702 Chr.9: 92720419-92720419 on Build GRCh38
rs1033255864 Chr.9: 92720237-92720237 on Build GRCh38
rs751891675 Chr.9: 92720295-92720295 on Build GRCh38
rs1031553880 Chr.9: 92720266-92720266 on Build GRCh38
rs377363410 Chr.9: 92720336-92720336 on Build GRCh38
rs555422625 Chr.9: 92720479-92720479 on Build GRCh38
rs540280844 Chr.9: 92720444-92720444 on Build GRCh38
rs765309802 Chr.9: 92720686-92720686 on Build GRCh38
rs751319533 Chr.9: 92720645-92720645 on Build GRCh38
rs776027761 Chr.9: 92720650-92720650 on Build GRCh38
rs150389188 Chr.9: 92720549-92720549 on Build GRCh38
rs373355173 Chr.9: 92720278-92720278 on Build GRCh38
rs368490787 Chr.9: 92720546-92720546 on Build GRCh38
rs1051133171 Chr.9: 92720241-92720241 on Build GRCh38
rs374569189 Chr.9: 92720375-92720375 on Build GRCh38
rs371444272 Chr.9: 92720372-92720372 on Build GRCh38

View Full Product Details