Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93318121-93318627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf129
Entrez Gene ID
445577
Gene Name
chromosome 9 open reading frame 129
Gene Aliases
bA165J3.3
UniGene
Hs.575636
Gene Chromosome Location
Chr.9: 93318199-93346414 on Build GRCh38
Transcripts
NM_001098808.1 XM_006717117.3 NM_001282394.1 NM_006648.3 XM_005252137.3 XM_005252140.2 XM_005252141.2 XM_011518926.2 XM_011518927.2 XM_011518928.2 XM_011518929.2 XM_011518930.2 XM_011518931.2 XM_011518932.2 XM_011518935.2 XM_011518936.2 XM_011518937.2 XM_011518939.2 XM_011518940.2 XM_017015045.1 XM_017015046.1 XM_017015047.1 XM_017015048.1 XM_017015049.1 XM_017015050.1 XM_017015051.1 XM_017015052.1 XM_017015053.1 XM_017015054.1 XM_017015055.1

Gene Symbol
WNK2
Entrez Gene ID
65268
Gene Name
WNK lysine deficient protein kinase 2
Gene Aliases
NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43
UniGene
Hs.654856
Gene Chromosome Location
Chr.9: 93184156-93327581 on Build GRCh38
Transcripts
NM_001098808.1 XM_006717117.3 NM_001282394.1 NM_006648.3 XM_005252137.3 XM_005252140.2 XM_005252141.2 XM_011518926.2 XM_011518927.2 XM_011518928.2 XM_011518929.2 XM_011518930.2 XM_011518931.2 XM_011518932.2 XM_011518935.2 XM_011518936.2 XM_011518937.2 XM_011518939.2 XM_011518940.2 XM_017015045.1 XM_017015046.1 XM_017015047.1 XM_017015048.1 XM_017015049.1 XM_017015050.1 XM_017015051.1 XM_017015052.1 XM_017015053.1 XM_017015054.1 XM_017015055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5669804 c.499G>A p.G167S Chr.9: 93318490-93318490 on Build GRCh38
375258 c.554C>A p.P185H Chr.9: 93318435-93318435 on Build GRCh38
5455809 c.540T>C p.A180A Chr.9: 93318449-93318449 on Build GRCh38
1199072 c.568C>T p.R190C Chr.9: 93318421-93318421 on Build GRCh38
1199073 c.500G>A p.G167D Chr.9: 93318489-93318489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966066305 Chr.9: 93318275-93318275 on Build GRCh38
rs538747987 Chr.9: 93318511-93318511 on Build GRCh38
rs377181211 Chr.9: 93318475-93318475 on Build GRCh38
rs749770864 Chr.9: 93318562-93318562 on Build GRCh38
rs201784838 Chr.9: 93318447-93318447 on Build GRCh38
rs774762460 Chr.9: 93318571-93318571 on Build GRCh38
rs567980823 Chr.9: 93318446-93318446 on Build GRCh38
rs901635585 Chr.9: 93318152-93318152 on Build GRCh38
rs75416305 Chr.9: 93318422-93318422 on Build GRCh38
rs747820340 Chr.9: 93318475-93318477 on Build GRCh38
rs561982759 Chr.9: 93318249-93318249 on Build GRCh38
rs41274360 Chr.9: 93318355-93318355 on Build GRCh38
rs755682487 Chr.9: 93318338-93318338 on Build GRCh38
rs767257978 Chr.9: 93318294-93318294 on Build GRCh38
rs879532008 Chr.9: 93318486-93318486 on Build GRCh38
rs771789174 Chr.9: 93318442-93318442 on Build GRCh38
rs754358380 Chr.9: 93318157-93318157 on Build GRCh38
rs1009778714 Chr.9: 93318331-93318331 on Build GRCh38
rs369434914 Chr.9: 93318502-93318502 on Build GRCh38
rs767783082 Chr.9: 93318577-93318577 on Build GRCh38
rs777655297 Chr.9: 93318492-93318492 on Build GRCh38
rs1050076066 Chr.9: 93318172-93318172 on Build GRCh38
rs201476852 Chr.9: 93318385-93318385 on Build GRCh38
rs367936667 Chr.9: 93318399-93318399 on Build GRCh38
rs774476176 Chr.9: 93318484-93318484 on Build GRCh38
rs763173625 Chr.9: 93318482-93318482 on Build GRCh38
rs202074953 Chr.9: 93318501-93318501 on Build GRCh38
rs752296557 Chr.9: 93318325-93318325 on Build GRCh38
rs200294546 Chr.9: 93318556-93318556 on Build GRCh38
rs200539322 Chr.9: 93318483-93318483 on Build GRCh38
rs373819150 Chr.9: 93318425-93318425 on Build GRCh38
rs375063816 Chr.9: 93318567-93318567 on Build GRCh38
rs967808203 Chr.9: 93318220-93318220 on Build GRCh38
rs573923553 Chr.9: 93318588-93318588 on Build GRCh38
rs771265875 Chr.9: 93318566-93318566 on Build GRCh38
rs143563466 Chr.9: 93318205-93318205 on Build GRCh38
rs200220940 Chr.9: 93318420-93318420 on Build GRCh38
rs190749975 Chr.9: 93318599-93318599 on Build GRCh38
rs751679011 Chr.9: 93318415-93318415 on Build GRCh38
rs372403641 Chr.9: 93318462-93318462 on Build GRCh38
rs376679853 Chr.9: 93318343-93318343 on Build GRCh38
rs762368880 Chr.9: 93318465-93318465 on Build GRCh38
rs990274961 Chr.9: 93318161-93318161 on Build GRCh38
rs931912191 Chr.9: 93318316-93318316 on Build GRCh38
rs759690207 Chr.9: 93318574-93318574 on Build GRCh38
rs550098324 Chr.9: 93318587-93318587 on Build GRCh38
rs185926577 Chr.9: 93318549-93318549 on Build GRCh38
rs746709350 Chr.9: 93318527-93318527 on Build GRCh38
rs760787782 Chr.9: 93318598-93318598 on Build GRCh38
rs763239658 Chr.9: 93318404-93318404 on Build GRCh38
rs1011810696 Chr.9: 93318426-93318426 on Build GRCh38
rs553818677 Chr.9: 93318512-93318512 on Build GRCh38
rs1002939530 Chr.9: 93318386-93318386 on Build GRCh38
rs115115786 Chr.9: 93318481-93318481 on Build GRCh38
rs2868 Chr.9: 93318302-93318302 on Build GRCh38
rs544222075 Chr.9: 93318271-93318271 on Build GRCh38
rs77860862 Chr.9: 93318201-93318201 on Build GRCh38
rs566343168 Chr.9: 93318537-93318537 on Build GRCh38
rs765659687 Chr.9: 93318390-93318390 on Build GRCh38
rs781389210 Chr.9: 93318418-93318418 on Build GRCh38
rs779872153 Chr.9: 93318519-93318519 on Build GRCh38
rs36081907 Chr.9: 93318417-93318417 on Build GRCh38
rs1034410114 Chr.9: 93318589-93318589 on Build GRCh38
rs533070917 Chr.9: 93318313-93318313 on Build GRCh38
rs771568842 Chr.9: 93318536-93318537 on Build GRCh38
rs766468316 Chr.9: 93318410-93318410 on Build GRCh38
rs776117409 Chr.9: 93318464-93318464 on Build GRCh38
rs933070414 Chr.9: 93318158-93318158 on Build GRCh38
rs745593217 Chr.9: 93318440-93318440 on Build GRCh38
rs142675606 Chr.9: 93318276-93318276 on Build GRCh38
rs757265933 Chr.9: 93318510-93318510 on Build GRCh38
rs148022448 Chr.9: 93318256-93318256 on Build GRCh38

View Full Product Details