Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93446529-93447075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120AOS
Entrez Gene ID
158293
Gene Name
family with sequence similarity 120A opposite strand
Gene Aliases
C9orf10OS
UniGene
Hs.350364
Gene Chromosome Location
Chr.9: 93446500-93453592 on Build GRCh38
Transcripts
NM_001322224.1 NM_198841.2 NR_136229.1 NR_136230.1 NR_136231.1 NR_136232.1 NR_136233.1 NR_136234.1 NR_136235.1 NR_136236.1 NR_136237.1 NR_136238.1

SNP Detail

SNP ID Chromosome Location
rs757019777 Chr.9: 93446685-93446685 on Build GRCh38
rs3898091 Chr.9: 93446915-93446915 on Build GRCh38
rs913408740 Chr.9: 93446916-93446916 on Build GRCh38
rs984953832 Chr.9: 93446798-93446798 on Build GRCh38
rs908969843 Chr.9: 93446810-93446810 on Build GRCh38
rs1033527031 Chr.9: 93446691-93446691 on Build GRCh38
rs142030308 Chr.9: 93447017-93447017 on Build GRCh38
rs980130328 Chr.9: 93446839-93446839 on Build GRCh38
rs549007893 Chr.9: 93446591-93446591 on Build GRCh38
rs768772996 Chr.9: 93446553-93446554 on Build GRCh38
rs957446312 Chr.9: 93446733-93446733 on Build GRCh38
rs989245770 Chr.9: 93446738-93446738 on Build GRCh38
rs1034582884 Chr.9: 93447040-93447040 on Build GRCh38
rs765594646 Chr.9: 93447020-93447020 on Build GRCh38
rs570946888 Chr.9: 93446662-93446662 on Build GRCh38
rs1007676696 Chr.9: 93446759-93446759 on Build GRCh38
rs993283267 Chr.9: 93446565-93446565 on Build GRCh38
rs781771560 Chr.9: 93446654-93446654 on Build GRCh38
rs768339833 Chr.9: 93446841-93446841 on Build GRCh38
rs778606236 Chr.9: 93446730-93446730 on Build GRCh38
rs41274380 Chr.9: 93446977-93446977 on Build GRCh38
rs1038688544 Chr.9: 93447001-93447001 on Build GRCh38
rs1051267897 Chr.9: 93446967-93446967 on Build GRCh38
rs566923427 Chr.9: 93446604-93446604 on Build GRCh38
rs901783460 Chr.9: 93446567-93446567 on Build GRCh38
rs530414172 Chr.9: 93446581-93446581 on Build GRCh38
rs547250481 Chr.9: 93446779-93446779 on Build GRCh38
rs1032161467 Chr.9: 93446974-93446974 on Build GRCh38
rs932938198 Chr.9: 93446864-93446864 on Build GRCh38
rs992668622 Chr.9: 93446813-93446813 on Build GRCh38
rs889010385 Chr.9: 93446886-93446886 on Build GRCh38
rs187624162 Chr.9: 93446633-93446633 on Build GRCh38
rs866283291 Chr.9: 93446745-93446745 on Build GRCh38
rs746513776 Chr.9: 93446822-93446822 on Build GRCh38
rs926219667 Chr.9: 93446653-93446653 on Build GRCh38
rs117128404 Chr.9: 93446878-93446878 on Build GRCh38
rs936258240 Chr.9: 93446860-93446860 on Build GRCh38
rs1026223943 Chr.9: 93447028-93447028 on Build GRCh38
rs536171660 Chr.9: 93446789-93446789 on Build GRCh38
rs532369296 Chr.9: 93446721-93446721 on Build GRCh38
rs1036476761 Chr.9: 93446835-93446835 on Build GRCh38
rs917360578 Chr.9: 93446815-93446815 on Build GRCh38
rs546304557 Chr.9: 93446906-93446907 on Build GRCh38
rs981369053 Chr.9: 93447018-93447018 on Build GRCh38
rs148588599 Chr.9: 93446924-93446924 on Build GRCh38
rs565837266 Chr.9: 93446786-93446786 on Build GRCh38
rs569311860 Chr.9: 93446857-93446857 on Build GRCh38
rs771917816 Chr.9: 93446792-93446792 on Build GRCh38
rs891944726 Chr.9: 93446689-93446689 on Build GRCh38
rs1044400097 Chr.9: 93446645-93446645 on Build GRCh38
rs963292744 Chr.9: 93446780-93446780 on Build GRCh38
rs1001117592 Chr.9: 93446922-93446922 on Build GRCh38
rs1009318981 Chr.9: 93446717-93446717 on Build GRCh38
rs943120771 Chr.9: 93446988-93446988 on Build GRCh38
rs192096876 Chr.9: 93446820-93446820 on Build GRCh38
rs971661886 Chr.9: 93446778-93446778 on Build GRCh38

View Full Product Details