Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93527063-93527561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120A
Entrez Gene ID
23196
Gene Name
family with sequence similarity 120A
Gene Aliases
C9orf10, OSSA
UniGene
Hs.372003
Gene Chromosome Location
Chr.9: 93451696-93566115 on Build GRCh38
Transcripts
NM_001286722.1 NM_001286723.1 NM_001286724.1 NM_014612.4 XM_005251842.4 XM_011518412.2 XM_011518413.2 XM_011518414.2 XM_011518416.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5023943 c.1434C>T p.N478N Chr.9: 93527170-93527170 on Build GRCh38
6292989 c.1498C>A p.Q500K Chr.9: 93527234-93527234 on Build GRCh38
5023944 c.1518C>T p.N506N Chr.9: 93527170-93527170 on Build GRCh38
4825187 c.1466C>G p.S489* Chr.9: 93527202-93527202 on Build GRCh38
2149040 c.1448delA p.E483fs*33 Chr.9: 93527184-93527184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765625977 Chr.9: 93527294-93527294 on Build GRCh38
rs1018165672 Chr.9: 93527451-93527451 on Build GRCh38
rs762138845 Chr.9: 93527287-93527287 on Build GRCh38
rs373899557 Chr.9: 93527148-93527148 on Build GRCh38
rs755996298 Chr.9: 93527193-93527193 on Build GRCh38
rs1003166419 Chr.9: 93527346-93527346 on Build GRCh38
rs923570961 Chr.9: 93527169-93527169 on Build GRCh38
rs376126436 Chr.9: 93527486-93527486 on Build GRCh38
rs924144590 Chr.9: 93527255-93527255 on Build GRCh38
rs367939753 Chr.9: 93527164-93527164 on Build GRCh38
rs780037097 Chr.9: 93527188-93527188 on Build GRCh38
rs368792160 Chr.9: 93527254-93527254 on Build GRCh38
rs977800075 Chr.9: 93527457-93527457 on Build GRCh38
rs1008068763 Chr.9: 93527436-93527436 on Build GRCh38
rs562880065 Chr.9: 93527329-93527329 on Build GRCh38
rs894670824 Chr.9: 93527400-93527400 on Build GRCh38
rs562270148 Chr.9: 93527132-93527132 on Build GRCh38
rs921824495 Chr.9: 93527218-93527218 on Build GRCh38
rs944610634 Chr.9: 93527191-93527191 on Build GRCh38
rs776668721 Chr.9: 93527182-93527184 on Build GRCh38
rs865939632 Chr.9: 93527378-93527378 on Build GRCh38
rs899468528 Chr.9: 93527460-93527460 on Build GRCh38
rs777574383 Chr.9: 93527199-93527199 on Build GRCh38
rs1999386 Chr.9: 93527461-93527461 on Build GRCh38
rs150637769 Chr.9: 93527098-93527098 on Build GRCh38
rs372086188 Chr.9: 93527256-93527256 on Build GRCh38
rs887881820 Chr.9: 93527302-93527302 on Build GRCh38
rs775094705 Chr.9: 93527258-93527258 on Build GRCh38
rs965696724 Chr.9: 93527468-93527468 on Build GRCh38
rs41274402 Chr.9: 93527109-93527109 on Build GRCh38
rs767053814 Chr.9: 93527128-93527128 on Build GRCh38
rs764740745 Chr.9: 93527270-93527270 on Build GRCh38
rs761206974 Chr.9: 93527137-93527137 on Build GRCh38
rs761299039 Chr.9: 93527269-93527269 on Build GRCh38
rs907173500 Chr.9: 93527345-93527345 on Build GRCh38
rs931894823 Chr.9: 93527491-93527491 on Build GRCh38
rs758470662 Chr.9: 93527186-93527186 on Build GRCh38
rs770448026 Chr.9: 93527108-93527108 on Build GRCh38
rs967802230 Chr.9: 93527433-93527433 on Build GRCh38
rs376249435 Chr.9: 93527165-93527165 on Build GRCh38
rs775739842 Chr.9: 93527238-93527238 on Build GRCh38
rs201255629 Chr.9: 93527264-93527264 on Build GRCh38
rs868017442 Chr.9: 93527185-93527193 on Build GRCh38
rs1021776595 Chr.9: 93527410-93527410 on Build GRCh38
rs976056719 Chr.9: 93527477-93527477 on Build GRCh38
rs939602558 Chr.9: 93527278-93527278 on Build GRCh38
rs533317805 Chr.9: 93527337-93527337 on Build GRCh38
rs1049171192 Chr.9: 93527272-93527272 on Build GRCh38
rs199688004 Chr.9: 93527184-93527184 on Build GRCh38
rs777253102 Chr.9: 93527275-93527275 on Build GRCh38
rs368222954 Chr.9: 93527496-93527496 on Build GRCh38
rs1003046217 Chr.9: 93527095-93527095 on Build GRCh38
rs375834512 Chr.9: 93527207-93527207 on Build GRCh38
rs747325410 Chr.9: 93527465-93527465 on Build GRCh38
rs1012139225 Chr.9: 93527409-93527409 on Build GRCh38
rs961174846 Chr.9: 93527517-93527517 on Build GRCh38
rs1036329085 Chr.9: 93527340-93527340 on Build GRCh38
rs977768891 Chr.9: 93527168-93527168 on Build GRCh38
rs1056763879 Chr.9: 93527330-93527330 on Build GRCh38
rs759029698 Chr.9: 93527127-93527127 on Build GRCh38
rs200312346 Chr.9: 93527154-93527154 on Build GRCh38
rs754216270 Chr.9: 93527149-93527149 on Build GRCh38
rs914326043 Chr.9: 93527187-93527187 on Build GRCh38
rs190429030 Chr.9: 93527307-93527307 on Build GRCh38
rs371519996 Chr.9: 93527190-93527190 on Build GRCh38
rs766627817 Chr.9: 93527170-93527170 on Build GRCh38
rs1035005791 Chr.9: 93527389-93527389 on Build GRCh38
rs760987402 Chr.9: 93527143-93527143 on Build GRCh38
rs968587151 Chr.9: 93527217-93527217 on Build GRCh38

View Full Product Details