Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94760370-94760867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf3
Entrez Gene ID
84909
Gene Name
chromosome 9 open reading frame 3
Gene Aliases
AOPEP, AP-O, APO, C90RF3, ONPEP
UniGene
Hs.434253
Gene Chromosome Location
Chr.9: 94726604-95148264 on Build GRCh38
Transcripts
NM_001193329.1 NM_001193331.2 NM_032823.5 XM_005252284.2 XM_006717306.1 XM_011519119.1 XM_011519120.2 XM_011519121.2 XM_011519122.2 XM_011519123.2 XM_011519127.1 XM_011519129.1 XM_011519130.1 XM_011519134.2 XM_017015229.1 XM_017015230.1 XM_017015231.1 XM_017015232.1 XM_017015233.1 XM_017015234.1 XM_017015235.1 XM_017015236.1 XM_017015237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6550257 c.634G>T p.A212S Chr.9: 94760417-94760417 on Build GRCh38
456277 c.757G>A p.E253K Chr.9: 94760540-94760540 on Build GRCh38
4589058 c.696A>G p.T232T Chr.9: 94760479-94760479 on Build GRCh38
6550258 c.103G>T p.A35S Chr.9: 94760417-94760417 on Build GRCh38
6230948 c.389C>T p.S130L Chr.9: 94760550-94760550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554031344 Chr.9: 94760814-94760814 on Build GRCh38
rs903834264 Chr.9: 94760566-94760566 on Build GRCh38
rs202175159 Chr.9: 94760432-94760432 on Build GRCh38
rs761340510 Chr.9: 94760632-94760632 on Build GRCh38
rs779393035 Chr.9: 94760546-94760546 on Build GRCh38
rs373613062 Chr.9: 94760802-94760804 on Build GRCh38
rs1030493256 Chr.9: 94760408-94760408 on Build GRCh38
rs371873852 Chr.9: 94760537-94760537 on Build GRCh38
rs751389139 Chr.9: 94760413-94760413 on Build GRCh38
rs369861586 Chr.9: 94760498-94760498 on Build GRCh38
rs768912234 Chr.9: 94760563-94760563 on Build GRCh38
rs867251931 Chr.9: 94760628-94760628 on Build GRCh38
rs772103194 Chr.9: 94760481-94760481 on Build GRCh38
rs749803059 Chr.9: 94760438-94760438 on Build GRCh38
rs368705058 Chr.9: 94760427-94760427 on Build GRCh38
rs1030972469 Chr.9: 94760612-94760612 on Build GRCh38
rs1045653216 Chr.9: 94760495-94760495 on Build GRCh38
rs962334583 Chr.9: 94760714-94760714 on Build GRCh38
rs375606396 Chr.9: 94760526-94760526 on Build GRCh38
rs750720840 Chr.9: 94760479-94760480 on Build GRCh38
rs976191132 Chr.9: 94760406-94760406 on Build GRCh38
rs1006601161 Chr.9: 94760393-94760393 on Build GRCh38
rs775605570 Chr.9: 94760460-94760460 on Build GRCh38
rs779305268 Chr.9: 94760440-94760440 on Build GRCh38
rs1043405296 Chr.9: 94760560-94760560 on Build GRCh38
rs953626386 Chr.9: 94760665-94760665 on Build GRCh38
rs769632059 Chr.9: 94760531-94760531 on Build GRCh38
rs145414026 Chr.9: 94760550-94760550 on Build GRCh38
rs746240882 Chr.9: 94760444-94760444 on Build GRCh38
rs767297321 Chr.9: 94760511-94760511 on Build GRCh38
rs781216214 Chr.9: 94760575-94760575 on Build GRCh38
rs1055598699 Chr.9: 94760543-94760543 on Build GRCh38
rs759528062 Chr.9: 94760596-94760596 on Build GRCh38
rs1007700273 Chr.9: 94760675-94760675 on Build GRCh38
rs747234731 Chr.9: 94760561-94760561 on Build GRCh38
rs763818285 Chr.9: 94760627-94760627 on Build GRCh38
rs760299708 Chr.9: 94760615-94760615 on Build GRCh38
rs12341814 Chr.9: 94760633-94760633 on Build GRCh38
rs945946832 Chr.9: 94760493-94760495 on Build GRCh38
rs776739942 Chr.9: 94760486-94760486 on Build GRCh38
rs752346164 Chr.9: 94760513-94760513 on Build GRCh38
rs971702121 Chr.9: 94760721-94760721 on Build GRCh38
rs777318703 Chr.9: 94760437-94760437 on Build GRCh38
rs140563732 Chr.9: 94760468-94760468 on Build GRCh38
rs753418720 Chr.9: 94760631-94760631 on Build GRCh38
rs775353831 Chr.9: 94760613-94760613 on Build GRCh38
rs16911681 Chr.9: 94760547-94760547 on Build GRCh38
rs754763664 Chr.9: 94760417-94760417 on Build GRCh38
rs774456692 Chr.9: 94760586-94760586 on Build GRCh38
rs763043975 Chr.9: 94760496-94760496 on Build GRCh38
rs769955454 Chr.9: 94760583-94760583 on Build GRCh38
rs909486196 Chr.9: 94760679-94760679 on Build GRCh38
rs141234450 Chr.9: 94760453-94760453 on Build GRCh38
rs767284344 Chr.9: 94760431-94760431 on Build GRCh38
rs937147057 Chr.9: 94760533-94760533 on Build GRCh38
rs746153185 Chr.9: 94760395-94760395 on Build GRCh38
rs367963020 Chr.9: 94760398-94760398 on Build GRCh38
rs922121046 Chr.9: 94760418-94760418 on Build GRCh38
rs756484922 Chr.9: 94760606-94760607 on Build GRCh38
rs989950393 Chr.9: 94760816-94760816 on Build GRCh38
rs774364750 Chr.9: 94760508-94760508 on Build GRCh38
rs201160070 Chr.9: 94760582-94760582 on Build GRCh38
rs538464704 Chr.9: 94760646-94760646 on Build GRCh38
rs771805367 Chr.9: 94760606-94760606 on Build GRCh38
rs375600932 Chr.9: 94760479-94760479 on Build GRCh38
rs910947056 Chr.9: 94760454-94760454 on Build GRCh38
rs747138673 Chr.9: 94760510-94760510 on Build GRCh38
rs985044878 Chr.9: 94760655-94760655 on Build GRCh38
rs928288600 Chr.9: 94760727-94760727 on Build GRCh38
rs778154453 Chr.9: 94760651-94760651 on Build GRCh38
rs41281172 Chr.9: 94760434-94760434 on Build GRCh38
rs76131437 Chr.9: 94760401-94760401 on Build GRCh38
rs768081427 Chr.9: 94760399-94760400 on Build GRCh38
rs1029081021 Chr.9: 94760635-94760635 on Build GRCh38
rs780441873 Chr.9: 94760557-94760557 on Build GRCh38
rs375030149 Chr.9: 94760625-94760625 on Build GRCh38
rs773044361 Chr.9: 94760403-94760403 on Build GRCh38
rs756695675 Chr.9: 94760542-94760542 on Build GRCh38

View Full Product Details