Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94928344-94928846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf3
Entrez Gene ID
84909
Gene Name
chromosome 9 open reading frame 3
Gene Aliases
AOPEP, AP-O, APO, C90RF3, ONPEP
UniGene
Hs.434253
Gene Chromosome Location
Chr.9: 94726604-95148264 on Build GRCh38
Transcripts
NM_001193329.1 NM_001193331.2 NM_032823.5 XM_005252284.2 XM_006717306.1 XM_011519119.1 XM_011519120.2 XM_011519121.2 XM_011519122.2 XM_011519123.2 XM_011519127.1 XM_011519129.1 XM_011519132.1 XM_017015229.1 XM_017015230.1 XM_017015231.1 XM_017015232.1 XM_017015233.1 XM_017015234.1 XM_017015235.1 XM_017015236.1 XM_017015237.1

Gene Symbol
LOC101928119
Entrez Gene ID
101928119
Gene Name
uncharacterized LOC101928119
Gene Aliases
-
UniGene
Hs.630906
Gene Chromosome Location
Chr.9: 94920339-94937841 on Build GRCh38
Transcripts
NM_001193329.1 NM_001193331.2 NM_032823.5 XM_005252284.2 XM_006717306.1 XM_011519119.1 XM_011519120.2 XM_011519121.2 XM_011519122.2 XM_011519123.2 XM_011519127.1 XM_011519129.1 XM_011519132.1 XM_017015229.1 XM_017015230.1 XM_017015231.1 XM_017015232.1 XM_017015233.1 XM_017015234.1 XM_017015235.1 XM_017015236.1 XM_017015237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6298938 c.1587G>T p.E529D Chr.9: 94928457-94928457 on Build GRCh38
4855677 c.1624G>A p.E542K Chr.9: 94928494-94928494 on Build GRCh38
6298937 c.1587G>T p.E529D Chr.9: 94928457-94928457 on Build GRCh38
4468951 c.1572C>T p.A524A Chr.9: 94928442-94928442 on Build GRCh38
7251291 c.1603C>T p.R535C Chr.9: 94928473-94928473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042781673 Chr.9: 94928774-94928774 on Build GRCh38
rs549129630 Chr.9: 94928493-94928493 on Build GRCh38
rs765776872 Chr.9: 94928474-94928474 on Build GRCh38
rs527504955 Chr.9: 94928663-94928663 on Build GRCh38
rs896182260 Chr.9: 94928482-94928482 on Build GRCh38
rs187589689 Chr.9: 94928766-94928766 on Build GRCh38
rs975798583 Chr.9: 94928566-94928566 on Build GRCh38
rs894162990 Chr.9: 94928437-94928437 on Build GRCh38
rs199727977 Chr.9: 94928439-94928439 on Build GRCh38
rs999008130 Chr.9: 94928473-94928473 on Build GRCh38
rs555954977 Chr.9: 94928810-94928810 on Build GRCh38
rs147369569 Chr.9: 94928513-94928513 on Build GRCh38
rs566940178 Chr.9: 94928782-94928782 on Build GRCh38
rs938832630 Chr.9: 94928764-94928764 on Build GRCh38
rs962158033 Chr.9: 94928542-94928542 on Build GRCh38
rs928830077 Chr.9: 94928672-94928672 on Build GRCh38
rs972525761 Chr.9: 94928602-94928602 on Build GRCh38
rs544966281 Chr.9: 94928827-94928827 on Build GRCh38
rs114022900 Chr.9: 94928715-94928715 on Build GRCh38
rs914865018 Chr.9: 94928698-94928698 on Build GRCh38
rs1028221753 Chr.9: 94928498-94928498 on Build GRCh38
rs142764274 Chr.9: 94928451-94928451 on Build GRCh38
rs569276103 Chr.9: 94928765-94928765 on Build GRCh38
rs928758938 Chr.9: 94928385-94928385 on Build GRCh38
rs1036573869 Chr.9: 94928373-94928373 on Build GRCh38
rs12338409 Chr.9: 94928807-94928807 on Build GRCh38
rs140120435 Chr.9: 94928699-94928699 on Build GRCh38
rs964068611 Chr.9: 94928656-94928656 on Build GRCh38
rs923686212 Chr.9: 94928443-94928443 on Build GRCh38
rs545129535 Chr.9: 94928511-94928511 on Build GRCh38
rs138602742 Chr.9: 94928411-94928411 on Build GRCh38
rs141943676 Chr.9: 94928716-94928716 on Build GRCh38
rs751025719 Chr.9: 94928468-94928468 on Build GRCh38
rs1056026703 Chr.9: 94928435-94928435 on Build GRCh38
rs12344211 Chr.9: 94928822-94928822 on Build GRCh38
rs544234232 Chr.9: 94928369-94928369 on Build GRCh38
rs767085215 Chr.9: 94928704-94928704 on Build GRCh38
rs750854535 Chr.9: 94928502-94928502 on Build GRCh38
rs975402219 Chr.9: 94928679-94928679 on Build GRCh38
rs548732943 Chr.9: 94928681-94928681 on Build GRCh38
rs975684800 Chr.9: 94928395-94928396 on Build GRCh38
rs567355121 Chr.9: 94928658-94928658 on Build GRCh38
rs41281176 Chr.9: 94928480-94928480 on Build GRCh38
rs770527092 Chr.9: 94928460-94928460 on Build GRCh38
rs1042730198 Chr.9: 94928444-94928444 on Build GRCh38
rs756437433 Chr.9: 94928519-94928519 on Build GRCh38
rs898416175 Chr.9: 94928512-94928512 on Build GRCh38
rs1019446111 Chr.9: 94928479-94928479 on Build GRCh38
rs1041168640 Chr.9: 94928812-94928812 on Build GRCh38
rs773155661 Chr.9: 94928383-94928383 on Build GRCh38
rs934440664 Chr.9: 94928808-94928808 on Build GRCh38
rs558257621 Chr.9: 94928781-94928781 on Build GRCh38
rs575942115 Chr.9: 94928526-94928526 on Build GRCh38
rs1025442450 Chr.9: 94928535-94928535 on Build GRCh38
rs1008288267 Chr.9: 94928597-94928597 on Build GRCh38

View Full Product Details