Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 95920700-95921248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC6L2
Entrez Gene ID
375748
Gene Name
ERCC excision repair 6 like 2
Gene Aliases
BMFS2, C9orf102, RAD26L, SR278
UniGene
Hs.432364
Gene Chromosome Location
Chr.9: 95875618-96041092 on Build GRCh38
Transcripts
NM_001010895.2 NM_020207.4 XM_011518641.2 XM_011518644.2 XM_011518645.2 XM_011518646.2 XM_011518647.1 XM_011518650.2 XM_017014707.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1111564 c.1255G>A p.D419N Chr.9: 95921238-95921238 on Build GRCh38
3909256 c.1198T>C p.Y400H Chr.9: 95921181-95921181 on Build GRCh38
5990745 c.694A>G p.T232A Chr.9: 95921244-95921244 on Build GRCh38
3395943 c.1246G>C p.E416Q Chr.9: 95921229-95921229 on Build GRCh38
3909257 c.631T>C p.Y211H Chr.9: 95921181-95921181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761223800 Chr.9: 95921174-95921174 on Build GRCh38
rs908724958 Chr.9: 95920973-95920973 on Build GRCh38
rs564726445 Chr.9: 95920984-95920984 on Build GRCh38
rs923011622 Chr.9: 95921117-95921117 on Build GRCh38
rs766145218 Chr.9: 95921215-95921215 on Build GRCh38
rs938078498 Chr.9: 95921185-95921185 on Build GRCh38
rs961556056 Chr.9: 95921092-95921092 on Build GRCh38
rs984408680 Chr.9: 95921083-95921083 on Build GRCh38
rs545366686 Chr.9: 95920803-95920803 on Build GRCh38
rs1046458449 Chr.9: 95920983-95920983 on Build GRCh38
rs751512066 Chr.9: 95921220-95921220 on Build GRCh38
rs769990735 Chr.9: 95921144-95921145 on Build GRCh38
rs940129550 Chr.9: 95920815-95920815 on Build GRCh38
rs112389297 Chr.9: 95920864-95920864 on Build GRCh38
rs771303268 Chr.9: 95921128-95921128 on Build GRCh38
rs975777975 Chr.9: 95921104-95921104 on Build GRCh38
rs368981276 Chr.9: 95921133-95921133 on Build GRCh38
rs182964911 Chr.9: 95920856-95920856 on Build GRCh38
rs117728766 Chr.9: 95921028-95921028 on Build GRCh38
rs577917388 Chr.9: 95921160-95921160 on Build GRCh38
rs150111606 Chr.9: 95920977-95920977 on Build GRCh38
rs1048585485 Chr.9: 95920807-95920807 on Build GRCh38
rs142685145 Chr.9: 95921211-95921211 on Build GRCh38
rs186123753 Chr.9: 95921066-95921066 on Build GRCh38
rs1001707851 Chr.9: 95921058-95921058 on Build GRCh38
rs966827457 Chr.9: 95920886-95920886 on Build GRCh38
rs536005781 Chr.9: 95921038-95921038 on Build GRCh38
rs982722982 Chr.9: 95921120-95921120 on Build GRCh38
rs984663972 Chr.9: 95920957-95920957 on Build GRCh38
rs1034600732 Chr.9: 95920863-95920863 on Build GRCh38
rs886037662 Chr.9: 95921219-95921222 on Build GRCh38
rs990785109 Chr.9: 95921222-95921222 on Build GRCh38
rs1010289762 Chr.9: 95920805-95920805 on Build GRCh38
rs768120288 Chr.9: 95921175-95921175 on Build GRCh38
rs547963748 Chr.9: 95921054-95921054 on Build GRCh38
rs73534661 Chr.9: 95921089-95921089 on Build GRCh38
rs1009168797 Chr.9: 95921049-95921049 on Build GRCh38
rs543263443 Chr.9: 95920922-95920922 on Build GRCh38
rs893687708 Chr.9: 95921003-95921003 on Build GRCh38
rs975849826 Chr.9: 95920958-95920958 on Build GRCh38
rs891814723 Chr.9: 95920996-95920996 on Build GRCh38
rs1055034781 Chr.9: 95920859-95920859 on Build GRCh38
rs761403334 Chr.9: 95921176-95921176 on Build GRCh38
rs557515996 Chr.9: 95920816-95920816 on Build GRCh38
rs373230343 Chr.9: 95921136-95921136 on Build GRCh38
rs750116560 Chr.9: 95921184-95921184 on Build GRCh38
rs776934402 Chr.9: 95920949-95920949 on Build GRCh38
rs952938951 Chr.9: 95921071-95921071 on Build GRCh38
rs764954422 Chr.9: 95921179-95921179 on Build GRCh38
rs1010835795 Chr.9: 95921017-95921017 on Build GRCh38
rs1034933790 Chr.9: 95921064-95921064 on Build GRCh38
rs891847257 Chr.9: 95920784-95920784 on Build GRCh38
rs35961465 Chr.9: 95921130-95921130 on Build GRCh38
rs12685318 Chr.9: 95921018-95921018 on Build GRCh38
rs1008659045 Chr.9: 95920895-95920895 on Build GRCh38
rs560930516 Chr.9: 95920924-95920924 on Build GRCh38
rs539004509 Chr.9: 95920793-95920793 on Build GRCh38
rs906553276 Chr.9: 95920831-95920831 on Build GRCh38
rs772387539 Chr.9: 95921138-95921138 on Build GRCh38
rs908583824 Chr.9: 95920808-95920808 on Build GRCh38
rs1020788583 Chr.9: 95920879-95920879 on Build GRCh38
rs762785173 Chr.9: 95920865-95920865 on Build GRCh38
rs1055168144 Chr.9: 95921000-95921000 on Build GRCh38
rs532479344 Chr.9: 95920993-95920993 on Build GRCh38
rs762803546 Chr.9: 95921213-95921213 on Build GRCh38
rs1033427375 Chr.9: 95920781-95920781 on Build GRCh38
rs766129892 Chr.9: 95920923-95920923 on Build GRCh38
rs2018478 Chr.9: 95921070-95921070 on Build GRCh38
rs10988581 Chr.9: 95920934-95920934 on Build GRCh38
rs573576839 Chr.9: 95921055-95921055 on Build GRCh38

View Full Product Details