Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96500659-96501171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC14B
Entrez Gene ID
8555
Gene Name
cell division cycle 14B
Gene Aliases
CDC14B3, Cdc14B1, Cdc14B2, hCDC14B
UniGene
Hs.40582
Gene Chromosome Location
Chr.9: 96492738-96619830 on Build GRCh38
Transcripts
NM_001077181.1 NM_003671.3 NM_033331.2 XM_011519147.2 XM_011519148.2 XM_011519149.2 XM_011519152.2 XM_011519153.2 XM_011519156.2 XM_011519158.1 XM_011519159.2 XM_017015240.1 XM_017015241.1 XM_017015242.1 XM_017015244.1 XM_017015247.1 XM_017015248.1

SNP Detail

SNP ID Chromosome Location
rs553037526 Chr.9: 96500986-96500986 on Build GRCh38
rs931395430 Chr.9: 96501121-96501121 on Build GRCh38
rs570653039 Chr.9: 96501023-96501023 on Build GRCh38
rs1021740282 Chr.9: 96501046-96501046 on Build GRCh38
rs368906646 Chr.9: 96501061-96501061 on Build GRCh38
rs959708089 Chr.9: 96501078-96501078 on Build GRCh38
rs992729257 Chr.9: 96500867-96500867 on Build GRCh38
rs188366564 Chr.9: 96501057-96501057 on Build GRCh38
rs112781850 Chr.9: 96501044-96501044 on Build GRCh38
rs931380604 Chr.9: 96500963-96500963 on Build GRCh38
rs959956008 Chr.9: 96500822-96500822 on Build GRCh38
rs968427271 Chr.9: 96500787-96500787 on Build GRCh38
rs761828344 Chr.9: 96500821-96500822 on Build GRCh38
rs60778979 Chr.9: 96500906-96500907 on Build GRCh38
rs373640645 Chr.9: 96500902-96500902 on Build GRCh38
rs1020211935 Chr.9: 96500741-96500742 on Build GRCh38
rs866531309 Chr.9: 96500752-96500752 on Build GRCh38
rs570309387 Chr.9: 96500724-96500724 on Build GRCh38
rs192728512 Chr.9: 96500689-96500689 on Build GRCh38
rs1008443167 Chr.9: 96500735-96500735 on Build GRCh38
rs1041277340 Chr.9: 96501038-96501038 on Build GRCh38
rs765396998 Chr.9: 96500984-96500984 on Build GRCh38
rs1031311850 Chr.9: 96501071-96501071 on Build GRCh38
rs920388870 Chr.9: 96501120-96501120 on Build GRCh38
rs532277405 Chr.9: 96500809-96500809 on Build GRCh38
rs1047098947 Chr.9: 96500981-96500981 on Build GRCh38
rs1021398785 Chr.9: 96500784-96500784 on Build GRCh38
rs752750808 Chr.9: 96501060-96501060 on Build GRCh38
rs534703451 Chr.9: 96500921-96500921 on Build GRCh38
rs749680874 Chr.9: 96500891-96500892 on Build GRCh38
rs973630243 Chr.9: 96500914-96500914 on Build GRCh38
rs777481163 Chr.9: 96501095-96501095 on Build GRCh38
rs977091020 Chr.9: 96500794-96500794 on Build GRCh38
rs997019074 Chr.9: 96500766-96500766 on Build GRCh38
rs149993411 Chr.9: 96500879-96500879 on Build GRCh38
rs146755174 Chr.9: 96501079-96501079 on Build GRCh38
rs75622124 Chr.9: 96501143-96501143 on Build GRCh38
rs755877056 Chr.9: 96501091-96501091 on Build GRCh38
rs868535546 Chr.9: 96500775-96500775 on Build GRCh38
rs145099618 Chr.9: 96500890-96500891 on Build GRCh38
rs944302023 Chr.9: 96501026-96501026 on Build GRCh38
rs185045930 Chr.9: 96500873-96500873 on Build GRCh38
rs900065246 Chr.9: 96501043-96501044 on Build GRCh38
rs754843158 Chr.9: 96500744-96500744 on Build GRCh38
rs553885008 Chr.9: 96500903-96500903 on Build GRCh38
rs920040073 Chr.9: 96500952-96500952 on Build GRCh38

View Full Product Details