Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96939807-96940310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUTM2G
Entrez Gene ID
441457
Gene Name
NUT family member 2G
Gene Aliases
FAM22G, NUTMG
UniGene
Hs.652383
Gene Chromosome Location
Chr.9: 96929004-96942290 on Build GRCh38
Transcripts
NM_001045477.2

SNP Detail

SNP ID Chromosome Location
rs893877540 Chr.9: 96940092-96940092 on Build GRCh38
rs867486447 Chr.9: 96940088-96940088 on Build GRCh38
rs933600485 Chr.9: 96939889-96939889 on Build GRCh38
rs560685788 Chr.9: 96940249-96940249 on Build GRCh38
rs868513064 Chr.9: 96940029-96940029 on Build GRCh38
rs913348 Chr.9: 96939830-96939830 on Build GRCh38
rs758724456 Chr.9: 96939880-96939880 on Build GRCh38
rs4641179 Chr.9: 96940184-96940184 on Build GRCh38
rs201932259 Chr.9: 96940290-96940291 on Build GRCh38
rs758222114 Chr.9: 96940079-96940079 on Build GRCh38
rs527764140 Chr.9: 96940285-96940285 on Build GRCh38
rs943361814 Chr.9: 96940030-96940030 on Build GRCh38
rs752916292 Chr.9: 96939863-96939863 on Build GRCh38
rs889125170 Chr.9: 96939983-96939983 on Build GRCh38
rs577628502 Chr.9: 96940224-96940224 on Build GRCh38
rs879842503 Chr.9: 96940181-96940183 on Build GRCh38
rs754166040 Chr.9: 96939898-96939899 on Build GRCh38
rs1052412335 Chr.9: 96939911-96939911 on Build GRCh38
rs1020760920 Chr.9: 96940189-96940189 on Build GRCh38
rs1053830032 Chr.9: 96940089-96940089 on Build GRCh38
rs1009377414 Chr.9: 96940164-96940164 on Build GRCh38
rs754304815 Chr.9: 96940009-96940010 on Build GRCh38
rs867076693 Chr.9: 96939965-96939965 on Build GRCh38
rs867714094 Chr.9: 96940011-96940011 on Build GRCh38
rs573276117 Chr.9: 96940225-96940225 on Build GRCh38

View Full Product Details