Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96940368-96940872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUTM2G
Entrez Gene ID
441457
Gene Name
NUT family member 2G
Gene Aliases
FAM22G, NUTMG
UniGene
Hs.652383
Gene Chromosome Location
Chr.9: 96929004-96942290 on Build GRCh38
Transcripts
NM_001045477.2

SNP Detail

SNP ID Chromosome Location
rs547055406 Chr.9: 96940476-96940476 on Build GRCh38
rs1005215186 Chr.9: 96940419-96940419 on Build GRCh38
rs1055086337 Chr.9: 96940835-96940835 on Build GRCh38
rs369418895 Chr.9: 96940465-96940465 on Build GRCh38
rs1810789 Chr.9: 96940594-96940594 on Build GRCh38
rs943400240 Chr.9: 96940675-96940675 on Build GRCh38
rs1042421422 Chr.9: 96940771-96940771 on Build GRCh38
rs987779742 Chr.9: 96940585-96940585 on Build GRCh38
rs1003787018 Chr.9: 96940815-96940815 on Build GRCh38
rs185203996 Chr.9: 96940639-96940639 on Build GRCh38
rs910786379 Chr.9: 96940651-96940651 on Build GRCh38
rs945475767 Chr.9: 96940740-96940740 on Build GRCh38
rs556482903 Chr.9: 96940735-96940735 on Build GRCh38
rs545415274 Chr.9: 96940791-96940791 on Build GRCh38
rs915217000 Chr.9: 96940719-96940719 on Build GRCh38
rs41306453 Chr.9: 96940490-96940490 on Build GRCh38
rs553866988 Chr.9: 96940817-96940817 on Build GRCh38
rs3895381 Chr.9: 96940689-96940689 on Build GRCh38
rs866497593 Chr.9: 96940666-96940666 on Build GRCh38
rs919440542 Chr.9: 96940555-96940555 on Build GRCh38
rs1053694286 Chr.9: 96940703-96940703 on Build GRCh38
rs1016722427 Chr.9: 96940431-96940431 on Build GRCh38
rs972454345 Chr.9: 96940483-96940483 on Build GRCh38
rs567520559 Chr.9: 96940619-96940619 on Build GRCh38
rs959111410 Chr.9: 96940397-96940397 on Build GRCh38
rs906370315 Chr.9: 96940794-96940794 on Build GRCh38
rs532190235 Chr.9: 96940403-96940403 on Build GRCh38
rs955003465 Chr.9: 96940580-96940580 on Build GRCh38
rs895042960 Chr.9: 96940847-96940847 on Build GRCh38
rs189761646 Chr.9: 96940853-96940853 on Build GRCh38
rs554618043 Chr.9: 96940544-96940544 on Build GRCh38
rs535978169 Chr.9: 96940537-96940537 on Build GRCh38
rs577916254 Chr.9: 96940767-96940767 on Build GRCh38
rs540694208 Chr.9: 96940473-96940474 on Build GRCh38

View Full Product Details