Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97031336-97031840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSV
Entrez Gene ID
1515
Gene Name
cathepsin V
Gene Aliases
CATL2, CTSL2, CTSU
UniGene
Hs.610096
Gene Chromosome Location
Chr.9: 97029677-97039643 on Build GRCh38
Transcripts
NM_001201575.1 NM_001333.3

SNP Detail

SNP ID Chromosome Location
rs1003999298 Chr.9: 97031743-97031743 on Build GRCh38
rs968159394 Chr.9: 97031511-97031511 on Build GRCh38
rs188072300 Chr.9: 97031599-97031599 on Build GRCh38
rs1053974258 Chr.9: 97031585-97031585 on Build GRCh38
rs970772725 Chr.9: 97031763-97031763 on Build GRCh38
rs899761772 Chr.9: 97031708-97031708 on Build GRCh38
rs952579908 Chr.9: 97031738-97031738 on Build GRCh38
rs762033250 Chr.9: 97031600-97031600 on Build GRCh38
rs970741502 Chr.9: 97031524-97031524 on Build GRCh38
rs926591255 Chr.9: 97031785-97031785 on Build GRCh38
rs982794753 Chr.9: 97031507-97031507 on Build GRCh38
rs547814752 Chr.9: 97031482-97031482 on Build GRCh38
rs931092910 Chr.9: 97031635-97031635 on Build GRCh38
rs886982769 Chr.9: 97031652-97031652 on Build GRCh38
rs776816271 Chr.9: 97031547-97031547 on Build GRCh38
rs367544636 Chr.9: 97031688-97031688 on Build GRCh38
rs35529217 Chr.9: 97031432-97031432 on Build GRCh38
rs537751775 Chr.9: 97031777-97031777 on Build GRCh38
rs1026823189 Chr.9: 97031735-97031735 on Build GRCh38
rs190993337 Chr.9: 97031473-97031473 on Build GRCh38
rs147635536 Chr.9: 97031793-97031793 on Build GRCh38
rs373786189 Chr.9: 97031492-97031492 on Build GRCh38
rs774203734 Chr.9: 97031373-97031373 on Build GRCh38
rs57488920 Chr.9: 97031666-97031666 on Build GRCh38
rs1027205318 Chr.9: 97031483-97031483 on Build GRCh38
rs1054347537 Chr.9: 97031369-97031369 on Build GRCh38
rs902818861 Chr.9: 97031410-97031410 on Build GRCh38
rs541141229 Chr.9: 97031370-97031370 on Build GRCh38
rs528043145 Chr.9: 97031545-97031545 on Build GRCh38
rs574706304 Chr.9: 97031396-97031396 on Build GRCh38
rs562856590 Chr.9: 97031610-97031610 on Build GRCh38
rs187353351 Chr.9: 97031460-97031460 on Build GRCh38
rs1007919735 Chr.9: 97031646-97031646 on Build GRCh38
rs371516339 Chr.9: 97031395-97031395 on Build GRCh38
rs899486284 Chr.9: 97031728-97031728 on Build GRCh38
rs144664700 Chr.9: 97031745-97031745 on Build GRCh38
rs952818116 Chr.9: 97031493-97031493 on Build GRCh38
rs530265163 Chr.9: 97031620-97031620 on Build GRCh38
rs567796877 Chr.9: 97031606-97031606 on Build GRCh38
rs1008381551 Chr.9: 97031656-97031656 on Build GRCh38
rs908624963 Chr.9: 97031523-97031523 on Build GRCh38
rs770922532 Chr.9: 97031364-97031364 on Build GRCh38
rs182753586 Chr.9: 97031542-97031542 on Build GRCh38
rs1015655241 Chr.9: 97031752-97031752 on Build GRCh38
rs1056581101 Chr.9: 97031561-97031561 on Build GRCh38
rs898421250 Chr.9: 97031633-97031633 on Build GRCh38
rs963710838 Chr.9: 97031470-97031470 on Build GRCh38
rs765516399 Chr.9: 97031726-97031726 on Build GRCh38
rs892903312 Chr.9: 97031371-97031371 on Build GRCh38

View Full Product Details