Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97032612-97033107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSV
Entrez Gene ID
1515
Gene Name
cathepsin V
Gene Aliases
CATL2, CTSL2, CTSU
UniGene
Hs.610096
Gene Chromosome Location
Chr.9: 97029677-97039643 on Build GRCh38
Transcripts
NM_001201575.1 NM_001333.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7079258 c.906C>A p.S302R Chr.9: 97033048-97033048 on Build GRCh38
1111658 c.927G>A p.S309S Chr.9: 97033027-97033027 on Build GRCh38
1111657 c.967T>C p.C323R Chr.9: 97032987-97032987 on Build GRCh38
6184319 c.978C>T p.A326A Chr.9: 97032976-97032976 on Build GRCh38
6325269 c.911G>A p.G304D Chr.9: 97033043-97033043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199850483 Chr.9: 97032931-97032931 on Build GRCh38
rs908416471 Chr.9: 97032676-97032676 on Build GRCh38
rs573259723 Chr.9: 97032884-97032884 on Build GRCh38
rs754024516 Chr.9: 97032979-97032979 on Build GRCh38
rs762798163 Chr.9: 97032950-97032950 on Build GRCh38
rs1019636869 Chr.9: 97032815-97032815 on Build GRCh38
rs780635985 Chr.9: 97033006-97033006 on Build GRCh38
rs939206375 Chr.9: 97032664-97032664 on Build GRCh38
rs996760664 Chr.9: 97032868-97032868 on Build GRCh38
rs769983150 Chr.9: 97033017-97033017 on Build GRCh38
rs755085359 Chr.9: 97032817-97032817 on Build GRCh38
rs754510537 Chr.9: 97033018-97033018 on Build GRCh38
rs539237474 Chr.9: 97032790-97032790 on Build GRCh38
rs905339408 Chr.9: 97032939-97032939 on Build GRCh38
rs149018053 Chr.9: 97032976-97032976 on Build GRCh38
rs561351246 Chr.9: 97032875-97032875 on Build GRCh38
rs780898535 Chr.9: 97033019-97033019 on Build GRCh38
rs932504048 Chr.9: 97032799-97032799 on Build GRCh38
rs764260070 Chr.9: 97032978-97032978 on Build GRCh38
rs759509224 Chr.9: 97033084-97033084 on Build GRCh38
rs138007477 Chr.9: 97032908-97032908 on Build GRCh38
rs371162982 Chr.9: 97032925-97032925 on Build GRCh38
rs377544223 Chr.9: 97032912-97032912 on Build GRCh38
rs1010974837 Chr.9: 97032746-97032746 on Build GRCh38
rs376239775 Chr.9: 97032958-97032958 on Build GRCh38
rs192537216 Chr.9: 97032686-97032686 on Build GRCh38
rs796609075 Chr.9: 97032958-97032958 on Build GRCh38
rs573045175 Chr.9: 97032835-97032835 on Build GRCh38
rs375508007 Chr.9: 97032809-97032809 on Build GRCh38
rs1013834256 Chr.9: 97032845-97032845 on Build GRCh38
rs376637491 Chr.9: 97032890-97032890 on Build GRCh38
rs11452112 Chr.9: 97032639-97032640 on Build GRCh38
rs147989421 Chr.9: 97032998-97032998 on Build GRCh38
rs200849576 Chr.9: 97033036-97033036 on Build GRCh38
rs775489939 Chr.9: 97032959-97032959 on Build GRCh38
rs373324162 Chr.9: 97032955-97032955 on Build GRCh38
rs976665619 Chr.9: 97032689-97032689 on Build GRCh38
rs1015588622 Chr.9: 97033007-97033007 on Build GRCh38
rs765503098 Chr.9: 97032988-97032988 on Build GRCh38
rs1052782636 Chr.9: 97032812-97032812 on Build GRCh38
rs397819831 Chr.9: 97032646-97032647 on Build GRCh38
rs15394 Chr.9: 97032843-97032843 on Build GRCh38
rs774302568 Chr.9: 97033055-97033055 on Build GRCh38
rs770668641 Chr.9: 97032663-97032663 on Build GRCh38
rs374126168 Chr.9: 97033027-97033027 on Build GRCh38
rs146846413 Chr.9: 97032956-97032956 on Build GRCh38
rs1029136869 Chr.9: 97032876-97032876 on Build GRCh38
rs140679970 Chr.9: 97033008-97033008 on Build GRCh38
rs895234734 Chr.9: 97032731-97032731 on Build GRCh38
rs770723846 Chr.9: 97033043-97033043 on Build GRCh38
rs1042758508 Chr.9: 97032800-97032800 on Build GRCh38
rs370604384 Chr.9: 97032996-97032996 on Build GRCh38
rs558069650 Chr.9: 97032805-97032805 on Build GRCh38
rs749119575 Chr.9: 97033037-97033037 on Build GRCh38
rs73548362 Chr.9: 97032762-97032762 on Build GRCh38
rs1036942975 Chr.9: 97032655-97032655 on Build GRCh38

View Full Product Details