Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97034647-97035158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSV
Entrez Gene ID
1515
Gene Name
cathepsin V
Gene Aliases
CATL2, CTSL2, CTSU
UniGene
Hs.610096
Gene Chromosome Location
Chr.9: 97029677-97039643 on Build GRCh38
Transcripts
NM_001201575.1 NM_001333.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6324005 c.881G>A p.S294N Chr.9: 97034750-97034750 on Build GRCh38
6550275 c.895G>A p.V299I Chr.9: 97034736-97034736 on Build GRCh38
1111660 c.853G>A p.G285S Chr.9: 97034778-97034778 on Build GRCh38
3721373 c.799G>C p.E267Q Chr.9: 97034832-97034832 on Build GRCh38
1111659 c.872C>T p.S291L Chr.9: 97034759-97034759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756835807 Chr.9: 97034885-97034885 on Build GRCh38
rs753507628 Chr.9: 97034793-97034793 on Build GRCh38
rs111792382 Chr.9: 97034779-97034779 on Build GRCh38
rs10980228 Chr.9: 97034852-97034852 on Build GRCh38
rs761976907 Chr.9: 97034755-97034755 on Build GRCh38
rs1055736311 Chr.9: 97035038-97035038 on Build GRCh38
rs761131065 Chr.9: 97034876-97034876 on Build GRCh38
rs997262964 Chr.9: 97035110-97035110 on Build GRCh38
rs111603525 Chr.9: 97034728-97034728 on Build GRCh38
rs766836951 Chr.9: 97034761-97034761 on Build GRCh38
rs377238256 Chr.9: 97034882-97034882 on Build GRCh38
rs1052111851 Chr.9: 97034890-97034890 on Build GRCh38
rs755600659 Chr.9: 97034705-97034705 on Build GRCh38
rs796284380 Chr.9: 97034990-97034990 on Build GRCh38
rs774924658 Chr.9: 97034762-97034762 on Build GRCh38
rs145877427 Chr.9: 97034758-97034758 on Build GRCh38
rs538608725 Chr.9: 97035089-97035089 on Build GRCh38
rs888810729 Chr.9: 97035123-97035123 on Build GRCh38
rs1029947642 Chr.9: 97035112-97035112 on Build GRCh38
rs372591601 Chr.9: 97034918-97034918 on Build GRCh38
rs770728451 Chr.9: 97034735-97034735 on Build GRCh38
rs763552087 Chr.9: 97034784-97034784 on Build GRCh38
rs745664930 Chr.9: 97034820-97034820 on Build GRCh38
rs746963041 Chr.9: 97034837-97034837 on Build GRCh38
rs981438379 Chr.9: 97034926-97034926 on Build GRCh38
rs750707622 Chr.9: 97034678-97034678 on Build GRCh38
rs371321461 Chr.9: 97035080-97035081 on Build GRCh38
rs761934241 Chr.9: 97034672-97034672 on Build GRCh38
rs778724315 Chr.9: 97034736-97034736 on Build GRCh38
rs776623784 Chr.9: 97034751-97034751 on Build GRCh38
rs551605839 Chr.9: 97034778-97034778 on Build GRCh38
rs778671195 Chr.9: 97034813-97034813 on Build GRCh38
rs1026692659 Chr.9: 97034746-97034746 on Build GRCh38
rs571168751 Chr.9: 97035053-97035053 on Build GRCh38
rs763410496 Chr.9: 97034682-97034682 on Build GRCh38
rs1016328682 Chr.9: 97034703-97034703 on Build GRCh38
rs897577894 Chr.9: 97035098-97035098 on Build GRCh38
rs200351452 Chr.9: 97034884-97034884 on Build GRCh38
rs756913407 Chr.9: 97034797-97034797 on Build GRCh38
rs1052156686 Chr.9: 97034924-97034924 on Build GRCh38
rs941884995 Chr.9: 97035054-97035054 on Build GRCh38
rs766925551 Chr.9: 97034866-97034866 on Build GRCh38
rs763138553 Chr.9: 97034760-97034760 on Build GRCh38
rs776030369 Chr.9: 97034873-97034873 on Build GRCh38
rs541142550 Chr.9: 97034862-97034862 on Build GRCh38
rs892092677 Chr.9: 97034896-97034896 on Build GRCh38
rs772062045 Chr.9: 97034737-97034737 on Build GRCh38
rs750180388 Chr.9: 97034892-97034892 on Build GRCh38
rs749956105 Chr.9: 97034877-97034877 on Build GRCh38
rs771264161 Chr.9: 97034759-97034759 on Build GRCh38
rs758285243 Chr.9: 97034823-97034823 on Build GRCh38
rs190631856 Chr.9: 97035014-97035014 on Build GRCh38
rs948889389 Chr.9: 97034830-97034830 on Build GRCh38
rs887583019 Chr.9: 97035049-97035049 on Build GRCh38
rs769898872 Chr.9: 97034844-97034844 on Build GRCh38
rs771286878 Chr.9: 97034850-97034850 on Build GRCh38
rs567267114 Chr.9: 97034905-97034905 on Build GRCh38
rs138696108 Chr.9: 97034768-97034768 on Build GRCh38
rs755661945 Chr.9: 97034724-97034724 on Build GRCh38
rs768153718 Chr.9: 97034773-97034773 on Build GRCh38
rs756565741 Chr.9: 97034928-97034928 on Build GRCh38
rs765512690 Chr.9: 97034675-97034675 on Build GRCh38
rs752090471 Chr.9: 97034704-97034704 on Build GRCh38
rs768094452 Chr.9: 97034863-97034863 on Build GRCh38
rs150639946 Chr.9: 97034911-97034911 on Build GRCh38
rs766894163 Chr.9: 97034691-97034691 on Build GRCh38
rs780000387 Chr.9: 97034829-97034829 on Build GRCh38
rs764881156 Chr.9: 97034889-97034889 on Build GRCh38
rs748846139 Chr.9: 97034991-97034991 on Build GRCh38
rs749524740 Chr.9: 97034849-97034849 on Build GRCh38
rs775172356 Chr.9: 97034848-97034848 on Build GRCh38
rs780845597 Chr.9: 97034717-97034717 on Build GRCh38
rs897457600 Chr.9: 97034721-97034721 on Build GRCh38
rs928653103 Chr.9: 97034949-97034949 on Build GRCh38

View Full Product Details