Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97922328-97922822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMO
Entrez Gene ID
51531
Gene Name
tRNA methyltransferase O
Gene Aliases
C9orf156, HSPC219, NAP1
UniGene
Hs.9196
Gene Chromosome Location
Chr.9: 97876321-97922570 on Build GRCh38
Transcripts
NM_016481.4 XM_005252047.3 XM_005252048.4 XM_005252049.4 XM_005252050.4 XM_011518776.2 XM_011518777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
455018 c.19T>C p.S7P Chr.9: 97922475-97922475 on Build GRCh38
3763479 c.13T>C p.S5P Chr.9: 97922475-97922475 on Build GRCh38
3763480 c.16T>C p.S6P Chr.9: 97922475-97922475 on Build GRCh38
6616843 c.36A>G p.T12T Chr.9: 97922458-97922458 on Build GRCh38
3763477 c.51T>G p.V17V Chr.9: 97922437-97922437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756586569 Chr.9: 97922450-97922450 on Build GRCh38
rs766164192 Chr.9: 97922479-97922479 on Build GRCh38
rs746189244 Chr.9: 97922405-97922405 on Build GRCh38
rs775767150 Chr.9: 97922468-97922468 on Build GRCh38
rs746950691 Chr.9: 97922383-97922383 on Build GRCh38
rs746111324 Chr.9: 97922462-97922462 on Build GRCh38
rs867758231 Chr.9: 97922456-97922456 on Build GRCh38
rs147482198 Chr.9: 97922757-97922757 on Build GRCh38
rs947833562 Chr.9: 97922634-97922634 on Build GRCh38
rs977661858 Chr.9: 97922606-97922606 on Build GRCh38
rs377466224 Chr.9: 97922376-97922376 on Build GRCh38
rs150014670 Chr.9: 97922469-97922469 on Build GRCh38
rs1055387083 Chr.9: 97922731-97922731 on Build GRCh38
rs932433735 Chr.9: 97922382-97922382 on Build GRCh38
rs372240346 Chr.9: 97922501-97922501 on Build GRCh38
rs770982896 Chr.9: 97922607-97922607 on Build GRCh38
rs939790842 Chr.9: 97922696-97922696 on Build GRCh38
rs1019393341 Chr.9: 97922617-97922617 on Build GRCh38
rs895355163 Chr.9: 97922605-97922605 on Build GRCh38
rs3183928 Chr.9: 97922437-97922437 on Build GRCh38
rs984251105 Chr.9: 97922656-97922656 on Build GRCh38
rs906765305 Chr.9: 97922581-97922581 on Build GRCh38
rs368353816 Chr.9: 97922489-97922489 on Build GRCh38
rs1018145481 Chr.9: 97922760-97922760 on Build GRCh38
rs1037664824 Chr.9: 97922570-97922570 on Build GRCh38
rs781168398 Chr.9: 97922623-97922623 on Build GRCh38
rs952737057 Chr.9: 97922430-97922430 on Build GRCh38
rs187366296 Chr.9: 97922407-97922407 on Build GRCh38
rs1028071458 Chr.9: 97922647-97922647 on Build GRCh38
rs965255679 Chr.9: 97922624-97922624 on Build GRCh38
rs894926078 Chr.9: 97922490-97922490 on Build GRCh38
rs964193823 Chr.9: 97922568-97922568 on Build GRCh38
rs901851686 Chr.9: 97922790-97922790 on Build GRCh38
rs552718531 Chr.9: 97922673-97922673 on Build GRCh38
rs764924410 Chr.9: 97922433-97922433 on Build GRCh38
rs909652527 Chr.9: 97922675-97922675 on Build GRCh38
rs972414364 Chr.9: 97922698-97922698 on Build GRCh38
rs534533836 Chr.9: 97922632-97922632 on Build GRCh38
rs760579392 Chr.9: 97922523-97922523 on Build GRCh38
rs939713610 Chr.9: 97922569-97922569 on Build GRCh38
rs895287842 Chr.9: 97922746-97922746 on Build GRCh38
rs761542305 Chr.9: 97922432-97922432 on Build GRCh38
rs776323798 Chr.9: 97922429-97922429 on Build GRCh38
rs376128350 Chr.9: 97922453-97922453 on Build GRCh38
rs756133526 Chr.9: 97922385-97922385 on Build GRCh38
rs532293548 Chr.9: 97922367-97922367 on Build GRCh38
rs3824496 Chr.9: 97922363-97922363 on Build GRCh38
rs750538803 Chr.9: 97922533-97922533 on Build GRCh38
rs200242824 Chr.9: 97922464-97922464 on Build GRCh38
rs570367410 Chr.9: 97922630-97922630 on Build GRCh38
rs764684064 Chr.9: 97922474-97922474 on Build GRCh38
rs929852539 Chr.9: 97922669-97922669 on Build GRCh38
rs927648629 Chr.9: 97922719-97922719 on Build GRCh38
rs370852825 Chr.9: 97922486-97922486 on Build GRCh38
rs574170108 Chr.9: 97922471-97922471 on Build GRCh38
rs780071211 Chr.9: 97922539-97922539 on Build GRCh38
rs547981563 Chr.9: 97922544-97922544 on Build GRCh38
rs776030045 Chr.9: 97922410-97922410 on Build GRCh38
rs148772479 Chr.9: 97922482-97922482 on Build GRCh38
rs765431805 Chr.9: 97922784-97922784 on Build GRCh38
rs759453936 Chr.9: 97922485-97922485 on Build GRCh38
rs999515871 Chr.9: 97922615-97922615 on Build GRCh38
rs199804891 Chr.9: 97922496-97922496 on Build GRCh38
rs879545148 Chr.9: 97922514-97922514 on Build GRCh38
rs1003730279 Chr.9: 97922454-97922454 on Build GRCh38
rs766106334 Chr.9: 97922446-97922446 on Build GRCh38
rs946885722 Chr.9: 97922763-97922763 on Build GRCh38
rs1022715159 Chr.9: 97922477-97922477 on Build GRCh38
rs970705746 Chr.9: 97922377-97922377 on Build GRCh38
rs1055291691 Chr.9: 97922602-97922602 on Build GRCh38
rs749477329 Chr.9: 97922395-97922395 on Build GRCh38
rs569949225 Chr.9: 97922610-97922610 on Build GRCh38
rs1046995866 Chr.9: 97922679-97922679 on Build GRCh38
rs374859311 Chr.9: 97922513-97922513 on Build GRCh38
rs1046451024 Chr.9: 97922649-97922649 on Build GRCh38
rs967387582 Chr.9: 97922794-97922794 on Build GRCh38
rs781749940 Chr.9: 97922502-97922502 on Build GRCh38
rs778886130 Chr.9: 97922491-97922491 on Build GRCh38
rs548590730 Chr.9: 97922609-97922609 on Build GRCh38
rs1018941809 Chr.9: 97922786-97922786 on Build GRCh38
rs939581710 Chr.9: 97922730-97922730 on Build GRCh38
rs769321252 Chr.9: 97922512-97922512 on Build GRCh38
rs772433249 Chr.9: 97922465-97922465 on Build GRCh38
rs964372825 Chr.9: 97922445-97922445 on Build GRCh38
rs201610182 Chr.9: 97922537-97922537 on Build GRCh38
rs749696407 Chr.9: 97922516-97922517 on Build GRCh38
rs193014744 Chr.9: 97922713-97922713 on Build GRCh38
rs574228649 Chr.9: 97922692-97922692 on Build GRCh38
rs771207689 Chr.9: 97922398-97922398 on Build GRCh38
rs369570790 Chr.9: 97922499-97922499 on Build GRCh38
rs752586451 Chr.9: 97922522-97922522 on Build GRCh38
rs1003825211 Chr.9: 97922597-97922597 on Build GRCh38
rs1044306489 Chr.9: 97922773-97922773 on Build GRCh38
rs115749354 Chr.9: 97922612-97922612 on Build GRCh38
rs780410911 Chr.9: 97922469-97922470 on Build GRCh38
rs777876778 Chr.9: 97922391-97922391 on Build GRCh38
rs767335042 Chr.9: 97922520-97922520 on Build GRCh38
rs12379031 Chr.9: 97922707-97922707 on Build GRCh38
rs139234146 Chr.9: 97922449-97922449 on Build GRCh38
rs765330669 Chr.9: 97922526-97922526 on Build GRCh38
rs761108572 Chr.9: 97922427-97922427 on Build GRCh38
rs369881999 Chr.9: 97922455-97922455 on Build GRCh38
rs530119768 Chr.9: 97922564-97922564 on Build GRCh38
rs922637195 Chr.9: 97922586-97922587 on Build GRCh38
rs776553099 Chr.9: 97922546-97922546 on Build GRCh38
rs758588929 Chr.9: 97922494-97922494 on Build GRCh38
rs3183927 Chr.9: 97922475-97922475 on Build GRCh38
rs372086299 Chr.9: 97922458-97922458 on Build GRCh38
rs139909617 Chr.9: 97922785-97922785 on Build GRCh38
rs777289698 Chr.9: 97922473-97922473 on Build GRCh38
rs758172189 Chr.9: 97922442-97922442 on Build GRCh38
rs751859231 Chr.9: 97922541-97922541 on Build GRCh38
rs565535676 Chr.9: 97922518-97922518 on Build GRCh38
rs370149200 Chr.9: 97922381-97922381 on Build GRCh38
rs779127924 Chr.9: 97922460-97922460 on Build GRCh38
rs906663591 Chr.9: 97922450-97922450 on Build GRCh38
rs575139961 Chr.9: 97922772-97922772 on Build GRCh38
rs762557863 Chr.9: 97922478-97922478 on Build GRCh38
rs952539856 Chr.9: 97922519-97922519 on Build GRCh38
rs774568709 Chr.9: 97922402-97922402 on Build GRCh38
rs748686651 Chr.9: 97922505-97922505 on Build GRCh38
rs968006235 Chr.9: 97922598-97922598 on Build GRCh38
rs559901684 Chr.9: 97922559-97922559 on Build GRCh38
rs1012319293 Chr.9: 97922493-97922493 on Build GRCh38
rs552063672 Chr.9: 97922625-97922625 on Build GRCh38
rs751488458 Chr.9: 97922451-97922451 on Build GRCh38
rs200865938 Chr.9: 97922487-97922487 on Build GRCh38

View Full Product Details