Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98944057-98944549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL15A1
Entrez Gene ID
1306
Gene Name
collagen type XV alpha 1 chain
Gene Aliases
-
UniGene
Hs.409034
Gene Chromosome Location
Chr.9: 98943392-99070792 on Build GRCh38
Transcripts
NM_001855.4 XM_011518214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361994 c.47T>C p.L16P Chr.9: 98944197-98944197 on Build GRCh38
3952106 c.62C>A p.T21K Chr.9: 98944212-98944212 on Build GRCh38
1103161 c.67C>A p.L23I Chr.9: 98944217-98944217 on Build GRCh38
4759494 c.89G>A p.R30H Chr.9: 98944239-98944239 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774761012 Chr.9: 98944214-98944214 on Build GRCh38
rs145179644 Chr.9: 98944241-98944241 on Build GRCh38
rs768245842 Chr.9: 98944285-98944285 on Build GRCh38
rs1030191501 Chr.9: 98944385-98944400 on Build GRCh38
rs776468256 Chr.9: 98944291-98944291 on Build GRCh38
rs991376688 Chr.9: 98944274-98944274 on Build GRCh38
rs997614391 Chr.9: 98944522-98944522 on Build GRCh38
rs761736585 Chr.9: 98944230-98944230 on Build GRCh38
rs527704357 Chr.9: 98944372-98944372 on Build GRCh38
rs536993581 Chr.9: 98944506-98944506 on Build GRCh38
rs758313397 Chr.9: 98944248-98944248 on Build GRCh38
rs1020932695 Chr.9: 98944238-98944238 on Build GRCh38
rs770192792 Chr.9: 98944526-98944526 on Build GRCh38
rs551852521 Chr.9: 98944339-98944339 on Build GRCh38
rs897554790 Chr.9: 98944336-98944336 on Build GRCh38
rs780194163 Chr.9: 98944082-98944082 on Build GRCh38
rs571525747 Chr.9: 98944349-98944349 on Build GRCh38
rs756021416 Chr.9: 98944355-98944355 on Build GRCh38
rs368381123 Chr.9: 98944388-98944388 on Build GRCh38
rs567940582 Chr.9: 98944477-98944477 on Build GRCh38
rs755780960 Chr.9: 98944147-98944147 on Build GRCh38
rs556824568 Chr.9: 98944109-98944109 on Build GRCh38
rs746396613 Chr.9: 98944265-98944265 on Build GRCh38
rs766815864 Chr.9: 98944116-98944116 on Build GRCh38
rs747112212 Chr.9: 98944088-98944088 on Build GRCh38
rs906500985 Chr.9: 98944467-98944467 on Build GRCh38
rs771506945 Chr.9: 98944216-98944216 on Build GRCh38
rs756174426 Chr.9: 98944250-98944250 on Build GRCh38
rs897482912 Chr.9: 98944084-98944084 on Build GRCh38
rs753626754 Chr.9: 98944152-98944152 on Build GRCh38
rs748399210 Chr.9: 98944180-98944180 on Build GRCh38
rs562741376 Chr.9: 98944293-98944293 on Build GRCh38
rs577588200 Chr.9: 98944225-98944225 on Build GRCh38
rs778778001 Chr.9: 98944167-98944167 on Build GRCh38
rs531668067 Chr.9: 98944316-98944316 on Build GRCh38
rs201993304 Chr.9: 98944172-98944172 on Build GRCh38
rs888039421 Chr.9: 98944402-98944402 on Build GRCh38
rs374798276 Chr.9: 98944199-98944199 on Build GRCh38
rs372448914 Chr.9: 98944081-98944081 on Build GRCh38
rs770165221 Chr.9: 98944105-98944105 on Build GRCh38
rs199761983 Chr.9: 98944259-98944259 on Build GRCh38
rs375160252 Chr.9: 98944146-98944146 on Build GRCh38
rs779292407 Chr.9: 98944262-98944262 on Build GRCh38
rs765102697 Chr.9: 98944098-98944106 on Build GRCh38
rs540101582 Chr.9: 98944228-98944228 on Build GRCh38
rs757102272 Chr.9: 98944158-98944158 on Build GRCh38
rs765058784 Chr.9: 98944233-98944233 on Build GRCh38
rs939611827 Chr.9: 98944497-98944497 on Build GRCh38
rs200866045 Chr.9: 98944196-98944196 on Build GRCh38
rs369351245 Chr.9: 98944171-98944171 on Build GRCh38
rs7039672 Chr.9: 98944309-98944309 on Build GRCh38
rs1055359742 Chr.9: 98944505-98944505 on Build GRCh38
rs749712321 Chr.9: 98944203-98944203 on Build GRCh38
rs556785769 Chr.9: 98944516-98944516 on Build GRCh38
rs772567563 Chr.9: 98944270-98944270 on Build GRCh38
rs760413730 Chr.9: 98944223-98944223 on Build GRCh38
rs575245670 Chr.9: 98944127-98944127 on Build GRCh38
rs1037441819 Chr.9: 98944405-98944405 on Build GRCh38
rs149989981 Chr.9: 98944237-98944237 on Build GRCh38
rs940105487 Chr.9: 98944380-98944380 on Build GRCh38
rs182109129 Chr.9: 98944134-98944134 on Build GRCh38
rs974656470 Chr.9: 98944326-98944326 on Build GRCh38
rs764015031 Chr.9: 98944149-98944149 on Build GRCh38
rs776125052 Chr.9: 98944276-98944276 on Build GRCh38
rs529177276 Chr.9: 98944249-98944249 on Build GRCh38
rs758301996 Chr.9: 98944163-98944164 on Build GRCh38
rs868652225 Chr.9: 98944278-98944278 on Build GRCh38
rs369174893 Chr.9: 98944373-98944373 on Build GRCh38
rs201106974 Chr.9: 98944282-98944282 on Build GRCh38
rs774909394 Chr.9: 98944126-98944126 on Build GRCh38
rs949858460 Chr.9: 98944297-98944297 on Build GRCh38
rs775028238 Chr.9: 98944219-98944219 on Build GRCh38
rs761507645 Chr.9: 98944299-98944299 on Build GRCh38
rs1008224362 Chr.9: 98944421-98944421 on Build GRCh38
rs748380307 Chr.9: 98944103-98944103 on Build GRCh38
rs1048866369 Chr.9: 98944371-98944371 on Build GRCh38
rs954312763 Chr.9: 98944078-98944078 on Build GRCh38
rs771281346 Chr.9: 98944210-98944210 on Build GRCh38
rs1039239988 Chr.9: 98944457-98944457 on Build GRCh38
rs752580822 Chr.9: 98944105-98944105 on Build GRCh38
rs760069779 Chr.9: 98944215-98944215 on Build GRCh38
rs752346135 Chr.9: 98944144-98944144 on Build GRCh38
rs776757383 Chr.9: 98944100-98944100 on Build GRCh38

View Full Product Details