Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98985486-98985983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL15A1
Entrez Gene ID
1306
Gene Name
collagen type XV alpha 1 chain
Gene Aliases
-
UniGene
Hs.409034
Gene Chromosome Location
Chr.9: 98943392-99070792 on Build GRCh38
Transcripts
NM_001855.4 XM_011518214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260210 c.399G>A p.R133R Chr.9: 98985863-98985863 on Build GRCh38
6632503 c.230G>A p.R77H Chr.9: 98985694-98985694 on Build GRCh38
1103163 c.273C>T p.F91F Chr.9: 98985737-98985737 on Build GRCh38
3652248 c.504C>T p.V168V Chr.9: 98985968-98985968 on Build GRCh38
4470943 c.168C>T p.S56S Chr.9: 98985632-98985632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570016118 Chr.9: 98985957-98985957 on Build GRCh38
rs773097539 Chr.9: 98985779-98985779 on Build GRCh38
rs778572972 Chr.9: 98985824-98985824 on Build GRCh38
rs373086087 Chr.9: 98985885-98985885 on Build GRCh38
rs768302859 Chr.9: 98985596-98985596 on Build GRCh38
rs974480059 Chr.9: 98985741-98985741 on Build GRCh38
rs137919829 Chr.9: 98985623-98985623 on Build GRCh38
rs767075199 Chr.9: 98985930-98985930 on Build GRCh38
rs367883631 Chr.9: 98985627-98985627 on Build GRCh38
rs199509430 Chr.9: 98985547-98985547 on Build GRCh38
rs374984385 Chr.9: 98985801-98985801 on Build GRCh38
rs756805553 Chr.9: 98985908-98985908 on Build GRCh38
rs766766042 Chr.9: 98985781-98985781 on Build GRCh38
rs770970455 Chr.9: 98985671-98985671 on Build GRCh38
rs755701903 Chr.9: 98985751-98985751 on Build GRCh38
rs779152552 Chr.9: 98985567-98985567 on Build GRCh38
rs759476425 Chr.9: 98985681-98985681 on Build GRCh38
rs751914677 Chr.9: 98985746-98985746 on Build GRCh38
rs902739986 Chr.9: 98985714-98985714 on Build GRCh38
rs754797361 Chr.9: 98985924-98985924 on Build GRCh38
rs141708640 Chr.9: 98985737-98985737 on Build GRCh38
rs753271019 Chr.9: 98985795-98985795 on Build GRCh38
rs199564502 Chr.9: 98985772-98985772 on Build GRCh38
rs774534609 Chr.9: 98985937-98985937 on Build GRCh38
rs780742930 Chr.9: 98985593-98985593 on Build GRCh38
rs775541891 Chr.9: 98985771-98985771 on Build GRCh38
rs781517725 Chr.9: 98985656-98985656 on Build GRCh38
rs750164683 Chr.9: 98985536-98985536 on Build GRCh38
rs763402396 Chr.9: 98985622-98985622 on Build GRCh38
rs746093934 Chr.9: 98985941-98985941 on Build GRCh38
rs747757801 Chr.9: 98985598-98985598 on Build GRCh38
rs756750402 Chr.9: 98985813-98985813 on Build GRCh38
rs150514154 Chr.9: 98985960-98985960 on Build GRCh38
rs773643079 Chr.9: 98985552-98985552 on Build GRCh38
rs995849677 Chr.9: 98985592-98985592 on Build GRCh38
rs372510443 Chr.9: 98985540-98985540 on Build GRCh38
rs767388264 Chr.9: 98985692-98985692 on Build GRCh38
rs753851581 Chr.9: 98985615-98985615 on Build GRCh38
rs1016086727 Chr.9: 98985716-98985716 on Build GRCh38
rs754085747 Chr.9: 98985555-98985555 on Build GRCh38
rs765895796 Chr.9: 98985699-98985699 on Build GRCh38
rs368284198 Chr.9: 98985766-98985766 on Build GRCh38
rs777101028 Chr.9: 98985755-98985755 on Build GRCh38
rs200391843 Chr.9: 98985861-98985861 on Build GRCh38
rs141642715 Chr.9: 98985934-98985934 on Build GRCh38
rs749004076 Chr.9: 98985763-98985763 on Build GRCh38
rs771819188 Chr.9: 98985769-98985769 on Build GRCh38
rs758682861 Chr.9: 98985584-98985584 on Build GRCh38
rs374938012 Chr.9: 98985922-98985922 on Build GRCh38
rs777271466 Chr.9: 98985539-98985539 on Build GRCh38
rs762264972 Chr.9: 98985956-98985956 on Build GRCh38
rs754341520 Chr.9: 98985906-98985906 on Build GRCh38
rs974429335 Chr.9: 98985526-98985526 on Build GRCh38
rs745493353 Chr.9: 98985825-98985825 on Build GRCh38
rs201906975 Chr.9: 98985780-98985780 on Build GRCh38
rs759553086 Chr.9: 98985831-98985831 on Build GRCh38
rs774763433 Chr.9: 98985862-98985862 on Build GRCh38
rs1051086 Chr.9: 98985610-98985610 on Build GRCh38
rs781031691 Chr.9: 98985925-98985925 on Build GRCh38
rs769183016 Chr.9: 98985663-98985663 on Build GRCh38
rs750104746 Chr.9: 98985920-98985920 on Build GRCh38
rs755467932 Chr.9: 98985638-98985638 on Build GRCh38
rs377517566 Chr.9: 98985541-98985541 on Build GRCh38
rs751826956 Chr.9: 98985788-98985788 on Build GRCh38
rs2075662 Chr.9: 98985952-98985952 on Build GRCh38
rs746299482 Chr.9: 98985579-98985579 on Build GRCh38
rs370426713 Chr.9: 98985644-98985644 on Build GRCh38
rs765559698 Chr.9: 98985619-98985619 on Build GRCh38
rs375324510 Chr.9: 98985626-98985626 on Build GRCh38
rs781781615 Chr.9: 98985749-98985749 on Build GRCh38
rs964444166 Chr.9: 98985512-98985512 on Build GRCh38
rs147085668 Chr.9: 98985800-98985800 on Build GRCh38
rs768418278 Chr.9: 98985842-98985842 on Build GRCh38
rs115245175 Chr.9: 98985603-98985603 on Build GRCh38
rs370081942 Chr.9: 98985750-98985750 on Build GRCh38
rs776616947 Chr.9: 98985851-98985851 on Build GRCh38
rs373530299 Chr.9: 98985880-98985880 on Build GRCh38
rs764194365 Chr.9: 98985614-98985614 on Build GRCh38
rs777141387 Chr.9: 98985953-98985953 on Build GRCh38
rs143274396 Chr.9: 98985659-98985659 on Build GRCh38
rs201085523 Chr.9: 98985881-98985881 on Build GRCh38
rs779670663 Chr.9: 98985832-98985832 on Build GRCh38
rs377632543 Chr.9: 98985597-98985597 on Build GRCh38
rs755380230 Chr.9: 98985747-98985747 on Build GRCh38
rs758507654 Chr.9: 98985721-98985721 on Build GRCh38
rs544454009 Chr.9: 98985789-98985789 on Build GRCh38
rs577671129 Chr.9: 98985606-98985606 on Build GRCh38
rs774322799 Chr.9: 98985613-98985613 on Build GRCh38
rs770506843 Chr.9: 98985765-98985765 on Build GRCh38
rs7045978 Chr.9: 98985902-98985902 on Build GRCh38
rs77414605 Chr.9: 98985595-98985595 on Build GRCh38
rs71501885 Chr.9: 98985840-98985840 on Build GRCh38
rs868065640 Chr.9: 98985575-98985575 on Build GRCh38
rs777809448 Chr.9: 98985554-98985554 on Build GRCh38
rs775770612 Chr.9: 98985949-98985949 on Build GRCh38
rs375715330 Chr.9: 98985605-98985605 on Build GRCh38
rs757437335 Chr.9: 98985616-98985616 on Build GRCh38
rs772130405 Chr.9: 98985942-98985942 on Build GRCh38
rs764971894 Chr.9: 98985530-98985530 on Build GRCh38
rs573608896 Chr.9: 98985675-98985675 on Build GRCh38
rs775282439 Chr.9: 98985693-98985693 on Build GRCh38
rs761016441 Chr.9: 98985951-98985951 on Build GRCh38
rs757462729 Chr.9: 98985559-98985559 on Build GRCh38
rs139245903 Chr.9: 98985694-98985694 on Build GRCh38
rs994542363 Chr.9: 98985635-98985635 on Build GRCh38
rs764674198 Chr.9: 98985913-98985913 on Build GRCh38
rs770735842 Chr.9: 98985672-98985672 on Build GRCh38
rs750678351 Chr.9: 98985711-98985711 on Build GRCh38
rs567540031 Chr.9: 98985923-98985923 on Build GRCh38
rs372460599 Chr.9: 98985543-98985543 on Build GRCh38
rs906527217 Chr.9: 98985628-98985628 on Build GRCh38
rs769945542 Chr.9: 98985778-98985778 on Build GRCh38
rs753231557 Chr.9: 98985804-98985804 on Build GRCh38
rs527804413 Chr.9: 98985893-98985893 on Build GRCh38
rs772958842 Chr.9: 98985665-98985665 on Build GRCh38

View Full Product Details