Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 99979959-99980507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERP44
Entrez Gene ID
23071
Gene Name
endoplasmic reticulum protein 44
Gene Aliases
PDIA10, TXNDC4
UniGene
Hs.154023
Gene Chromosome Location
Chr.9: 99979179-100099052 on Build GRCh38
Transcripts
NM_015051.2

SNP Detail

SNP ID Chromosome Location
rs995132026 Chr.9: 99980480-99980480 on Build GRCh38
rs372603092 Chr.9: 99980093-99980094 on Build GRCh38
rs184075701 Chr.9: 99980204-99980204 on Build GRCh38
rs372776467 Chr.9: 99980465-99980466 on Build GRCh38
rs1054453717 Chr.9: 99980449-99980450 on Build GRCh38
rs989874420 Chr.9: 99980216-99980216 on Build GRCh38
rs755886874 Chr.9: 99980180-99980181 on Build GRCh38
rs936656039 Chr.9: 99980149-99980149 on Build GRCh38
rs139174595 Chr.9: 99980205-99980205 on Build GRCh38
rs1025049769 Chr.9: 99980163-99980163 on Build GRCh38
rs181875924 Chr.9: 99980399-99980399 on Build GRCh38
rs910847487 Chr.9: 99980284-99980284 on Build GRCh38
rs895141779 Chr.9: 99980460-99980460 on Build GRCh38
rs936899190 Chr.9: 99980446-99980446 on Build GRCh38
rs759778991 Chr.9: 99980288-99980288 on Build GRCh38
rs970534010 Chr.9: 99980178-99980182 on Build GRCh38
rs899428490 Chr.9: 99980409-99980409 on Build GRCh38
rs775682862 Chr.9: 99980437-99980437 on Build GRCh38
rs769579199 Chr.9: 99980072-99980072 on Build GRCh38
rs189241300 Chr.9: 99980243-99980243 on Build GRCh38
rs942367034 Chr.9: 99980286-99980286 on Build GRCh38
rs532969828 Chr.9: 99980077-99980077 on Build GRCh38
rs895141106 Chr.9: 99980037-99980037 on Build GRCh38
rs924124959 Chr.9: 99980419-99980419 on Build GRCh38
rs370319464 Chr.9: 99980249-99980249 on Build GRCh38
rs1051831234 Chr.9: 99980289-99980289 on Build GRCh38
rs560092323 Chr.9: 99980397-99980397 on Build GRCh38
rs150753794 Chr.9: 99980132-99980132 on Build GRCh38
rs781440683 Chr.9: 99980171-99980172 on Build GRCh38
rs940865917 Chr.9: 99980396-99980396 on Build GRCh38
rs933463745 Chr.9: 99980309-99980309 on Build GRCh38
rs1042553285 Chr.9: 99980324-99980324 on Build GRCh38
rs555882561 Chr.9: 99980336-99980336 on Build GRCh38
rs952630445 Chr.9: 99980240-99980240 on Build GRCh38
rs977961572 Chr.9: 99980093-99980093 on Build GRCh38

View Full Product Details