Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101421042-101421582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOB
Entrez Gene ID
229
Gene Name
aldolase, fructose-bisphosphate B
Gene Aliases
ALDB, ALDO2
UniGene
Hs.530274
Gene Chromosome Location
Chr.9: 101420560-101435780 on Build GRCh38
Transcripts
NM_000035.3

SNP Detail

SNP ID Chromosome Location
rs937856450 Chr.9: 101421154-101421154 on Build GRCh38
rs1017488987 Chr.9: 101421136-101421136 on Build GRCh38
rs762406910 Chr.9: 101421518-101421518 on Build GRCh38
rs564268678 Chr.9: 101421111-101421111 on Build GRCh38
rs896497786 Chr.9: 101421247-101421247 on Build GRCh38
rs539546507 Chr.9: 101421374-101421374 on Build GRCh38
rs748992077 Chr.9: 101421296-101421296 on Build GRCh38
rs951719614 Chr.9: 101421105-101421105 on Build GRCh38
rs886063289 Chr.9: 101421095-101421095 on Build GRCh38
rs754731873 Chr.9: 101421151-101421151 on Build GRCh38
rs949099709 Chr.9: 101421435-101421435 on Build GRCh38
rs12347519 Chr.9: 101421098-101421098 on Build GRCh38
rs765455562 Chr.9: 101421527-101421527 on Build GRCh38
rs540272117 Chr.9: 101421242-101421242 on Build GRCh38
rs566368466 Chr.9: 101421522-101421522 on Build GRCh38
rs760952735 Chr.9: 101421421-101421421 on Build GRCh38
rs41296049 Chr.9: 101421197-101421197 on Build GRCh38
rs530643699 Chr.9: 101421305-101421305 on Build GRCh38
rs887552586 Chr.9: 101421549-101421549 on Build GRCh38
rs949381406 Chr.9: 101421514-101421514 on Build GRCh38
rs916578819 Chr.9: 101421484-101421484 on Build GRCh38
rs886063290 Chr.9: 101421520-101421520 on Build GRCh38
rs945169695 Chr.9: 101421207-101421207 on Build GRCh38
rs983455988 Chr.9: 101421329-101421329 on Build GRCh38
rs1042042177 Chr.9: 101421447-101421447 on Build GRCh38
rs773210631 Chr.9: 101421434-101421434 on Build GRCh38
rs896226667 Chr.9: 101421179-101421179 on Build GRCh38
rs1032585724 Chr.9: 101421285-101421285 on Build GRCh38
rs978298249 Chr.9: 101421434-101421434 on Build GRCh38
rs768345214 Chr.9: 101421160-101421163 on Build GRCh38
rs907842528 Chr.9: 101421332-101421332 on Build GRCh38
rs966587043 Chr.9: 101421415-101421415 on Build GRCh38
rs144337754 Chr.9: 101421365-101421365 on Build GRCh38
rs148781781 Chr.9: 101421385-101421385 on Build GRCh38
rs764461347 Chr.9: 101421459-101421459 on Build GRCh38
rs142431256 Chr.9: 101421541-101421541 on Build GRCh38
rs781001201 Chr.9: 101421163-101421163 on Build GRCh38
rs902219663 Chr.9: 101421457-101421457 on Build GRCh38
rs897330283 Chr.9: 101421283-101421283 on Build GRCh38
rs777064471 Chr.9: 101421461-101421461 on Build GRCh38
rs984398166 Chr.9: 101421127-101421127 on Build GRCh38
rs978202633 Chr.9: 101421241-101421241 on Build GRCh38
rs1005924855 Chr.9: 101421552-101421552 on Build GRCh38
rs575767946 Chr.9: 101421087-101421087 on Build GRCh38
rs1013218532 Chr.9: 101421182-101421182 on Build GRCh38
rs936666980 Chr.9: 101421250-101421250 on Build GRCh38
rs529257579 Chr.9: 101421274-101421274 on Build GRCh38
rs1047037430 Chr.9: 101421269-101421269 on Build GRCh38
rs150017672 Chr.9: 101421299-101421299 on Build GRCh38
rs918812534 Chr.9: 101421529-101421529 on Build GRCh38
rs774019250 Chr.9: 101421366-101421366 on Build GRCh38
rs17772845 Chr.9: 101421304-101421304 on Build GRCh38
rs1054937198 Chr.9: 101421386-101421386 on Build GRCh38
rs917684057 Chr.9: 101421403-101421403 on Build GRCh38
rs550789144 Chr.9: 101421357-101421357 on Build GRCh38
rs562463127 Chr.9: 101421401-101421401 on Build GRCh38
rs974436732 Chr.9: 101421369-101421369 on Build GRCh38
rs1013594139 Chr.9: 101421251-101421251 on Build GRCh38
rs746649622 Chr.9: 101421078-101421078 on Build GRCh38
rs895422053 Chr.9: 101421255-101421255 on Build GRCh38
rs996768943 Chr.9: 101421538-101421538 on Build GRCh38
rs965283551 Chr.9: 101421295-101421295 on Build GRCh38
rs990856931 Chr.9: 101421470-101421470 on Build GRCh38
rs954756064 Chr.9: 101421306-101421306 on Build GRCh38
rs919836112 Chr.9: 101421373-101421373 on Build GRCh38
rs1055115783 Chr.9: 101421254-101421254 on Build GRCh38
rs886063291 Chr.9: 101421523-101421523 on Build GRCh38
rs900725762 Chr.9: 101421523-101421523 on Build GRCh38
rs41308902 Chr.9: 101421257-101421257 on Build GRCh38

View Full Product Details