Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101425364-101425861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOB
Entrez Gene ID
229
Gene Name
aldolase, fructose-bisphosphate B
Gene Aliases
ALDB, ALDO2
UniGene
Hs.530274
Gene Chromosome Location
Chr.9: 101420560-101435780 on Build GRCh38
Transcripts
NM_000035.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7320026 c.677G>A p.G226D Chr.9: 101425575-101425575 on Build GRCh38
5410049 c.654T>C p.N218N Chr.9: 101425598-101425598 on Build GRCh38
1103331 c.689A>C p.K230T Chr.9: 101425563-101425563 on Build GRCh38
5587158 c.696C>T p.N232N Chr.9: 101425556-101425556 on Build GRCh38
3652366 c.775C>T p.R259C Chr.9: 101425477-101425477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866549991 Chr.9: 101425513-101425513 on Build GRCh38
rs201424676 Chr.9: 101425477-101425477 on Build GRCh38
rs978432591 Chr.9: 101425672-101425672 on Build GRCh38
rs748157721 Chr.9: 101425612-101425612 on Build GRCh38
rs968428089 Chr.9: 101425389-101425389 on Build GRCh38
rs756759302 Chr.9: 101425410-101425410 on Build GRCh38
rs966168711 Chr.9: 101425748-101425748 on Build GRCh38
rs968834739 Chr.9: 101425671-101425671 on Build GRCh38
rs182245025 Chr.9: 101425442-101425442 on Build GRCh38
rs1803086 Chr.9: 101425606-101425606 on Build GRCh38
rs930171273 Chr.9: 101425801-101425801 on Build GRCh38
rs61757689 Chr.9: 101425571-101425571 on Build GRCh38
rs141888548 Chr.9: 101425447-101425447 on Build GRCh38
rs35869228 Chr.9: 101425773-101425773 on Build GRCh38
rs184958489 Chr.9: 101425804-101425804 on Build GRCh38
rs190206611 Chr.9: 101425825-101425825 on Build GRCh38
rs745937509 Chr.9: 101425679-101425679 on Build GRCh38
rs142156974 Chr.9: 101425685-101425685 on Build GRCh38
rs764701775 Chr.9: 101425482-101425482 on Build GRCh38
rs771092366 Chr.9: 101425529-101425529 on Build GRCh38
rs759655888 Chr.9: 101425461-101425461 on Build GRCh38
rs772197524 Chr.9: 101425535-101425535 on Build GRCh38
rs752431371 Chr.9: 101425551-101425551 on Build GRCh38
rs376978142 Chr.9: 101425562-101425562 on Build GRCh38
rs751133985 Chr.9: 101425538-101425538 on Build GRCh38
rs117738198 Chr.9: 101425425-101425425 on Build GRCh38
rs778655699 Chr.9: 101425417-101425417 on Build GRCh38
rs786204503 Chr.9: 101425629-101425629 on Build GRCh38
rs977026708 Chr.9: 101425693-101425693 on Build GRCh38
rs978401483 Chr.9: 101425416-101425416 on Build GRCh38
rs754791588 Chr.9: 101425581-101425581 on Build GRCh38
rs751175021 Chr.9: 101425579-101425579 on Build GRCh38
rs775786023 Chr.9: 101425537-101425537 on Build GRCh38
rs905795695 Chr.9: 101425516-101425516 on Build GRCh38
rs777324536 Chr.9: 101425554-101425554 on Build GRCh38
rs764927361 Chr.9: 101425494-101425494 on Build GRCh38
rs1019102997 Chr.9: 101425738-101425738 on Build GRCh38
rs774656173 Chr.9: 101425453-101425453 on Build GRCh38
rs1055615924 Chr.9: 101425558-101425558 on Build GRCh38
rs776496714 Chr.9: 101425434-101425434 on Build GRCh38
rs1035568052 Chr.9: 101425539-101425539 on Build GRCh38
rs937202326 Chr.9: 101425536-101425536 on Build GRCh38
rs766027880 Chr.9: 101425573-101425573 on Build GRCh38
rs767286399 Chr.9: 101425837-101425837 on Build GRCh38
rs546476555 Chr.9: 101425699-101425699 on Build GRCh38
rs922833334 Chr.9: 101425697-101425697 on Build GRCh38
rs781473466 Chr.9: 101425637-101425638 on Build GRCh38
rs909969038 Chr.9: 101425813-101425813 on Build GRCh38
rs1045119481 Chr.9: 101425826-101425826 on Build GRCh38
rs777708190 Chr.9: 101425677-101425677 on Build GRCh38
rs766032602 Chr.9: 101425511-101425511 on Build GRCh38
rs776649110 Chr.9: 101425492-101425492 on Build GRCh38
rs779885811 Chr.9: 101425495-101425495 on Build GRCh38
rs781139407 Chr.9: 101425514-101425514 on Build GRCh38
rs773439417 Chr.9: 101425433-101425433 on Build GRCh38
rs749892001 Chr.9: 101425567-101425567 on Build GRCh38
rs897846555 Chr.9: 101425595-101425595 on Build GRCh38
rs118204426 Chr.9: 101425532-101425532 on Build GRCh38
rs541973411 Chr.9: 101425812-101425812 on Build GRCh38
rs868200339 Chr.9: 101425455-101425455 on Build GRCh38
rs201551409 Chr.9: 101425486-101425486 on Build GRCh38
rs912961119 Chr.9: 101425387-101425387 on Build GRCh38
rs371219301 Chr.9: 101425572-101425572 on Build GRCh38
rs922806049 Chr.9: 101425448-101425448 on Build GRCh38
rs146360505 Chr.9: 101425493-101425493 on Build GRCh38
rs762765534 Chr.9: 101425505-101425505 on Build GRCh38
rs967049159 Chr.9: 101425810-101425810 on Build GRCh38
rs772207233 Chr.9: 101425395-101425395 on Build GRCh38
rs1020052060 Chr.9: 101425670-101425670 on Build GRCh38
rs761061896 Chr.9: 101425480-101425480 on Build GRCh38
rs764501544 Chr.9: 101425546-101425546 on Build GRCh38
rs564662987 Chr.9: 101425836-101425836 on Build GRCh38
rs200102975 Chr.9: 101425578-101425578 on Build GRCh38
rs748031490 Chr.9: 101425519-101425519 on Build GRCh38
rs150644101 Chr.9: 101425502-101425502 on Build GRCh38
rs780943027 Chr.9: 101425598-101425598 on Build GRCh38
rs758133069 Chr.9: 101425579-101425579 on Build GRCh38
rs764872458 Chr.9: 101425564-101425564 on Build GRCh38
rs761151751 Chr.9: 101425544-101425544 on Build GRCh38
rs41281035 Chr.9: 101425504-101425504 on Build GRCh38
rs549697821 Chr.9: 101425390-101425390 on Build GRCh38
rs888499183 Chr.9: 101425488-101425488 on Build GRCh38
rs775516821 Chr.9: 101425398-101425398 on Build GRCh38
rs976501192 Chr.9: 101425783-101425783 on Build GRCh38
rs755991230 Chr.9: 101425658-101425658 on Build GRCh38
rs965153604 Chr.9: 101425472-101425472 on Build GRCh38
rs747021015 Chr.9: 101425429-101425429 on Build GRCh38
rs565827286 Chr.9: 101425476-101425476 on Build GRCh38
rs202031050 Chr.9: 101425580-101425580 on Build GRCh38
rs774265180 Chr.9: 101425530-101425530 on Build GRCh38
rs779686777 Chr.9: 101425428-101425428 on Build GRCh38
rs577601048 Chr.9: 101425686-101425686 on Build GRCh38
rs976450613 Chr.9: 101425474-101425474 on Build GRCh38

View Full Product Details