Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 103005384-103005889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYLC2
Entrez Gene ID
1539
Gene Name
cylicin 2
Gene Aliases
-
UniGene
Hs.3232
Gene Chromosome Location
Chr.9: 102995311-103018488 on Build GRCh38
Transcripts
NM_001340.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1103424 c.954T>C p.D318D Chr.9: 103005585-103005585 on Build GRCh38
1202803 c.871G>T p.E291* Chr.9: 103005502-103005502 on Build GRCh38
4389212 c.802A>T p.K268* Chr.9: 103005433-103005433 on Build GRCh38
3167495 c.765T>A p.D255E Chr.9: 103005396-103005396 on Build GRCh38
6114524 c.895G>T p.D299Y Chr.9: 103005526-103005526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930975245 Chr.9: 103005805-103005805 on Build GRCh38
rs750777444 Chr.9: 103005440-103005440 on Build GRCh38
rs936070852 Chr.9: 103005683-103005683 on Build GRCh38
rs781201790 Chr.9: 103005640-103005640 on Build GRCh38
rs778748789 Chr.9: 103005482-103005482 on Build GRCh38
rs757958924 Chr.9: 103005433-103005433 on Build GRCh38
rs377524091 Chr.9: 103005650-103005650 on Build GRCh38
rs16922388 Chr.9: 103005516-103005516 on Build GRCh38
rs571875621 Chr.9: 103005409-103005409 on Build GRCh38
rs199562230 Chr.9: 103005517-103005517 on Build GRCh38
rs376299522 Chr.9: 103005705-103005705 on Build GRCh38
rs149633662 Chr.9: 103005539-103005539 on Build GRCh38
rs778658742 Chr.9: 103005544-103005544 on Build GRCh38
rs768616436 Chr.9: 103005419-103005419 on Build GRCh38
rs755969012 Chr.9: 103005466-103005466 on Build GRCh38
rs201302612 Chr.9: 103005474-103005474 on Build GRCh38
rs369659683 Chr.9: 103005562-103005562 on Build GRCh38
rs912345821 Chr.9: 103005500-103005500 on Build GRCh38
rs780549272 Chr.9: 103005462-103005462 on Build GRCh38
rs367661428 Chr.9: 103005572-103005572 on Build GRCh38
rs765188712 Chr.9: 103005631-103005631 on Build GRCh38
rs777713576 Chr.9: 103005472-103005472 on Build GRCh38
rs369835418 Chr.9: 103005444-103005444 on Build GRCh38
rs773488089 Chr.9: 103005566-103005566 on Build GRCh38
rs965181568 Chr.9: 103005732-103005732 on Build GRCh38
rs772111575 Chr.9: 103005661-103005663 on Build GRCh38
rs140754596 Chr.9: 103005536-103005536 on Build GRCh38
rs745714965 Chr.9: 103005547-103005547 on Build GRCh38
rs769996075 Chr.9: 103005564-103005564 on Build GRCh38
rs748422105 Chr.9: 103005653-103005654 on Build GRCh38
rs752103381 Chr.9: 103005532-103005532 on Build GRCh38
rs201540530 Chr.9: 103005720-103005720 on Build GRCh38
rs748393005 Chr.9: 103005642-103005642 on Build GRCh38
rs754723609 Chr.9: 103005460-103005460 on Build GRCh38
rs144603458 Chr.9: 103005744-103005744 on Build GRCh38
rs776540560 Chr.9: 103005492-103005492 on Build GRCh38
rs185998342 Chr.9: 103005619-103005619 on Build GRCh38
rs763398202 Chr.9: 103005745-103005745 on Build GRCh38
rs773239550 Chr.9: 103005663-103005663 on Build GRCh38
rs781003105 Chr.9: 103005601-103005612 on Build GRCh38
rs774539013 Chr.9: 103005620-103005631 on Build GRCh38
rs532036391 Chr.9: 103005579-103005579 on Build GRCh38
rs767233678 Chr.9: 103005527-103005527 on Build GRCh38
rs1019855149 Chr.9: 103005726-103005726 on Build GRCh38
rs777465483 Chr.9: 103005451-103005453 on Build GRCh38
rs2298052 Chr.9: 103005420-103005420 on Build GRCh38
rs761982928 Chr.9: 103005499-103005499 on Build GRCh38
rs372929483 Chr.9: 103005613-103005613 on Build GRCh38
rs771609795 Chr.9: 103005581-103005581 on Build GRCh38
rs765726759 Chr.9: 103005585-103005585 on Build GRCh38
rs1031614542 Chr.9: 103005415-103005415 on Build GRCh38
rs964182319 Chr.9: 103005591-103005591 on Build GRCh38
rs370950999 Chr.9: 103005701-103005701 on Build GRCh38
rs759082916 Chr.9: 103005523-103005523 on Build GRCh38
rs540168296 Chr.9: 103005760-103005760 on Build GRCh38
rs762344627 Chr.9: 103005580-103005580 on Build GRCh38
rs145773594 Chr.9: 103005555-103005555 on Build GRCh38
rs200924574 Chr.9: 103005491-103005491 on Build GRCh38
rs570803926 Chr.9: 103005825-103005825 on Build GRCh38
rs749026661 Chr.9: 103005473-103005473 on Build GRCh38
rs373184793 Chr.9: 103005671-103005671 on Build GRCh38
rs763582781 Chr.9: 103005618-103005618 on Build GRCh38
rs775213952 Chr.9: 103005607-103005607 on Build GRCh38
rs769540404 Chr.9: 103005608-103005631 on Build GRCh38
rs567806323 Chr.9: 103005718-103005718 on Build GRCh38
rs774940202 Chr.9: 103005699-103005699 on Build GRCh38
rs762202245 Chr.9: 103005431-103005431 on Build GRCh38
rs747185979 Chr.9: 103005487-103005487 on Build GRCh38
rs568757462 Chr.9: 103005528-103005539 on Build GRCh38
rs972496902 Chr.9: 103005736-103005736 on Build GRCh38
rs745606773 Chr.9: 103005601-103005636 on Build GRCh38
rs138337835 Chr.9: 103005521-103005521 on Build GRCh38
rs527747098 Chr.9: 103005606-103005606 on Build GRCh38
rs924758805 Chr.9: 103005508-103005510 on Build GRCh38
rs977472835 Chr.9: 103005638-103005638 on Build GRCh38
rs755113842 Chr.9: 103005552-103005552 on Build GRCh38
rs763338109 Chr.9: 103005520-103005520 on Build GRCh38
rs373291704 Chr.9: 103005457-103005457 on Build GRCh38
rs3763636 Chr.9: 103005587-103005587 on Build GRCh38
rs984331772 Chr.9: 103005428-103005428 on Build GRCh38
rs772481741 Chr.9: 103005700-103005700 on Build GRCh38
rs760694867 Chr.9: 103005718-103005718 on Build GRCh38
rs184093078 Chr.9: 103005702-103005702 on Build GRCh38
rs757154630 Chr.9: 103005621-103005621 on Build GRCh38
rs766067392 Chr.9: 103005445-103005445 on Build GRCh38
rs777621557 Chr.9: 103005538-103005538 on Build GRCh38
rs771518182 Chr.9: 103005675-103005675 on Build GRCh38
rs756460777 Chr.9: 103005644-103005644 on Build GRCh38
rs752293876 Chr.9: 103005465-103005465 on Build GRCh38
rs1047365923 Chr.9: 103005858-103005858 on Build GRCh38
rs765811661 Chr.9: 103005433-103005433 on Build GRCh38
rs975652915 Chr.9: 103005458-103005458 on Build GRCh38
rs142695278 Chr.9: 103005635-103005635 on Build GRCh38

View Full Product Details