Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105619625-105620170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKTN
Entrez Gene ID
2218
Gene Name
fukutin
Gene Aliases
CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
UniGene
Hs.55777
Gene Chromosome Location
Chr.9: 105558117-105655950 on Build GRCh38
Transcripts
NM_001079802.1 NM_001198963.1 NM_006731.2 XM_006717014.2 XM_011518368.2 XM_011518369.2 XM_011518370.2 XM_011518371.2 XM_011518373.2 XM_011518374.2 XM_011518375.2 XM_011518376.2 XM_011518378.2 XM_011518379.2 XM_011518381.2 XM_011518387.2 XM_011518389.2 XM_011518390.2 XM_011518391.2 XM_017014462.1 XM_017014463.1 XM_017014464.1 XM_017014465.1 XM_017014466.1 XM_017014467.1 XM_017014468.1 XM_017014469.1 XM_017014470.1 XM_017014471.1 XM_017014472.1 XM_017014473.1 XM_017014474.1 XM_017014475.1 XM_017014476.1 XM_017014477.1 XM_017014478.1 XM_017014479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1103575 c.1056C>A p.S352R Chr.9: 105619945-105619945 on Build GRCh38
5531292 c.1068C>T p.S356S Chr.9: 105619957-105619957 on Build GRCh38
6847815 c.1093C>G p.L365V Chr.9: 105619982-105619982 on Build GRCh38
1103576 c.1107C>A p.F369L Chr.9: 105619996-105619996 on Build GRCh38
3716107 c.1048G>C p.E350Q Chr.9: 105619937-105619937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751317602 Chr.9: 105619954-105619954 on Build GRCh38
rs119463993 Chr.9: 105619962-105619962 on Build GRCh38
rs1034845210 Chr.9: 105620091-105620091 on Build GRCh38
rs970499794 Chr.9: 105620067-105620067 on Build GRCh38
rs1007950406 Chr.9: 105619694-105619694 on Build GRCh38
rs891588561 Chr.9: 105620072-105620072 on Build GRCh38
rs1024735310 Chr.9: 105619818-105619818 on Build GRCh38
rs748766811 Chr.9: 105619684-105619685 on Build GRCh38
rs995219029 Chr.9: 105619754-105619754 on Build GRCh38
rs374381691 Chr.9: 105619991-105619991 on Build GRCh38
rs398123555 Chr.9: 105620056-105620057 on Build GRCh38
rs780791268 Chr.9: 105619995-105619995 on Build GRCh38
rs1039466532 Chr.9: 105619956-105619956 on Build GRCh38
rs911005943 Chr.9: 105619697-105619697 on Build GRCh38
rs747639725 Chr.9: 105619999-105619999 on Build GRCh38
rs148975262 Chr.9: 105620048-105620048 on Build GRCh38
rs761813807 Chr.9: 105620104-105620104 on Build GRCh38
rs187457647 Chr.9: 105619773-105619773 on Build GRCh38
rs905981141 Chr.9: 105619996-105619996 on Build GRCh38
rs563259780 Chr.9: 105619677-105619677 on Build GRCh38
rs946346185 Chr.9: 105619898-105619898 on Build GRCh38
rs1057516999 Chr.9: 105619995-105619995 on Build GRCh38
rs970553894 Chr.9: 105619823-105619823 on Build GRCh38
rs750176716 Chr.9: 105619989-105619989 on Build GRCh38
rs119464998 Chr.9: 105620001-105620001 on Build GRCh38
rs765709686 Chr.9: 105620111-105620111 on Build GRCh38
rs776723808 Chr.9: 105620076-105620076 on Build GRCh38
rs573229695 Chr.9: 105619768-105619768 on Build GRCh38
rs958948173 Chr.9: 105619862-105619862 on Build GRCh38
rs539089647 Chr.9: 105619935-105619935 on Build GRCh38
rs758841311 Chr.9: 105619943-105619943 on Build GRCh38
rs189299628 Chr.9: 105619852-105619852 on Build GRCh38
rs1048088668 Chr.9: 105620024-105620024 on Build GRCh38
rs180759073 Chr.9: 105619767-105619767 on Build GRCh38
rs371776972 Chr.9: 105620008-105620008 on Build GRCh38
rs549221329 Chr.9: 105619740-105619740 on Build GRCh38
rs775692665 Chr.9: 105619907-105619907 on Build GRCh38
rs569184532 Chr.9: 105620142-105620142 on Build GRCh38
rs149085844 Chr.9: 105619945-105619945 on Build GRCh38
rs374554922 Chr.9: 105619772-105619772 on Build GRCh38
rs990541478 Chr.9: 105620133-105620133 on Build GRCh38
rs942060927 Chr.9: 105619942-105619942 on Build GRCh38
rs776627363 Chr.9: 105619826-105619826 on Build GRCh38
rs770778747 Chr.9: 105620032-105620032 on Build GRCh38
rs958841363 Chr.9: 105620110-105620110 on Build GRCh38
rs768792475 Chr.9: 105620074-105620074 on Build GRCh38
rs368199437 Chr.9: 105619806-105619806 on Build GRCh38
rs754270605 Chr.9: 105619928-105619928 on Build GRCh38
rs145883833 Chr.9: 105619894-105619894 on Build GRCh38
rs764291359 Chr.9: 105619912-105619912 on Build GRCh38
rs750759323 Chr.9: 105619937-105619937 on Build GRCh38
rs756416366 Chr.9: 105619917-105619917 on Build GRCh38
rs760739538 Chr.9: 105619911-105619911 on Build GRCh38
rs1003989506 Chr.9: 105619784-105619784 on Build GRCh38
rs1021999352 Chr.9: 105620137-105620137 on Build GRCh38
rs990205460 Chr.9: 105619881-105619881 on Build GRCh38
rs530428813 Chr.9: 105619692-105619692 on Build GRCh38
rs762343906 Chr.9: 105619931-105619931 on Build GRCh38
rs201402850 Chr.9: 105619654-105619655 on Build GRCh38
rs749014759 Chr.9: 105620012-105620012 on Build GRCh38
rs768509805 Chr.9: 105619812-105619812 on Build GRCh38
rs747280605 Chr.9: 105620058-105620058 on Build GRCh38
rs772221503 Chr.9: 105619906-105619906 on Build GRCh38
rs886044528 Chr.9: 105620047-105620047 on Build GRCh38
rs781004059 Chr.9: 105619838-105619838 on Build GRCh38

View Full Product Details