Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 40223213-40223722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD20A2
Entrez Gene ID
441430
Gene Name
ankyrin repeat domain 20 family member A2
Gene Aliases
-
UniGene
Hs.632663
Gene Chromosome Location
Chr.9: 40223285-40282360 on Build GRCh38
Transcripts
NM_001012421.1 XM_011517877.2 XM_011517878.2 XM_011517879.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1490043 c.200A>G p.H67R Chr.9: 40223596-40223596 on Build GRCh38
116722 c.105G>T p.Q35H Chr.9: 40223501-40223501 on Build GRCh38
7328233 c.153C>G p.R51R Chr.9: 40223549-40223549 on Build GRCh38
3907487 c.183C>T p.D61D Chr.9: 40223579-40223579 on Build GRCh38
299705 c.31G>T p.G11C Chr.9: 40223427-40223427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200348657 Chr.9: 40223596-40223596 on Build GRCh38
rs776784579 Chr.9: 40223586-40223586 on Build GRCh38
rs755828388 Chr.9: 40223563-40223563 on Build GRCh38
rs755597137 Chr.9: 40223533-40223533 on Build GRCh38
rs761029255 Chr.9: 40223547-40223547 on Build GRCh38
rs749215832 Chr.9: 40223535-40223535 on Build GRCh38
rs754654814 Chr.9: 40223362-40223362 on Build GRCh38
rs952298294 Chr.9: 40223440-40223440 on Build GRCh38
rs192233891 Chr.9: 40223311-40223311 on Build GRCh38
rs745659187 Chr.9: 40223541-40223541 on Build GRCh38
rs576099039 Chr.9: 40223257-40223257 on Build GRCh38
rs757721360 Chr.9: 40223348-40223348 on Build GRCh38
rs753310002 Chr.9: 40223605-40223605 on Build GRCh38
rs75899060 Chr.9: 40223324-40223324 on Build GRCh38
rs563254670 Chr.9: 40223270-40223270 on Build GRCh38
rs200102177 Chr.9: 40223403-40223403 on Build GRCh38
rs756743289 Chr.9: 40223606-40223606 on Build GRCh38
rs777301271 Chr.9: 40223534-40223534 on Build GRCh38
rs769677519 Chr.9: 40223376-40223376 on Build GRCh38
rs537180839 Chr.9: 40223559-40223559 on Build GRCh38
rs764648779 Chr.9: 40223438-40223438 on Build GRCh38
rs769058424 Chr.9: 40223584-40223584 on Build GRCh38
rs748334582 Chr.9: 40223596-40223597 on Build GRCh38
rs541028622 Chr.9: 40223631-40223631 on Build GRCh38
rs775599549 Chr.9: 40223546-40223546 on Build GRCh38
rs781587220 Chr.9: 40223642-40223642 on Build GRCh38
rs757879213 Chr.9: 40223476-40223476 on Build GRCh38
rs77881930 Chr.9: 40223531-40223531 on Build GRCh38
rs529865515 Chr.9: 40223386-40223386 on Build GRCh38
rs754004482 Chr.9: 40223478-40223482 on Build GRCh38
rs558257066 Chr.9: 40223564-40223564 on Build GRCh38
rs754590877 Chr.9: 40223528-40223528 on Build GRCh38
rs901978252 Chr.9: 40223397-40223397 on Build GRCh38
rs752312776 Chr.9: 40223562-40223562 on Build GRCh38
rs576030596 Chr.9: 40223557-40223557 on Build GRCh38
rs781252587 Chr.9: 40223429-40223437 on Build GRCh38
rs548411696 Chr.9: 40223388-40223388 on Build GRCh38
rs534978018 Chr.9: 40223421-40223421 on Build GRCh38
rs764239844 Chr.9: 40223550-40223550 on Build GRCh38
rs1026567566 Chr.9: 40223416-40223416 on Build GRCh38
rs746814375 Chr.9: 40223622-40223622 on Build GRCh38
rs552265330 Chr.9: 40223408-40223408 on Build GRCh38
rs778749415 Chr.9: 40223579-40223579 on Build GRCh38
rs199914729 Chr.9: 40223536-40223536 on Build GRCh38
rs772235896 Chr.9: 40223605-40223605 on Build GRCh38
rs568417904 Chr.9: 40223514-40223514 on Build GRCh38
rs201683465 Chr.9: 40223504-40223504 on Build GRCh38
rs200874570 Chr.9: 40223443-40223443 on Build GRCh38
rs994108193 Chr.9: 40223406-40223406 on Build GRCh38
rs757300447 Chr.9: 40223381-40223383 on Build GRCh38
rs184426325 Chr.9: 40223319-40223319 on Build GRCh38
rs373509797 Chr.9: 40223472-40223472 on Build GRCh38
rs779470684 Chr.9: 40223353-40223353 on Build GRCh38
rs770285075 Chr.9: 40223591-40223591 on Build GRCh38
rs369158633 Chr.9: 40223434-40223434 on Build GRCh38
rs531056161 Chr.9: 40223405-40223405 on Build GRCh38
rs947033322 Chr.9: 40223347-40223347 on Build GRCh38
rs755005645 Chr.9: 40223523-40223524 on Build GRCh38
rs557549890 Chr.9: 40223532-40223532 on Build GRCh38
rs62537988 Chr.9: 40223610-40223610 on Build GRCh38
rs751974807 Chr.9: 40223597-40223597 on Build GRCh38
rs773568246 Chr.9: 40223592-40223592 on Build GRCh38
rs535613628 Chr.9: 40223522-40223522 on Build GRCh38
rs762014444 Chr.9: 40223590-40223590 on Build GRCh38
rs766814670 Chr.9: 40223561-40223561 on Build GRCh38
rs758430225 Chr.9: 40223619-40223619 on Build GRCh38
rs543018902 Chr.9: 40223300-40223300 on Build GRCh38
rs749270932 Chr.9: 40223382-40223382 on Build GRCh38
rs778145776 Chr.9: 40223604-40223605 on Build GRCh38
rs778982515 Chr.9: 40223578-40223579 on Build GRCh38
rs780695080 Chr.9: 40223364-40223364 on Build GRCh38
rs763248453 Chr.9: 40223560-40223560 on Build GRCh38
rs779982251 Chr.9: 40223621-40223621 on Build GRCh38
rs376153150 Chr.9: 40223366-40223366 on Build GRCh38
rs774142152 Chr.9: 40223537-40223537 on Build GRCh38
rs75596179 Chr.9: 40223582-40223582 on Build GRCh38
rs763744291 Chr.9: 40223599-40223599 on Build GRCh38
rs759795559 Chr.9: 40223392-40223392 on Build GRCh38
rs771743863 Chr.9: 40223580-40223580 on Build GRCh38
rs914530029 Chr.9: 40223343-40223343 on Build GRCh38
rs576817733 Chr.9: 40223567-40223567 on Build GRCh38
rs762396744 Chr.9: 40223551-40223551 on Build GRCh38
rs747231113 Chr.9: 40223583-40223583 on Build GRCh38
rs570858369 Chr.9: 40223409-40223409 on Build GRCh38
rs750298271 Chr.9: 40223618-40223618 on Build GRCh38
rs772300529 Chr.9: 40223544-40223544 on Build GRCh38
rs879493222 Chr.9: 40223652-40223652 on Build GRCh38
rs746643024 Chr.9: 40223354-40223354 on Build GRCh38
rs763553428 Chr.9: 40223594-40223594 on Build GRCh38
rs1006213000 Chr.9: 40223516-40223516 on Build GRCh38

View Full Product Details