Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 40226748-40227267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD20A2
Entrez Gene ID
441430
Gene Name
ankyrin repeat domain 20 family member A2
Gene Aliases
-
UniGene
Hs.632663
Gene Chromosome Location
Chr.9: 40223285-40282360 on Build GRCh38
Transcripts
NM_001012421.1 XM_011517877.2 XM_011517878.2 XM_011517879.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6410313 c.320C>T p.A107V Chr.9: 40227185-40227185 on Build GRCh38
3685764 c.366T>A p.H122Q Chr.9: 40227231-40227231 on Build GRCh38
3685763 c.347C>G p.A116G Chr.9: 40227212-40227212 on Build GRCh38
6374167 c.382C>T p.L128F Chr.9: 40227247-40227247 on Build GRCh38
4417101 c.315A>G p.I105M Chr.9: 40227018-40227018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529530672 Chr.9: 40227010-40227010 on Build GRCh38
rs776247315 Chr.9: 40227022-40227022 on Build GRCh38
rs769729537 Chr.9: 40226929-40226929 on Build GRCh38
rs537005509 Chr.9: 40226827-40226827 on Build GRCh38
rs561777986 Chr.9: 40227008-40227008 on Build GRCh38
rs4929513 Chr.9: 40227177-40227177 on Build GRCh38
rs747346429 Chr.9: 40226905-40226931 on Build GRCh38
rs983862591 Chr.9: 40227038-40227038 on Build GRCh38
rs763353720 Chr.9: 40226947-40226947 on Build GRCh38
rs951031105 Chr.9: 40227020-40227020 on Build GRCh38
rs774627216 Chr.9: 40226994-40226994 on Build GRCh38
rs61114609 Chr.9: 40227195-40227195 on Build GRCh38
rs771478591 Chr.9: 40226972-40226972 on Build GRCh38
rs748427212 Chr.9: 40226917-40226917 on Build GRCh38
rs761568016 Chr.9: 40227026-40227026 on Build GRCh38
rs528580340 Chr.9: 40226806-40226806 on Build GRCh38
rs62537990 Chr.9: 40227115-40227115 on Build GRCh38
rs773405137 Chr.9: 40226944-40226944 on Build GRCh38
rs546564086 Chr.9: 40226920-40226920 on Build GRCh38
rs918225638 Chr.9: 40226820-40226820 on Build GRCh38

View Full Product Details