Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 41953473-41952967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP3B
Entrez Gene ID
728577
Gene Name
contactin associated protein-like 3B
Gene Aliases
-
UniGene
Hs.521495
Gene Chromosome Location
Chr.9: 41891887-42129504 on Build GRCh38
Transcripts
NM_001201380.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5697348 c.1986C>T p.G662G Chr.9: 41953277-41953277 on Build GRCh38
6631951 c.2005G>A p.A669T Chr.9: 41953258-41953258 on Build GRCh38
5475293 c.2080+7G>A p.? Chr.9: 41953176-41953176 on Build GRCh38
4803305 c.1914C>T p.P638P Chr.9: 41953349-41953349 on Build GRCh38
6631950 c.1945G>A p.G649R Chr.9: 41953318-41953318 on Build GRCh38
View More

View Full Product Details