Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 68536853-68537352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM252
Entrez Gene ID
169693
Gene Name
transmembrane protein 252
Gene Aliases
C9orf71
UniGene
Hs.663056
Gene Chromosome Location
Chr.9: 68536580-68540867 on Build GRCh38
Transcripts
NM_153237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4163998 c.487G>A p.A163T Chr.9: 68537285-68537285 on Build GRCh38
6803949 c.421C>A p.P141T Chr.9: 68537351-68537351 on Build GRCh38
4847733 c.497G>C p.R166P Chr.9: 68537275-68537275 on Build GRCh38
3732505 c.476T>C p.I159T Chr.9: 68537296-68537296 on Build GRCh38
6866431 c.488C>T p.A163V Chr.9: 68537284-68537284 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs265075 Chr.9: 68537032-68537032 on Build GRCh38
rs147022534 Chr.9: 68537261-68537261 on Build GRCh38
rs1027742366 Chr.9: 68536985-68536985 on Build GRCh38
rs746346576 Chr.9: 68537282-68537282 on Build GRCh38
rs201004451 Chr.9: 68537317-68537317 on Build GRCh38
rs779836311 Chr.9: 68537233-68537233 on Build GRCh38
rs35122443 Chr.9: 68537077-68537078 on Build GRCh38
rs138207550 Chr.9: 68537289-68537289 on Build GRCh38
rs960846616 Chr.9: 68537011-68537011 on Build GRCh38
rs373105492 Chr.9: 68537286-68537286 on Build GRCh38
rs549073206 Chr.9: 68537003-68537003 on Build GRCh38
rs777294637 Chr.9: 68537053-68537053 on Build GRCh38
rs558683460 Chr.9: 68537056-68537056 on Build GRCh38
rs539308117 Chr.9: 68537109-68537109 on Build GRCh38
rs754908291 Chr.9: 68537248-68537248 on Build GRCh38
rs1023890394 Chr.9: 68537324-68537324 on Build GRCh38
rs141633262 Chr.9: 68537285-68537285 on Build GRCh38
rs1041958298 Chr.9: 68536919-68536919 on Build GRCh38
rs925062995 Chr.9: 68537115-68537115 on Build GRCh38
rs1019531486 Chr.9: 68537134-68537134 on Build GRCh38
rs774316383 Chr.9: 68537277-68537277 on Build GRCh38
rs554462807 Chr.9: 68537123-68537123 on Build GRCh38
rs373137990 Chr.9: 68537237-68537237 on Build GRCh38
rs781063883 Chr.9: 68537256-68537256 on Build GRCh38
rs935113971 Chr.9: 68537142-68537142 on Build GRCh38
rs1055990913 Chr.9: 68537284-68537284 on Build GRCh38
rs187233962 Chr.9: 68537042-68537042 on Build GRCh38
rs147880223 Chr.9: 68537276-68537276 on Build GRCh38
rs992438740 Chr.9: 68537014-68537014 on Build GRCh38
rs947763530 Chr.9: 68537084-68537084 on Build GRCh38
rs751039467 Chr.9: 68537314-68537314 on Build GRCh38
rs964755981 Chr.9: 68536884-68536884 on Build GRCh38
rs965249152 Chr.9: 68537148-68537148 on Build GRCh38
rs778116807 Chr.9: 68537328-68537328 on Build GRCh38
rs112283706 Chr.9: 68537122-68537122 on Build GRCh38
rs1018597358 Chr.9: 68536882-68536882 on Build GRCh38
rs41305537 Chr.9: 68537158-68537158 on Build GRCh38
rs996558544 Chr.9: 68536977-68536977 on Build GRCh38
rs1039644740 Chr.9: 68537201-68537201 on Build GRCh38
rs756389231 Chr.9: 68537273-68537273 on Build GRCh38
rs772902912 Chr.9: 68537296-68537296 on Build GRCh38
rs536221957 Chr.9: 68537178-68537178 on Build GRCh38
rs746716463 Chr.9: 68537035-68537035 on Build GRCh38
rs943847234 Chr.9: 68537189-68537189 on Build GRCh38
rs983528051 Chr.9: 68537009-68537009 on Build GRCh38
rs996200908 Chr.9: 68537270-68537270 on Build GRCh38
rs574570095 Chr.9: 68537188-68537188 on Build GRCh38
rs975247042 Chr.9: 68537157-68537157 on Build GRCh38
rs41312182 Chr.9: 68537108-68537108 on Build GRCh38
rs754882905 Chr.9: 68537315-68537315 on Build GRCh38
rs371615871 Chr.9: 68536877-68536877 on Build GRCh38
rs567386476 Chr.9: 68537010-68537010 on Build GRCh38
rs375725598 Chr.9: 68537275-68537275 on Build GRCh38
rs1008124605 Chr.9: 68536878-68536878 on Build GRCh38
rs202084316 Chr.9: 68537312-68537312 on Build GRCh38
rs762539176 Chr.9: 68537311-68537311 on Build GRCh38
rs111242031 Chr.9: 68537316-68537316 on Build GRCh38

View Full Product Details