Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 71865512-71866061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD17B
Entrez Gene ID
51104
Gene Name
abhydrolase domain containing 17B
Gene Aliases
C9orf77, CGI-67, FAM108B1
UniGene
Hs.380389
Gene Chromosome Location
Chr.9: 71859461-71911245 on Build GRCh38
Transcripts
NM_001025780.2 NM_016014.3 XM_006717134.3 XM_017014787.1 XM_017014788.1 XM_017014789.1 XM_017014790.1 XM_017014791.1

SNP Detail

SNP ID Chromosome Location
rs765252189 Chr.9: 71865556-71865556 on Build GRCh38
rs758580283 Chr.9: 71865584-71865584 on Build GRCh38
rs41287391 Chr.9: 71865722-71865722 on Build GRCh38
rs187603844 Chr.9: 71865721-71865721 on Build GRCh38
rs551726810 Chr.9: 71865582-71865582 on Build GRCh38
rs112801047 Chr.9: 71865926-71865926 on Build GRCh38
rs932129717 Chr.9: 71865774-71865774 on Build GRCh38
rs750517084 Chr.9: 71865583-71865583 on Build GRCh38
rs958584275 Chr.9: 71865818-71865818 on Build GRCh38
rs777357764 Chr.9: 71865882-71865882 on Build GRCh38
rs573545876 Chr.9: 71865953-71865953 on Build GRCh38
rs901999211 Chr.9: 71865548-71865548 on Build GRCh38
rs1003190648 Chr.9: 71865628-71865628 on Build GRCh38
rs940914715 Chr.9: 71865538-71865538 on Build GRCh38
rs984826116 Chr.9: 71865687-71865687 on Build GRCh38
rs538872290 Chr.9: 71865816-71865816 on Build GRCh38
rs1035555233 Chr.9: 71865649-71865649 on Build GRCh38
rs139715879 Chr.9: 71865554-71865554 on Build GRCh38
rs912831029 Chr.9: 71865775-71865775 on Build GRCh38
rs1040558443 Chr.9: 71865544-71865544 on Build GRCh38
rs1012823296 Chr.9: 71865986-71865986 on Build GRCh38
rs916673086 Chr.9: 71865673-71865673 on Build GRCh38
rs755275458 Chr.9: 71865725-71865725 on Build GRCh38
rs979572373 Chr.9: 71865689-71865689 on Build GRCh38
rs572416293 Chr.9: 71865922-71865922 on Build GRCh38
rs892840806 Chr.9: 71865583-71865583 on Build GRCh38
rs2030700 Chr.9: 71865807-71865807 on Build GRCh38
rs886842634 Chr.9: 71865902-71865902 on Build GRCh38
rs1041412267 Chr.9: 71865798-71865798 on Build GRCh38
rs962350358 Chr.9: 71865934-71865934 on Build GRCh38
rs952141544 Chr.9: 71865887-71865887 on Build GRCh38
rs556368711 Chr.9: 71865795-71865795 on Build GRCh38
rs57767774 Chr.9: 71865716-71865716 on Build GRCh38
rs1026750362 Chr.9: 71865857-71865857 on Build GRCh38
rs538915215 Chr.9: 71865817-71865817 on Build GRCh38
rs1025814033 Chr.9: 71865567-71865567 on Build GRCh38
rs1046842093 Chr.9: 71865879-71865879 on Build GRCh38
rs985265261 Chr.9: 71865900-71865900 on Build GRCh38
rs946714381 Chr.9: 71865659-71865659 on Build GRCh38
rs766503811 Chr.9: 71865652-71865652 on Build GRCh38
rs548875521 Chr.9: 71865868-71865869 on Build GRCh38
rs145307925 Chr.9: 71865626-71865626 on Build GRCh38
rs774564058 Chr.9: 71865827-71865827 on Build GRCh38
rs898203618 Chr.9: 71865833-71865833 on Build GRCh38
rs932198709 Chr.9: 71865982-71865982 on Build GRCh38
rs192613409 Chr.9: 71865943-71865943 on Build GRCh38
rs923523760 Chr.9: 71865963-71865963 on Build GRCh38
rs796316208 Chr.9: 71865592-71865592 on Build GRCh38
rs916540508 Chr.9: 71865656-71865656 on Build GRCh38
rs1021487216 Chr.9: 71865989-71865989 on Build GRCh38
rs377288125 Chr.9: 71865874-71865874 on Build GRCh38
rs558693025 Chr.9: 71865876-71865876 on Build GRCh38
rs2030701 Chr.9: 71865785-71865785 on Build GRCh38
rs914731067 Chr.9: 71866013-71866013 on Build GRCh38
rs1020347847 Chr.9: 71865615-71865615 on Build GRCh38
rs976394485 Chr.9: 71865955-71865955 on Build GRCh38
rs958616601 Chr.9: 71865973-71865973 on Build GRCh38

View Full Product Details