Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 71866732-71867242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD17B
Entrez Gene ID
51104
Gene Name
abhydrolase domain containing 17B
Gene Aliases
C9orf77, CGI-67, FAM108B1
UniGene
Hs.380389
Gene Chromosome Location
Chr.9: 71859461-71911245 on Build GRCh38
Transcripts
NM_001025780.2 NM_016014.3 XM_006717134.3 XM_017014787.1 XM_017014788.1 XM_017014789.1 XM_017014790.1 XM_017014791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5685043 c.694G>T p.G232W Chr.9: 71866960-71866960 on Build GRCh38
5411210 c.671C>T p.T224I Chr.9: 71866983-71866983 on Build GRCh38
6590877 c.732C>T p.L244L Chr.9: 71866922-71866922 on Build GRCh38
3657978 c.829T>C p.L277L Chr.9: 71866825-71866825 on Build GRCh38
6115658 c.732C>G p.L244L Chr.9: 71866922-71866922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975594682 Chr.9: 71867190-71867190 on Build GRCh38
rs763321044 Chr.9: 71867030-71867030 on Build GRCh38
rs376028645 Chr.9: 71866895-71866895 on Build GRCh38
rs112513232 Chr.9: 71867197-71867197 on Build GRCh38
rs915614887 Chr.9: 71867105-71867105 on Build GRCh38
rs986417591 Chr.9: 71866983-71866983 on Build GRCh38
rs372004707 Chr.9: 71866894-71866894 on Build GRCh38
rs867131302 Chr.9: 71867109-71867109 on Build GRCh38
rs563004240 Chr.9: 71867116-71867116 on Build GRCh38
rs76374020 Chr.9: 71866930-71866930 on Build GRCh38
rs112162987 Chr.9: 71867101-71867101 on Build GRCh38
rs900988700 Chr.9: 71867213-71867213 on Build GRCh38
rs776895458 Chr.9: 71866892-71866892 on Build GRCh38
rs554947530 Chr.9: 71866860-71866860 on Build GRCh38
rs113634289 Chr.9: 71867085-71867085 on Build GRCh38
rs766798354 Chr.9: 71867037-71867037 on Build GRCh38
rs948382143 Chr.9: 71867128-71867128 on Build GRCh38
rs758796707 Chr.9: 71866909-71866909 on Build GRCh38
rs904447151 Chr.9: 71867047-71867047 on Build GRCh38
rs751699852 Chr.9: 71867022-71867022 on Build GRCh38
rs760746048 Chr.9: 71866846-71866846 on Build GRCh38
rs766595687 Chr.9: 71866917-71866917 on Build GRCh38
rs565932773 Chr.9: 71866814-71866814 on Build GRCh38
rs773783520 Chr.9: 71867025-71867025 on Build GRCh38
rs776983268 Chr.9: 71867015-71867015 on Build GRCh38
rs200191910 Chr.9: 71866841-71866841 on Build GRCh38
rs755653161 Chr.9: 71866767-71866767 on Build GRCh38
rs756745882 Chr.9: 71866964-71866964 on Build GRCh38
rs377444855 Chr.9: 71866922-71866922 on Build GRCh38
rs746792403 Chr.9: 71867012-71867012 on Build GRCh38
rs879701112 Chr.9: 71866766-71866766 on Build GRCh38
rs368657844 Chr.9: 71867028-71867029 on Build GRCh38
rs201931606 Chr.9: 71867050-71867050 on Build GRCh38
rs756620259 Chr.9: 71867217-71867217 on Build GRCh38
rs7858585 Chr.9: 71867130-71867130 on Build GRCh38
rs373175774 Chr.9: 71866921-71866921 on Build GRCh38
rs771910241 Chr.9: 71866992-71866992 on Build GRCh38
rs757076067 Chr.9: 71867023-71867024 on Build GRCh38
rs143555778 Chr.9: 71867079-71867079 on Build GRCh38
rs775797358 Chr.9: 71866845-71866845 on Build GRCh38
rs369334690 Chr.9: 71866902-71866902 on Build GRCh38
rs1056835984 Chr.9: 71867009-71867009 on Build GRCh38
rs573608768 Chr.9: 71867044-71867044 on Build GRCh38
rs749495612 Chr.9: 71867042-71867043 on Build GRCh38
rs752418170 Chr.9: 71866919-71866919 on Build GRCh38
rs753788257 Chr.9: 71867034-71867034 on Build GRCh38
rs774243027 Chr.9: 71866829-71866829 on Build GRCh38
rs948573720 Chr.9: 71867029-71867029 on Build GRCh38
rs1031584915 Chr.9: 71867065-71867065 on Build GRCh38
rs576211447 Chr.9: 71867129-71867129 on Build GRCh38
rs778974377 Chr.9: 71866976-71866976 on Build GRCh38
rs369905464 Chr.9: 71867024-71867024 on Build GRCh38
rs770905338 Chr.9: 71866826-71866826 on Build GRCh38
rs766528806 Chr.9: 71866997-71866997 on Build GRCh38
rs1042842056 Chr.9: 71867038-71867038 on Build GRCh38
rs777225213 Chr.9: 71866806-71866806 on Build GRCh38
rs184492262 Chr.9: 71867156-71867156 on Build GRCh38
rs745722851 Chr.9: 71866987-71866987 on Build GRCh38
rs952905119 Chr.9: 71867203-71867203 on Build GRCh38

View Full Product Details