Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77647516-77648022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA14
Entrez Gene ID
9630
Gene Name
G protein subunit alpha 14
Gene Aliases
-
UniGene
Hs.657795
Gene Chromosome Location
Chr.9: 77423079-77648316 on Build GRCh38
Transcripts
NM_004297.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6244367 c.84C>A p.D28E Chr.9: 77647710-77647710 on Build GRCh38
1314941 c.28G>A p.E10K Chr.9: 77647766-77647766 on Build GRCh38
1110322 c.25G>A p.A9T Chr.9: 77647769-77647769 on Build GRCh38
4999330 c.56C>T p.A19V Chr.9: 77647738-77647738 on Build GRCh38
1208389 c.8G>A p.G3D Chr.9: 77647786-77647786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759027847 Chr.9: 77647648-77647648 on Build GRCh38
rs927258579 Chr.9: 77647841-77647841 on Build GRCh38
rs758409416 Chr.9: 77647692-77647692 on Build GRCh38
rs947340795 Chr.9: 77647750-77647750 on Build GRCh38
rs767195221 Chr.9: 77647655-77647655 on Build GRCh38
rs1022083782 Chr.9: 77647999-77647999 on Build GRCh38
rs998308242 Chr.9: 77647727-77647727 on Build GRCh38
rs752297329 Chr.9: 77647665-77647665 on Build GRCh38
rs762103572 Chr.9: 77647795-77647795 on Build GRCh38
rs763015311 Chr.9: 77647635-77647635 on Build GRCh38
rs773887644 Chr.9: 77647641-77647641 on Build GRCh38
rs569754038 Chr.9: 77647574-77647574 on Build GRCh38
rs745814997 Chr.9: 77647763-77647763 on Build GRCh38
rs897282909 Chr.9: 77647613-77647613 on Build GRCh38
rs750228033 Chr.9: 77647636-77647636 on Build GRCh38
rs969271823 Chr.9: 77647829-77647829 on Build GRCh38
rs927418796 Chr.9: 77647607-77647607 on Build GRCh38
rs1050042869 Chr.9: 77647572-77647572 on Build GRCh38
rs138686336 Chr.9: 77647697-77647697 on Build GRCh38
rs924241397 Chr.9: 77647989-77647989 on Build GRCh38
rs930164192 Chr.9: 77647893-77647893 on Build GRCh38
rs977054700 Chr.9: 77647915-77647915 on Build GRCh38
rs190995452 Chr.9: 77647546-77647546 on Build GRCh38
rs1034961597 Chr.9: 77647694-77647694 on Build GRCh38
rs757124524 Chr.9: 77647758-77647758 on Build GRCh38
rs938789180 Chr.9: 77647846-77647846 on Build GRCh38
rs768674028 Chr.9: 77647895-77647895 on Build GRCh38
rs144900584 Chr.9: 77647671-77647671 on Build GRCh38
rs765954210 Chr.9: 77647770-77647770 on Build GRCh38
rs943849774 Chr.9: 77647832-77647832 on Build GRCh38
rs374751150 Chr.9: 77647819-77647819 on Build GRCh38
rs548941450 Chr.9: 77647896-77647896 on Build GRCh38
rs751699547 Chr.9: 77647747-77647747 on Build GRCh38
rs775163253 Chr.9: 77647729-77647729 on Build GRCh38
rs771496591 Chr.9: 77647724-77647724 on Build GRCh38
rs1010731432 Chr.9: 77647996-77647996 on Build GRCh38
rs371558607 Chr.9: 77647589-77647589 on Build GRCh38
rs980112751 Chr.9: 77647830-77647830 on Build GRCh38
rs144744914 Chr.9: 77647568-77647568 on Build GRCh38
rs765788385 Chr.9: 77647676-77647676 on Build GRCh38
rs7033216 Chr.9: 77647581-77647581 on Build GRCh38
rs556776699 Chr.9: 77647878-77647878 on Build GRCh38
rs993293114 Chr.9: 77647868-77647868 on Build GRCh38
rs993842292 Chr.9: 77647611-77647611 on Build GRCh38
rs376031163 Chr.9: 77647967-77647967 on Build GRCh38
rs759758045 Chr.9: 77647842-77647842 on Build GRCh38
rs879863953 Chr.9: 77647569-77647569 on Build GRCh38
rs147981179 Chr.9: 77647938-77647938 on Build GRCh38
rs749143230 Chr.9: 77647706-77647706 on Build GRCh38
rs918510206 Chr.9: 77647889-77647889 on Build GRCh38
rs754441712 Chr.9: 77647945-77647945 on Build GRCh38
rs1018064446 Chr.9: 77647544-77647544 on Build GRCh38
rs746573983 Chr.9: 77647785-77647785 on Build GRCh38
rs777678411 Chr.9: 77647704-77647704 on Build GRCh38
rs185897814 Chr.9: 77648000-77648000 on Build GRCh38
rs140078098 Chr.9: 77647712-77647712 on Build GRCh38
rs750509298 Chr.9: 77647679-77647679 on Build GRCh38
rs758450822 Chr.9: 77647735-77647735 on Build GRCh38
rs141591381 Chr.9: 77647723-77647723 on Build GRCh38
rs756079982 Chr.9: 77647671-77647673 on Build GRCh38
rs760355213 Chr.9: 77647730-77647730 on Build GRCh38
rs1031212363 Chr.9: 77647553-77647553 on Build GRCh38
rs765537922 Chr.9: 77647797-77647797 on Build GRCh38
rs773728283 Chr.9: 77647799-77647799 on Build GRCh38
rs925951180 Chr.9: 77647774-77647774 on Build GRCh38
rs763084688 Chr.9: 77647660-77647660 on Build GRCh38
rs866268844 Chr.9: 77647907-77647907 on Build GRCh38
rs771574912 Chr.9: 77647766-77647766 on Build GRCh38
rs751673982 Chr.9: 77647833-77647833 on Build GRCh38
rs181066673 Chr.9: 77647909-77647909 on Build GRCh38
rs568792361 Chr.9: 77647898-77647898 on Build GRCh38
rs930257662 Chr.9: 77647622-77647622 on Build GRCh38
rs1057303089 Chr.9: 77647612-77647612 on Build GRCh38
rs776267674 Chr.9: 77647788-77647788 on Build GRCh38
rs773234193 Chr.9: 77647634-77647634 on Build GRCh38
rs754306577 Chr.9: 77647673-77647673 on Build GRCh38
rs757733279 Chr.9: 77647674-77647674 on Build GRCh38
rs766217642 Chr.9: 77647685-77647686 on Build GRCh38
rs754882574 Chr.9: 77647758-77647760 on Build GRCh38
rs766957756 Chr.9: 77647821-77647821 on Build GRCh38
rs766400790 Chr.9: 77647743-77647743 on Build GRCh38
rs942888312 Chr.9: 77647914-77647914 on Build GRCh38
rs150308184 Chr.9: 77647760-77647760 on Build GRCh38
rs765705506 Chr.9: 77647734-77647734 on Build GRCh38
rs769720441 Chr.9: 77647629-77647629 on Build GRCh38
rs569245471 Chr.9: 77647545-77647545 on Build GRCh38
rs753672787 Chr.9: 77647705-77647706 on Build GRCh38
rs537798179 Chr.9: 77647951-77647951 on Build GRCh38
rs755932543 Chr.9: 77647701-77647701 on Build GRCh38
rs896863191 Chr.9: 77647610-77647610 on Build GRCh38
rs781620877 Chr.9: 77647753-77647753 on Build GRCh38
rs576655122 Chr.9: 77647899-77647899 on Build GRCh38
rs747218320 Chr.9: 77647698-77647698 on Build GRCh38
rs565812769 Chr.9: 77647596-77647596 on Build GRCh38
rs534693963 Chr.9: 77647638-77647638 on Build GRCh38
rs545632262 Chr.9: 77647908-77647908 on Build GRCh38
rs143500751 Chr.9: 77647937-77647937 on Build GRCh38
rs1006543657 Chr.9: 77647536-77647536 on Build GRCh38
rs768290236 Chr.9: 77647787-77647787 on Build GRCh38
rs749092434 Chr.9: 77647756-77647756 on Build GRCh38
rs926035549 Chr.9: 77647556-77647556 on Build GRCh38
rs370899063 Chr.9: 77647731-77647731 on Build GRCh38
rs980060619 Chr.9: 77647595-77647595 on Build GRCh38
rs770980701 Chr.9: 77647708-77647708 on Build GRCh38
rs371439397 Chr.9: 77647917-77647917 on Build GRCh38

View Full Product Details