Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77720459-77720966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAQ
Entrez Gene ID
2776
Gene Name
G protein subunit alpha q
Gene Aliases
CMC1, G-ALPHA-q, GAQ, SWS
UniGene
Hs.269782
Gene Chromosome Location
Chr.9: 77716274-78031449 on Build GRCh38
Transcripts
NM_002072.4 XM_017014628.1

SNP Detail

SNP ID Chromosome Location
rs1033886410 Chr.9: 77720838-77720838 on Build GRCh38
rs180831863 Chr.9: 77720767-77720767 on Build GRCh38
rs867959406 Chr.9: 77720501-77720501 on Build GRCh38
rs151176741 Chr.9: 77720566-77720566 on Build GRCh38
rs918192322 Chr.9: 77720865-77720865 on Build GRCh38
rs941764813 Chr.9: 77720684-77720684 on Build GRCh38
rs547929961 Chr.9: 77720545-77720545 on Build GRCh38
rs765576034 Chr.9: 77720609-77720609 on Build GRCh38
rs574238631 Chr.9: 77720740-77720740 on Build GRCh38
rs763774267 Chr.9: 77720862-77720862 on Build GRCh38
rs536724620 Chr.9: 77720553-77720553 on Build GRCh38
rs878892684 Chr.9: 77720581-77720581 on Build GRCh38
rs979922289 Chr.9: 77720837-77720837 on Build GRCh38
rs867919099 Chr.9: 77720852-77720852 on Build GRCh38
rs1854326 Chr.9: 77720522-77720522 on Build GRCh38
rs887548109 Chr.9: 77720605-77720605 on Build GRCh38
rs959534596 Chr.9: 77720850-77720850 on Build GRCh38
rs941831053 Chr.9: 77720932-77720932 on Build GRCh38
rs142106028 Chr.9: 77720547-77720547 on Build GRCh38
rs554321056 Chr.9: 77720733-77720733 on Build GRCh38
rs543103476 Chr.9: 77720751-77720751 on Build GRCh38
rs968183711 Chr.9: 77720774-77720774 on Build GRCh38
rs938282579 Chr.9: 77720509-77720509 on Build GRCh38
rs908855178 Chr.9: 77720925-77720925 on Build GRCh38
rs140218242 Chr.9: 77720826-77720826 on Build GRCh38
rs891689065 Chr.9: 77720752-77720752 on Build GRCh38
rs1038893168 Chr.9: 77720705-77720705 on Build GRCh38
rs758650921 Chr.9: 77720736-77720736 on Build GRCh38
rs558769766 Chr.9: 77720637-77720637 on Build GRCh38
rs878907883 Chr.9: 77720818-77720818 on Build GRCh38
rs759018642 Chr.9: 77720514-77720514 on Build GRCh38
rs780389784 Chr.9: 77720768-77720768 on Build GRCh38
rs189599873 Chr.9: 77720568-77720568 on Build GRCh38
rs186763431 Chr.9: 77720829-77720829 on Build GRCh38
rs750630718 Chr.9: 77720649-77720649 on Build GRCh38
rs540860825 Chr.9: 77720706-77720706 on Build GRCh38
rs926766068 Chr.9: 77720507-77720507 on Build GRCh38
rs929515721 Chr.9: 77720903-77720903 on Build GRCh38
rs556644357 Chr.9: 77720554-77720554 on Build GRCh38
rs764641995 Chr.9: 77720637-77720637 on Build GRCh38
rs751735382 Chr.9: 77720924-77720924 on Build GRCh38
rs572170496 Chr.9: 77720667-77720667 on Build GRCh38
rs997424524 Chr.9: 77720735-77720736 on Build GRCh38

View Full Product Details