Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77721248-77721750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAQ
Entrez Gene ID
2776
Gene Name
G protein subunit alpha q
Gene Aliases
CMC1, G-ALPHA-q, GAQ, SWS
UniGene
Hs.269782
Gene Chromosome Location
Chr.9: 77716274-78031449 on Build GRCh38
Transcripts
NM_002072.4 XM_017014628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3658636 c.899G>A p.R300K Chr.9: 77721504-77721504 on Build GRCh38
123394 c.974A>T p.Y325F Chr.9: 77721429-77721429 on Build GRCh38
6960798 c.1025C>T p.A342V Chr.9: 77721378-77721378 on Build GRCh38
3658635 c.1018G>C p.V340L Chr.9: 77721385-77721385 on Build GRCh38
6672195 c.1012C>T p.R338C Chr.9: 77721391-77721391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775693911 Chr.9: 77721503-77721503 on Build GRCh38
rs1059530 Chr.9: 77721352-77721352 on Build GRCh38
rs149624211 Chr.9: 77721569-77721569 on Build GRCh38
rs759108484 Chr.9: 77721629-77721629 on Build GRCh38
rs1051567389 Chr.9: 77721297-77721297 on Build GRCh38
rs1025189626 Chr.9: 77721543-77721543 on Build GRCh38
rs774951302 Chr.9: 77721496-77721496 on Build GRCh38
rs771611377 Chr.9: 77721635-77721635 on Build GRCh38
rs1059529 Chr.9: 77721377-77721377 on Build GRCh38
rs936701973 Chr.9: 77721530-77721530 on Build GRCh38
rs755652968 Chr.9: 77721548-77721548 on Build GRCh38
rs760869735 Chr.9: 77721504-77721504 on Build GRCh38
rs554382469 Chr.9: 77721538-77721538 on Build GRCh38
rs376987002 Chr.9: 77721493-77721493 on Build GRCh38
rs751516475 Chr.9: 77721452-77721452 on Build GRCh38
rs183399236 Chr.9: 77721621-77721621 on Build GRCh38
rs1059532 Chr.9: 77721323-77721323 on Build GRCh38
rs541739421 Chr.9: 77721648-77721648 on Build GRCh38
rs776922380 Chr.9: 77721512-77721512 on Build GRCh38
rs748971807 Chr.9: 77721545-77721545 on Build GRCh38
rs1059533 Chr.9: 77721314-77721314 on Build GRCh38
rs778608199 Chr.9: 77721280-77721280 on Build GRCh38
rs574519710 Chr.9: 77721565-77721565 on Build GRCh38
rs576775511 Chr.9: 77721572-77721572 on Build GRCh38
rs959918799 Chr.9: 77721658-77721658 on Build GRCh38
rs1059534 Chr.9: 77721290-77721290 on Build GRCh38
rs777234154 Chr.9: 77721550-77721550 on Build GRCh38
rs746447398 Chr.9: 77721291-77721291 on Build GRCh38
rs772749242 Chr.9: 77721499-77721499 on Build GRCh38
rs762604216 Chr.9: 77721518-77721518 on Build GRCh38
rs185734369 Chr.9: 77721551-77721551 on Build GRCh38
rs954500251 Chr.9: 77721606-77721606 on Build GRCh38
rs558410927 Chr.9: 77721526-77721526 on Build GRCh38
rs760152072 Chr.9: 77721497-77721497 on Build GRCh38
rs545662450 Chr.9: 77721540-77721541 on Build GRCh38
rs780771168 Chr.9: 77721304-77721304 on Build GRCh38
rs752142063 Chr.9: 77721547-77721547 on Build GRCh38
rs945872879 Chr.9: 77721646-77721646 on Build GRCh38
rs773426924 Chr.9: 77721322-77721322 on Build GRCh38
rs543065799 Chr.9: 77721586-77721586 on Build GRCh38
rs1059528 Chr.9: 77721383-77721383 on Build GRCh38
rs757656161 Chr.9: 77721556-77721556 on Build GRCh38
rs558664347 Chr.9: 77721356-77721356 on Build GRCh38
rs766188046 Chr.9: 77721519-77721519 on Build GRCh38
rs752981660 Chr.9: 77721272-77721272 on Build GRCh38
rs769525825 Chr.9: 77721320-77721320 on Build GRCh38
rs369282648 Chr.9: 77721401-77721401 on Build GRCh38
rs561477377 Chr.9: 77721684-77721684 on Build GRCh38
rs1047392836 Chr.9: 77721589-77721589 on Build GRCh38
rs369796989 Chr.9: 77721303-77721303 on Build GRCh38
rs879087534 Chr.9: 77721577-77721577 on Build GRCh38
rs144945100 Chr.9: 77721479-77721479 on Build GRCh38
rs61736823 Chr.9: 77721386-77721386 on Build GRCh38
rs974407780 Chr.9: 77721600-77721600 on Build GRCh38
rs183713546 Chr.9: 77721446-77721446 on Build GRCh38
rs113500437 Chr.9: 77721559-77721559 on Build GRCh38
rs887440226 Chr.9: 77721599-77721599 on Build GRCh38
rs747826333 Chr.9: 77721311-77721311 on Build GRCh38
rs559066778 Chr.9: 77721584-77721584 on Build GRCh38
rs1059527 Chr.9: 77721394-77721394 on Build GRCh38
rs373212005 Chr.9: 77721374-77721374 on Build GRCh38
rs374303316 Chr.9: 77721506-77721506 on Build GRCh38
rs190985140 Chr.9: 77721576-77721576 on Build GRCh38
rs750275446 Chr.9: 77721282-77721282 on Build GRCh38
rs1059531 Chr.9: 77721338-77721338 on Build GRCh38
rs758215116 Chr.9: 77721422-77721422 on Build GRCh38
rs375995906 Chr.9: 77721289-77721289 on Build GRCh38
rs921580622 Chr.9: 77721603-77721603 on Build GRCh38
rs769315863 Chr.9: 77721486-77721486 on Build GRCh38
rs199754984 Chr.9: 77721464-77721464 on Build GRCh38
rs777546823 Chr.9: 77721492-77721492 on Build GRCh38
rs1013677517 Chr.9: 77721688-77721688 on Build GRCh38
rs397825484 Chr.9: 77721439-77721439 on Build GRCh38
rs764430308 Chr.9: 77721413-77721413 on Build GRCh38
rs775004834 Chr.9: 77721359-77721359 on Build GRCh38
rs777013529 Chr.9: 77721689-77721689 on Build GRCh38
rs758249489 Chr.9: 77721286-77721286 on Build GRCh38
rs778106531 Chr.9: 77721533-77721534 on Build GRCh38
rs190993985 Chr.9: 77721279-77721279 on Build GRCh38
rs372746608 Chr.9: 77721469-77721469 on Build GRCh38
rs1059525 Chr.9: 77721416-77721416 on Build GRCh38
rs751391950 Chr.9: 77721533-77721533 on Build GRCh38

View Full Product Details