Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 78236185-78236705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP78
Entrez Gene ID
84131
Gene Name
centrosomal protein 78
Gene Aliases
C9orf81, IP63
UniGene
Hs.374421
Gene Chromosome Location
Chr.9: 78236062-78278431 on Build GRCh38
Transcripts
NM_001098802.1 NM_032171.1 XM_005252263.4 XM_005252265.2 XM_005252266.4 XM_017015192.1 XM_017015193.1 XM_017015194.1 XM_017015195.1 XM_017015196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7189153 c.151C>T p.R51C Chr.9: 78236501-78236501 on Build GRCh38
4164038 c.98C>A p.A33D Chr.9: 78236448-78236448 on Build GRCh38
5160336 c.133T>C p.F45L Chr.9: 78236483-78236483 on Build GRCh38
3218475 c.157G>T p.V53L Chr.9: 78236507-78236507 on Build GRCh38
3218465 c.42_44delCTT p.F16delF Chr.9: 78236392-78236394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373622498 Chr.9: 78236603-78236603 on Build GRCh38
rs777863022 Chr.9: 78236403-78236403 on Build GRCh38
rs942527547 Chr.9: 78236242-78236242 on Build GRCh38
rs908462973 Chr.9: 78236525-78236525 on Build GRCh38
rs372654847 Chr.9: 78236476-78236476 on Build GRCh38
rs770486787 Chr.9: 78236578-78236578 on Build GRCh38
rs752556378 Chr.9: 78236636-78236636 on Build GRCh38
rs201121890 Chr.9: 78236439-78236439 on Build GRCh38
rs756323667 Chr.9: 78236402-78236402 on Build GRCh38
rs13292584 Chr.9: 78236611-78236611 on Build GRCh38
rs746720724 Chr.9: 78236241-78236241 on Build GRCh38
rs1028191296 Chr.9: 78236588-78236588 on Build GRCh38
rs546848947 Chr.9: 78236422-78236422 on Build GRCh38
rs764038233 Chr.9: 78236513-78236513 on Build GRCh38
rs763730469 Chr.9: 78236345-78236345 on Build GRCh38
rs769215485 Chr.9: 78236481-78236481 on Build GRCh38
rs760713104 Chr.9: 78236511-78236511 on Build GRCh38
rs534732604 Chr.9: 78236566-78236566 on Build GRCh38
rs775304883 Chr.9: 78236336-78236336 on Build GRCh38
rs753251020 Chr.9: 78236510-78236510 on Build GRCh38
rs995744807 Chr.9: 78236586-78236586 on Build GRCh38
rs767578924 Chr.9: 78236509-78236509 on Build GRCh38
rs11137567 Chr.9: 78236659-78236659 on Build GRCh38
rs749504811 Chr.9: 78236462-78236462 on Build GRCh38
rs878905366 Chr.9: 78236517-78236517 on Build GRCh38
rs751391396 Chr.9: 78236534-78236534 on Build GRCh38
rs373441923 Chr.9: 78236483-78236483 on Build GRCh38
rs182452644 Chr.9: 78236385-78236385 on Build GRCh38
rs375884026 Chr.9: 78236487-78236487 on Build GRCh38
rs936476107 Chr.9: 78236665-78236665 on Build GRCh38
rs573626737 Chr.9: 78236557-78236557 on Build GRCh38
rs752035535 Chr.9: 78236373-78236373 on Build GRCh38
rs1051765497 Chr.9: 78236315-78236315 on Build GRCh38
rs768713389 Chr.9: 78236472-78236472 on Build GRCh38
rs920677287 Chr.9: 78236569-78236569 on Build GRCh38
rs746861398 Chr.9: 78236470-78236470 on Build GRCh38
rs1023786396 Chr.9: 78236390-78236390 on Build GRCh38
rs975667081 Chr.9: 78236255-78236255 on Build GRCh38
rs1012885930 Chr.9: 78236207-78236207 on Build GRCh38
rs765352298 Chr.9: 78236497-78236497 on Build GRCh38
rs778003732 Chr.9: 78236456-78236456 on Build GRCh38
rs933920757 Chr.9: 78236288-78236288 on Build GRCh38
rs745619126 Chr.9: 78236407-78236407 on Build GRCh38
rs748284703 Chr.9: 78236395-78236395 on Build GRCh38
rs753236016 Chr.9: 78236516-78236516 on Build GRCh38
rs376672047 Chr.9: 78236549-78236549 on Build GRCh38
rs776323982 Chr.9: 78236346-78236346 on Build GRCh38
rs750515617 Chr.9: 78236627-78236627 on Build GRCh38
rs779196527 Chr.9: 78236527-78236527 on Build GRCh38
rs774031337 Chr.9: 78236325-78236325 on Build GRCh38
rs187080779 Chr.9: 78236409-78236409 on Build GRCh38
rs774048983 Chr.9: 78236580-78236580 on Build GRCh38
rs564286061 Chr.9: 78236507-78236507 on Build GRCh38
rs760184319 Chr.9: 78236340-78236340 on Build GRCh38
rs772758636 Chr.9: 78236430-78236430 on Build GRCh38
rs549492461 Chr.9: 78236233-78236233 on Build GRCh38
rs776271026 Chr.9: 78236426-78236426 on Build GRCh38
rs907227210 Chr.9: 78236536-78236536 on Build GRCh38
rs758728798 Chr.9: 78236632-78236632 on Build GRCh38
rs201790064 Chr.9: 78236584-78236584 on Build GRCh38
rs765143446 Chr.9: 78236392-78236394 on Build GRCh38
rs373992755 Chr.9: 78236522-78236522 on Build GRCh38
rs748336278 Chr.9: 78236316-78236316 on Build GRCh38
rs761036809 Chr.9: 78236598-78236598 on Build GRCh38
rs765043440 Chr.9: 78236355-78236355 on Build GRCh38
rs761538280 Chr.9: 78236351-78236351 on Build GRCh38
rs775862101 Chr.9: 78236593-78236593 on Build GRCh38
rs777834985 Chr.9: 78236652-78236652 on Build GRCh38
rs768766296 Chr.9: 78236425-78236425 on Build GRCh38
rs757542494 Chr.9: 78236465-78236465 on Build GRCh38
rs774692436 Chr.9: 78236498-78236498 on Build GRCh38
rs768077721 Chr.9: 78236444-78236444 on Build GRCh38
rs922502016 Chr.9: 78236283-78236283 on Build GRCh38
rs369765677 Chr.9: 78236519-78236519 on Build GRCh38
rs777091174 Chr.9: 78236560-78236560 on Build GRCh38
rs766857594 Chr.9: 78236363-78236363 on Build GRCh38
rs754874101 Chr.9: 78236544-78236544 on Build GRCh38
rs757517647 Chr.9: 78236543-78236547 on Build GRCh38
rs200483715 Chr.9: 78236453-78236453 on Build GRCh38
rs906774238 Chr.9: 78236400-78236400 on Build GRCh38
rs945109292 Chr.9: 78236406-78236406 on Build GRCh38
rs750799292 Chr.9: 78236357-78236357 on Build GRCh38
rs781069275 Chr.9: 78236446-78236446 on Build GRCh38
rs756200672 Chr.9: 78236454-78236454 on Build GRCh38
rs781331158 Chr.9: 78236593-78236593 on Build GRCh38
rs772816803 Chr.9: 78236482-78236482 on Build GRCh38
rs189860505 Chr.9: 78236630-78236630 on Build GRCh38
rs781225406 Chr.9: 78236392-78236392 on Build GRCh38
rs759316369 Chr.9: 78236329-78236329 on Build GRCh38
rs10867166 Chr.9: 78236434-78236434 on Build GRCh38
rs751667495 Chr.9: 78236473-78236473 on Build GRCh38
rs764920634 Chr.9: 78236429-78236429 on Build GRCh38
rs781088538 Chr.9: 78236634-78236634 on Build GRCh38
rs748836436 Chr.9: 78236571-78236571 on Build GRCh38
rs956451098 Chr.9: 78236205-78236205 on Build GRCh38
rs752002017 Chr.9: 78236440-78236440 on Build GRCh38
rs779694885 Chr.9: 78236466-78236466 on Build GRCh38
rs899263613 Chr.9: 78236349-78236349 on Build GRCh38
rs770049121 Chr.9: 78236317-78236317 on Build GRCh38
rs756227232 Chr.9: 78236652-78236653 on Build GRCh38
rs758699168 Chr.9: 78236361-78236361 on Build GRCh38
rs201300510 Chr.9: 78236623-78236623 on Build GRCh38
rs755462996 Chr.9: 78236443-78236443 on Build GRCh38
rs115171840 Chr.9: 78236227-78236227 on Build GRCh38
rs781349320 Chr.9: 78236314-78236314 on Build GRCh38
rs973947175 Chr.9: 78236328-78236328 on Build GRCh38
rs35540488 Chr.9: 78236677-78236678 on Build GRCh38
rs940677343 Chr.9: 78236533-78236533 on Build GRCh38
rs762015609 Chr.9: 78236616-78236616 on Build GRCh38
rs879332654 Chr.9: 78236307-78236307 on Build GRCh38
rs761485253 Chr.9: 78236428-78236428 on Build GRCh38
rs755940154 Chr.9: 78236637-78236637 on Build GRCh38
rs770673349 Chr.9: 78236489-78236489 on Build GRCh38
rs371076007 Chr.9: 78236591-78236591 on Build GRCh38
rs536688125 Chr.9: 78236477-78236477 on Build GRCh38

View Full Product Details