Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 81949024-81949518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA31D3
Entrez Gene ID
389762
Gene Name
SPATA31 subfamily D member 3
Gene Aliases
FAM75D3
UniGene
Hs.522246
Gene Chromosome Location
Chr.9: 81943500-81950094 on Build GRCh38
Transcripts
NM_207416.2

SNP Detail

SNP ID Chromosome Location
rs886106433 Chr.9: 81949294-81949294 on Build GRCh38
rs766537529 Chr.9: 81949145-81949145 on Build GRCh38
rs563437752 Chr.9: 81949046-81949046 on Build GRCh38
rs900146007 Chr.9: 81949131-81949131 on Build GRCh38
rs148192526 Chr.9: 81949299-81949299 on Build GRCh38
rs182467412 Chr.9: 81949356-81949356 on Build GRCh38
rs879017049 Chr.9: 81949336-81949336 on Build GRCh38
rs559946896 Chr.9: 81949183-81949183 on Build GRCh38
rs189130041 Chr.9: 81949407-81949407 on Build GRCh38
rs891265993 Chr.9: 81949241-81949241 on Build GRCh38
rs2782405 Chr.9: 81949195-81949195 on Build GRCh38
rs1012025276 Chr.9: 81949449-81949449 on Build GRCh38
rs542489645 Chr.9: 81949320-81949320 on Build GRCh38
rs1047351619 Chr.9: 81949428-81949428 on Build GRCh38
rs545345794 Chr.9: 81949429-81949429 on Build GRCh38
rs567316741 Chr.9: 81949432-81949432 on Build GRCh38
rs202023903 Chr.9: 81949146-81949146 on Build GRCh38
rs747000117 Chr.9: 81949375-81949375 on Build GRCh38
rs1035192733 Chr.9: 81949300-81949300 on Build GRCh38
rs559603743 Chr.9: 81949364-81949364 on Build GRCh38
rs979322685 Chr.9: 81949351-81949351 on Build GRCh38
rs144862035 Chr.9: 81949193-81949193 on Build GRCh38
rs771963958 Chr.9: 81949104-81949104 on Build GRCh38
rs537713394 Chr.9: 81949196-81949196 on Build GRCh38
rs746788933 Chr.9: 81949177-81949177 on Build GRCh38
rs376706713 Chr.9: 81949350-81949350 on Build GRCh38
rs990722191 Chr.9: 81949188-81949188 on Build GRCh38
rs983686723 Chr.9: 81949168-81949168 on Build GRCh38
rs575185943 Chr.9: 81949302-81949302 on Build GRCh38
rs1051311184 Chr.9: 81949099-81949099 on Build GRCh38
rs942881416 Chr.9: 81949254-81949254 on Build GRCh38
rs924027193 Chr.9: 81949208-81949208 on Build GRCh38
rs546533057 Chr.9: 81949475-81949475 on Build GRCh38
rs994496851 Chr.9: 81949289-81949289 on Build GRCh38
rs935392840 Chr.9: 81949217-81949217 on Build GRCh38
rs1027393497 Chr.9: 81949159-81949159 on Build GRCh38
rs549029395 Chr.9: 81949202-81949202 on Build GRCh38
rs547554443 Chr.9: 81949171-81949171 on Build GRCh38
rs1023493438 Chr.9: 81949328-81949328 on Build GRCh38
rs923933097 Chr.9: 81949354-81949354 on Build GRCh38
rs552788646 Chr.9: 81949258-81949258 on Build GRCh38
rs549690588 Chr.9: 81949427-81949427 on Build GRCh38
rs898779193 Chr.9: 81949261-81949261 on Build GRCh38
rs11795190 Chr.9: 81949343-81949343 on Build GRCh38
rs1018551203 Chr.9: 81949116-81949116 on Build GRCh38
rs967979099 Chr.9: 81949342-81949342 on Build GRCh38
rs538462798 Chr.9: 81949293-81949293 on Build GRCh38
rs138977703 Chr.9: 81949134-81949134 on Build GRCh38
rs529372049 Chr.9: 81949139-81949139 on Build GRCh38
rs868663519 Chr.9: 81949073-81949073 on Build GRCh38
rs886076234 Chr.9: 81949430-81949430 on Build GRCh38
rs561778160 Chr.9: 81949323-81949323 on Build GRCh38
rs780214099 Chr.9: 81949388-81949388 on Build GRCh38
rs1034820480 Chr.9: 81949175-81949175 on Build GRCh38
rs565134715 Chr.9: 81949439-81949439 on Build GRCh38
rs967865029 Chr.9: 81949467-81949467 on Build GRCh38
rs979437386 Chr.9: 81949206-81949206 on Build GRCh38
rs1887453 Chr.9: 81949121-81949121 on Build GRCh38
rs896252268 Chr.9: 81949447-81949447 on Build GRCh38
rs528077264 Chr.9: 81949474-81949474 on Build GRCh38
rs960545395 Chr.9: 81949308-81949308 on Build GRCh38
rs920171171 Chr.9: 81949417-81949417 on Build GRCh38
rs641486 Chr.9: 81949237-81949237 on Build GRCh38
rs543425935 Chr.9: 81949120-81949120 on Build GRCh38
rs935491909 Chr.9: 81949086-81949086 on Build GRCh38
rs541372310 Chr.9: 81949357-81949357 on Build GRCh38
rs755277924 Chr.9: 81949248-81949248 on Build GRCh38
rs563851740 Chr.9: 81949385-81949385 on Build GRCh38
rs575153602 Chr.9: 81949100-81949100 on Build GRCh38
rs367833743 Chr.9: 81949373-81949373 on Build GRCh38
rs9644989 Chr.9: 81949203-81949203 on Build GRCh38
rs1031354876 Chr.9: 81949493-81949493 on Build GRCh38
rs950529299 Chr.9: 81949164-81949164 on Build GRCh38
rs1039931078 Chr.9: 81949412-81949412 on Build GRCh38
rs577619267 Chr.9: 81949292-81949292 on Build GRCh38
rs142232161 Chr.9: 81949184-81949184 on Build GRCh38

View Full Product Details