Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 81991047-81991542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA31D1
Entrez Gene ID
389763
Gene Name
SPATA31 subfamily D member 1
Gene Aliases
FAM75D1
UniGene
Hs.209545
Gene Chromosome Location
Chr.9: 81987404-81995256 on Build GRCh38
Transcripts
NM_001001670.2 XM_017014710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4895970 c.730C>G p.P244A Chr.9: 81991200-81991200 on Build GRCh38
754094 c.696G>A p.K232K Chr.9: 81991166-81991166 on Build GRCh38
4609888 c.581C>A p.P194Q Chr.9: 81991051-81991051 on Build GRCh38
3658857 c.737C>T p.P246L Chr.9: 81991207-81991207 on Build GRCh38
3848905 c.582A>C p.P194P Chr.9: 81991052-81991052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369205312 Chr.9: 81991473-81991473 on Build GRCh38
rs747823541 Chr.9: 81991322-81991322 on Build GRCh38
rs779182971 Chr.9: 81991440-81991440 on Build GRCh38
rs770537333 Chr.9: 81991518-81991522 on Build GRCh38
rs765399068 Chr.9: 81991075-81991075 on Build GRCh38
rs763327408 Chr.9: 81991453-81991454 on Build GRCh38
rs752368118 Chr.9: 81991353-81991353 on Build GRCh38
rs766275420 Chr.9: 81991348-81991348 on Build GRCh38
rs747027613 Chr.9: 81991451-81991451 on Build GRCh38
rs968478246 Chr.9: 81991072-81991072 on Build GRCh38
rs534939187 Chr.9: 81991259-81991259 on Build GRCh38
rs745318325 Chr.9: 81991121-81991121 on Build GRCh38
rs201322548 Chr.9: 81991254-81991254 on Build GRCh38
rs752085906 Chr.9: 81991511-81991511 on Build GRCh38
rs775126000 Chr.9: 81991373-81991373 on Build GRCh38
rs751484980 Chr.9: 81991350-81991350 on Build GRCh38
rs776964194 Chr.9: 81991261-81991261 on Build GRCh38
rs752950305 Chr.9: 81991294-81991294 on Build GRCh38
rs755815174 Chr.9: 81991097-81991097 on Build GRCh38
rs767131278 Chr.9: 81991426-81991426 on Build GRCh38
rs1011110966 Chr.9: 81991311-81991311 on Build GRCh38
rs879070901 Chr.9: 81991183-81991183 on Build GRCh38
rs767714129 Chr.9: 81991284-81991284 on Build GRCh38
rs374887411 Chr.9: 81991495-81991495 on Build GRCh38
rs779659652 Chr.9: 81991234-81991234 on Build GRCh38
rs939444123 Chr.9: 81991467-81991467 on Build GRCh38
rs746728707 Chr.9: 81991137-81991137 on Build GRCh38
rs746379227 Chr.9: 81991444-81991444 on Build GRCh38
rs528626632 Chr.9: 81991499-81991499 on Build GRCh38
rs766502040 Chr.9: 81991188-81991188 on Build GRCh38
rs776726585 Chr.9: 81991392-81991392 on Build GRCh38
rs766442011 Chr.9: 81991087-81991087 on Build GRCh38
rs959649850 Chr.9: 81991298-81991298 on Build GRCh38
rs376936477 Chr.9: 81991186-81991186 on Build GRCh38
rs749219893 Chr.9: 81991108-81991108 on Build GRCh38
rs770365086 Chr.9: 81991179-81991179 on Build GRCh38
rs373731758 Chr.9: 81991089-81991089 on Build GRCh38
rs538227393 Chr.9: 81991297-81991297 on Build GRCh38
rs753572962 Chr.9: 81991206-81991206 on Build GRCh38
rs762445757 Chr.9: 81991073-81991073 on Build GRCh38
rs192256999 Chr.9: 81991224-81991224 on Build GRCh38
rs369239362 Chr.9: 81991498-81991498 on Build GRCh38
rs773857998 Chr.9: 81991180-81991180 on Build GRCh38
rs192666773 Chr.9: 81991489-81991489 on Build GRCh38
rs750585491 Chr.9: 81991080-81991080 on Build GRCh38
rs201304983 Chr.9: 81991305-81991305 on Build GRCh38
rs527931753 Chr.9: 81991475-81991475 on Build GRCh38
rs376216540 Chr.9: 81991332-81991332 on Build GRCh38
rs200251028 Chr.9: 81991514-81991514 on Build GRCh38
rs780503248 Chr.9: 81991505-81991505 on Build GRCh38
rs777581939 Chr.9: 81991358-81991358 on Build GRCh38
rs144337990 Chr.9: 81991342-81991342 on Build GRCh38
rs113761190 Chr.9: 81991336-81991336 on Build GRCh38
rs758771158 Chr.9: 81991504-81991504 on Build GRCh38
rs770260787 Chr.9: 81991323-81991323 on Build GRCh38
rs779424406 Chr.9: 81991368-81991368 on Build GRCh38
rs746633707 Chr.9: 81991236-81991236 on Build GRCh38
rs759533865 Chr.9: 81991482-81991482 on Build GRCh38
rs906576821 Chr.9: 81991443-81991443 on Build GRCh38
rs1051701953 Chr.9: 81991171-81991171 on Build GRCh38
rs201295400 Chr.9: 81991508-81991508 on Build GRCh38
rs779413636 Chr.9: 81991313-81991313 on Build GRCh38
rs770312079 Chr.9: 81991247-81991247 on Build GRCh38
rs866975830 Chr.9: 81991244-81991244 on Build GRCh38
rs778836622 Chr.9: 81991116-81991116 on Build GRCh38
rs1031425866 Chr.9: 81991516-81991516 on Build GRCh38
rs536489449 Chr.9: 81991326-81991326 on Build GRCh38
rs773609357 Chr.9: 81991438-81991438 on Build GRCh38
rs370859354 Chr.9: 81991335-81991335 on Build GRCh38
rs749745831 Chr.9: 81991327-81991327 on Build GRCh38
rs184877684 Chr.9: 81991420-81991420 on Build GRCh38
rs771411463 Chr.9: 81991328-81991328 on Build GRCh38
rs577988203 Chr.9: 81991387-81991387 on Build GRCh38
rs757244629 Chr.9: 81991115-81991115 on Build GRCh38
rs370991238 Chr.9: 81991272-81991272 on Build GRCh38
rs933118806 Chr.9: 81991170-81991170 on Build GRCh38
rs781428577 Chr.9: 81991461-81991461 on Build GRCh38
rs755897741 Chr.9: 81991354-81991354 on Build GRCh38
rs189290500 Chr.9: 81991428-81991428 on Build GRCh38
rs761008316 Chr.9: 81991506-81991508 on Build GRCh38
rs759549394 Chr.9: 81991283-81991283 on Build GRCh38
rs372238180 Chr.9: 81991362-81991362 on Build GRCh38
rs766112490 Chr.9: 81991405-81991405 on Build GRCh38
rs755688898 Chr.9: 81991427-81991427 on Build GRCh38
rs377083125 Chr.9: 81991394-81991394 on Build GRCh38
rs748697344 Chr.9: 81991300-81991300 on Build GRCh38
rs750691392 Chr.9: 81991501-81991501 on Build GRCh38
rs780556657 Chr.9: 81991449-81991449 on Build GRCh38
rs757616539 Chr.9: 81991433-81991433 on Build GRCh38
rs1004081601 Chr.9: 81991256-81991256 on Build GRCh38
rs779250759 Chr.9: 81991265-81991265 on Build GRCh38
rs546849459 Chr.9: 81991222-81991222 on Build GRCh38
rs557466931 Chr.9: 81991445-81991445 on Build GRCh38
rs758562392 Chr.9: 81991082-81991082 on Build GRCh38
rs765068419 Chr.9: 81991304-81991304 on Build GRCh38
rs763784115 Chr.9: 81991202-81991202 on Build GRCh38
rs202197407 Chr.9: 81991351-81991351 on Build GRCh38
rs771646286 Chr.9: 81991125-81991125 on Build GRCh38
rs367809394 Chr.9: 81991360-81991360 on Build GRCh38
rs768409615 Chr.9: 81991241-81991241 on Build GRCh38
rs568845059 Chr.9: 81991319-81991319 on Build GRCh38
rs191924906 Chr.9: 81991388-81991388 on Build GRCh38
rs778785302 Chr.9: 81991221-81991221 on Build GRCh38
rs369682294 Chr.9: 81991070-81991070 on Build GRCh38
rs753454037 Chr.9: 81991430-81991430 on Build GRCh38
rs200253689 Chr.9: 81991242-81991242 on Build GRCh38
rs372453419 Chr.9: 81991246-81991246 on Build GRCh38
rs182874647 Chr.9: 81991343-81991343 on Build GRCh38
rs561290889 Chr.9: 81991178-81991178 on Build GRCh38
rs373603059 Chr.9: 81991359-81991359 on Build GRCh38
rs748369357 Chr.9: 81991470-81991470 on Build GRCh38
rs774959316 Chr.9: 81991133-81991133 on Build GRCh38
rs187210922 Chr.9: 81991369-81991369 on Build GRCh38
rs777116696 Chr.9: 81991173-81991173 on Build GRCh38
rs924330965 Chr.9: 81991330-81991330 on Build GRCh38
rs1033943946 Chr.9: 81991264-81991264 on Build GRCh38
rs200478585 Chr.9: 81991208-81991208 on Build GRCh38
rs377471076 Chr.9: 81991318-81991318 on Build GRCh38
rs368251732 Chr.9: 81991385-81991385 on Build GRCh38
rs772105230 Chr.9: 81991372-81991372 on Build GRCh38
rs772271242 Chr.9: 81991332-81991332 on Build GRCh38
rs755029847 Chr.9: 81991518-81991518 on Build GRCh38
rs759432930 Chr.9: 81991424-81991424 on Build GRCh38
rs777671720 Chr.9: 81991104-81991104 on Build GRCh38
rs751708255 Chr.9: 81991189-81991189 on Build GRCh38
rs762394418 Chr.9: 81991176-81991176 on Build GRCh38
rs369218974 Chr.9: 81991190-81991190 on Build GRCh38
rs768876074 Chr.9: 81991146-81991146 on Build GRCh38

View Full Product Details