Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 81992215-81992723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA31D1
Entrez Gene ID
389763
Gene Name
SPATA31 subfamily D member 1
Gene Aliases
FAM75D1
UniGene
Hs.209545
Gene Chromosome Location
Chr.9: 81987404-81995256 on Build GRCh38
Transcripts
NM_001001670.2 XM_017014710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5091760 c.2070C>G p.H690Q Chr.9: 81992540-81992540 on Build GRCh38
6788231 c.1803C>A p.I601I Chr.9: 81992273-81992273 on Build GRCh38
6190729 c.1755A>T p.Q585H Chr.9: 81992225-81992225 on Build GRCh38
6116184 c.2128C>T p.L710L Chr.9: 81992598-81992598 on Build GRCh38
7000702 c.1774C>A p.L592I Chr.9: 81992244-81992244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201182137 Chr.9: 81992376-81992376 on Build GRCh38
rs758289804 Chr.9: 81992357-81992357 on Build GRCh38
rs762149951 Chr.9: 81992484-81992484 on Build GRCh38
rs745710584 Chr.9: 81992657-81992657 on Build GRCh38
rs754763558 Chr.9: 81992491-81992491 on Build GRCh38
rs553658681 Chr.9: 81992580-81992580 on Build GRCh38
rs907560592 Chr.9: 81992674-81992674 on Build GRCh38
rs760157715 Chr.9: 81992402-81992402 on Build GRCh38
rs774576065 Chr.9: 81992698-81992698 on Build GRCh38
rs774838223 Chr.9: 81992468-81992468 on Build GRCh38
rs201473485 Chr.9: 81992303-81992303 on Build GRCh38
rs752130307 Chr.9: 81992325-81992325 on Build GRCh38
rs199964077 Chr.9: 81992285-81992285 on Build GRCh38
rs558976705 Chr.9: 81992588-81992588 on Build GRCh38
rs141219530 Chr.9: 81992431-81992431 on Build GRCh38
rs370865376 Chr.9: 81992562-81992562 on Build GRCh38
rs772371244 Chr.9: 81992555-81992555 on Build GRCh38
rs751531699 Chr.9: 81992273-81992273 on Build GRCh38
rs527382695 Chr.9: 81992439-81992439 on Build GRCh38
rs761518143 Chr.9: 81992313-81992313 on Build GRCh38
rs142280401 Chr.9: 81992443-81992443 on Build GRCh38
rs752509724 Chr.9: 81992516-81992516 on Build GRCh38
rs754991223 Chr.9: 81992326-81992326 on Build GRCh38
rs200176100 Chr.9: 81992243-81992243 on Build GRCh38
rs369084497 Chr.9: 81992696-81992696 on Build GRCh38
rs769372029 Chr.9: 81992379-81992379 on Build GRCh38
rs374159797 Chr.9: 81992678-81992678 on Build GRCh38
rs754105524 Chr.9: 81992479-81992479 on Build GRCh38
rs750073015 Chr.9: 81992661-81992661 on Build GRCh38
rs748647179 Chr.9: 81992681-81992681 on Build GRCh38
rs768840052 Chr.9: 81992564-81992564 on Build GRCh38
rs964208834 Chr.9: 81992648-81992648 on Build GRCh38
rs771134839 Chr.9: 81992324-81992324 on Build GRCh38
rs374596895 Chr.9: 81992548-81992548 on Build GRCh38
rs368739177 Chr.9: 81992424-81992424 on Build GRCh38
rs183342985 Chr.9: 81992660-81992660 on Build GRCh38
rs554676830 Chr.9: 81992252-81992252 on Build GRCh38
rs140486017 Chr.9: 81992493-81992493 on Build GRCh38
rs371164738 Chr.9: 81992267-81992267 on Build GRCh38
rs754158778 Chr.9: 81992435-81992435 on Build GRCh38
rs747003329 Chr.9: 81992364-81992364 on Build GRCh38
rs186455044 Chr.9: 81992466-81992466 on Build GRCh38
rs374319352 Chr.9: 81992581-81992581 on Build GRCh38
rs544594797 Chr.9: 81992338-81992338 on Build GRCh38
rs763969001 Chr.9: 81992659-81992659 on Build GRCh38
rs763316004 Chr.9: 81992321-81992321 on Build GRCh38
rs199808243 Chr.9: 81992620-81992620 on Build GRCh38
rs370390438 Chr.9: 81992291-81992291 on Build GRCh38
rs747405552 Chr.9: 81992310-81992310 on Build GRCh38
rs1000735265 Chr.9: 81992608-81992608 on Build GRCh38
rs765628606 Chr.9: 81992256-81992256 on Build GRCh38
rs370707399 Chr.9: 81992352-81992352 on Build GRCh38
rs768044363 Chr.9: 81992372-81992372 on Build GRCh38
rs778459117 Chr.9: 81992545-81992545 on Build GRCh38
rs781224286 Chr.9: 81992279-81992279 on Build GRCh38
rs367867592 Chr.9: 81992609-81992609 on Build GRCh38
rs752965070 Chr.9: 81992335-81992335 on Build GRCh38
rs764756162 Chr.9: 81992451-81992451 on Build GRCh38
rs200547091 Chr.9: 81992249-81992249 on Build GRCh38
rs755591044 Chr.9: 81992519-81992519 on Build GRCh38
rs374174863 Chr.9: 81992611-81992611 on Build GRCh38
rs187675943 Chr.9: 81992663-81992663 on Build GRCh38
rs769216202 Chr.9: 81992624-81992624 on Build GRCh38
rs151266667 Chr.9: 81992489-81992489 on Build GRCh38
rs558888712 Chr.9: 81992311-81992311 on Build GRCh38
rs745842429 Chr.9: 81992240-81992240 on Build GRCh38
rs748980110 Chr.9: 81992464-81992464 on Build GRCh38
rs777773457 Chr.9: 81992692-81992692 on Build GRCh38
rs752684622 Chr.9: 81992498-81992498 on Build GRCh38
rs756739364 Chr.9: 81992610-81992610 on Build GRCh38
rs780620369 Chr.9: 81992616-81992616 on Build GRCh38
rs776364895 Chr.9: 81992253-81992253 on Build GRCh38
rs369669501 Chr.9: 81992437-81992437 on Build GRCh38
rs766887483 Chr.9: 81992271-81992271 on Build GRCh38
rs751304370 Chr.9: 81992490-81992490 on Build GRCh38
rs769817458 Chr.9: 81992651-81992651 on Build GRCh38
rs776792441 Chr.9: 81992568-81992568 on Build GRCh38
rs777382775 Chr.9: 81992462-81992462 on Build GRCh38
rs142037367 Chr.9: 81992433-81992433 on Build GRCh38
rs374940088 Chr.9: 81992401-81992401 on Build GRCh38
rs376217865 Chr.9: 81992544-81992544 on Build GRCh38
rs201213048 Chr.9: 81992684-81992684 on Build GRCh38
rs575202702 Chr.9: 81992662-81992662 on Build GRCh38
rs747673865 Chr.9: 81992378-81992378 on Build GRCh38
rs772887082 Chr.9: 81992384-81992384 on Build GRCh38
rs760237044 Chr.9: 81992245-81992245 on Build GRCh38
rs373743285 Chr.9: 81992441-81992441 on Build GRCh38
rs1044972077 Chr.9: 81992590-81992590 on Build GRCh38
rs201821505 Chr.9: 81992320-81992320 on Build GRCh38
rs369209091 Chr.9: 81992455-81992455 on Build GRCh38
rs200441343 Chr.9: 81992673-81992673 on Build GRCh38
rs763259162 Chr.9: 81992388-81992388 on Build GRCh38
rs765575764 Chr.9: 81992319-81992319 on Build GRCh38
rs368257035 Chr.9: 81992658-81992658 on Build GRCh38
rs748212998 Chr.9: 81992642-81992642 on Build GRCh38
rs769320705 Chr.9: 81992459-81992459 on Build GRCh38
rs759822657 Chr.9: 81992472-81992472 on Build GRCh38
rs201356308 Chr.9: 81992447-81992447 on Build GRCh38
rs367832515 Chr.9: 81992615-81992615 on Build GRCh38
rs376003108 Chr.9: 81992636-81992636 on Build GRCh38
rs750923092 Chr.9: 81992265-81992265 on Build GRCh38
rs777331876 Chr.9: 81992538-81992538 on Build GRCh38
rs755419107 Chr.9: 81992670-81992670 on Build GRCh38
rs377116387 Chr.9: 81992672-81992672 on Build GRCh38
rs771693898 Chr.9: 81992241-81992241 on Build GRCh38
rs371185097 Chr.9: 81992391-81992391 on Build GRCh38
rs767677479 Chr.9: 81992331-81992331 on Build GRCh38
rs375054373 Chr.9: 81992282-81992282 on Build GRCh38
rs145735905 Chr.9: 81992654-81992654 on Build GRCh38
rs190878253 Chr.9: 81992577-81992577 on Build GRCh38
rs536287199 Chr.9: 81992477-81992477 on Build GRCh38
rs775527578 Chr.9: 81992304-81992304 on Build GRCh38
rs149127563 Chr.9: 81992255-81992255 on Build GRCh38
rs560197350 Chr.9: 81992699-81992699 on Build GRCh38
rs190903361 Chr.9: 81992288-81992288 on Build GRCh38
rs768478334 Chr.9: 81992700-81992700 on Build GRCh38
rs756102985 Chr.9: 81992598-81992598 on Build GRCh38
rs751935060 Chr.9: 81992668-81992668 on Build GRCh38
rs147641372 Chr.9: 81992236-81992236 on Build GRCh38
rs767483666 Chr.9: 81992422-81992422 on Build GRCh38
rs563171357 Chr.9: 81992351-81992351 on Build GRCh38
rs756346242 Chr.9: 81992336-81992336 on Build GRCh38
rs376227501 Chr.9: 81992476-81992476 on Build GRCh38
rs767341094 Chr.9: 81992596-81992596 on Build GRCh38
rs764034521 Chr.9: 81992607-81992607 on Build GRCh38
rs752627498 Chr.9: 81992597-81992597 on Build GRCh38
rs747582758 Chr.9: 81992563-81992563 on Build GRCh38

View Full Product Details