Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83780244-83780784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GKAP1
Entrez Gene ID
80318
Gene Name
G kinase anchoring protein 1
Gene Aliases
FKSG21, GKAP42
UniGene
Hs.522255
Gene Chromosome Location
Chr.9: 83739421-83817837 on Build GRCh38
Transcripts
NM_001135953.1 NM_025211.3 XM_005252241.2 XM_006717293.1 XM_011519058.2 XM_017015171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6428228 c.565C>T p.H189Y Chr.9: 83780402-83780402 on Build GRCh38
7290131 c.577A>G p.K193E Chr.9: 83780390-83780390 on Build GRCh38
3780145 c.563A>G p.D188G Chr.9: 83780404-83780404 on Build GRCh38
1463301 c.585G>A p.E195E Chr.9: 83780382-83780382 on Build GRCh38
5165214 c.563-5T>C p.? Chr.9: 83780409-83780409 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs562453749 Chr.9: 83780627-83780627 on Build GRCh38
rs565198228 Chr.9: 83780536-83780536 on Build GRCh38
rs982614698 Chr.9: 83780542-83780551 on Build GRCh38
rs199669659 Chr.9: 83780439-83780439 on Build GRCh38
rs200663185 Chr.9: 83780466-83780466 on Build GRCh38
rs1005725330 Chr.9: 83780466-83780469 on Build GRCh38
rs532297276 Chr.9: 83780715-83780715 on Build GRCh38
rs189435010 Chr.9: 83780412-83780412 on Build GRCh38
rs951444878 Chr.9: 83780532-83780532 on Build GRCh38
rs934987118 Chr.9: 83780680-83780680 on Build GRCh38
rs548267106 Chr.9: 83780644-83780644 on Build GRCh38
rs750500967 Chr.9: 83780415-83780415 on Build GRCh38
rs1009619848 Chr.9: 83780413-83780413 on Build GRCh38
rs1056266496 Chr.9: 83780419-83780419 on Build GRCh38
rs538665011 Chr.9: 83780281-83780281 on Build GRCh38
rs1040688882 Chr.9: 83780663-83780663 on Build GRCh38
rs376311607 Chr.9: 83780367-83780367 on Build GRCh38
rs1038578045 Chr.9: 83780753-83780753 on Build GRCh38
rs766596017 Chr.9: 83780435-83780435 on Build GRCh38
rs10868061 Chr.9: 83780756-83780756 on Build GRCh38
rs1026051195 Chr.9: 83780529-83780529 on Build GRCh38
rs747364180 Chr.9: 83780750-83780750 on Build GRCh38
rs891718698 Chr.9: 83780368-83780368 on Build GRCh38
rs765364735 Chr.9: 83780394-83780394 on Build GRCh38
rs115745898 Chr.9: 83780558-83780558 on Build GRCh38
rs771732099 Chr.9: 83780358-83780358 on Build GRCh38
rs529857543 Chr.9: 83780638-83780638 on Build GRCh38
rs1038890123 Chr.9: 83780299-83780299 on Build GRCh38
rs1043677350 Chr.9: 83780676-83780676 on Build GRCh38
rs761059174 Chr.9: 83780384-83780384 on Build GRCh38
rs75303439 Chr.9: 83780483-83780483 on Build GRCh38
rs1049717611 Chr.9: 83780698-83780698 on Build GRCh38
rs577179251 Chr.9: 83780543-83780543 on Build GRCh38
rs140108826 Chr.9: 83780653-83780653 on Build GRCh38
rs112907047 Chr.9: 83780284-83780284 on Build GRCh38
rs373163192 Chr.9: 83780343-83780343 on Build GRCh38
rs768310879 Chr.9: 83780413-83780414 on Build GRCh38
rs1045621356 Chr.9: 83780741-83780741 on Build GRCh38
rs778923783 Chr.9: 83780420-83780423 on Build GRCh38
rs769173593 Chr.9: 83780388-83780388 on Build GRCh38
rs745575801 Chr.9: 83780344-83780344 on Build GRCh38
rs758907095 Chr.9: 83780522-83780522 on Build GRCh38
rs946375762 Chr.9: 83780315-83780315 on Build GRCh38
rs1023998154 Chr.9: 83780535-83780535 on Build GRCh38
rs927170259 Chr.9: 83780329-83780329 on Build GRCh38
rs112400576 Chr.9: 83780659-83780659 on Build GRCh38
rs986846512 Chr.9: 83780539-83780539 on Build GRCh38
rs989962154 Chr.9: 83780604-83780604 on Build GRCh38
rs748833746 Chr.9: 83780332-83780332 on Build GRCh38
rs957080990 Chr.9: 83780593-83780593 on Build GRCh38
rs755617843 Chr.9: 83780524-83780524 on Build GRCh38
rs376240218 Chr.9: 83780688-83780688 on Build GRCh38
rs28696030 Chr.9: 83780355-83780355 on Build GRCh38
rs921853799 Chr.9: 83780310-83780310 on Build GRCh38
rs911720821 Chr.9: 83780276-83780276 on Build GRCh38
rs986872860 Chr.9: 83780272-83780272 on Build GRCh38
rs979911418 Chr.9: 83780547-83780547 on Build GRCh38
rs777061715 Chr.9: 83780389-83780389 on Build GRCh38
rs768925250 Chr.9: 83780720-83780720 on Build GRCh38
rs1023863010 Chr.9: 83780484-83780484 on Build GRCh38
rs923599786 Chr.9: 83780677-83780677 on Build GRCh38
rs780544607 Chr.9: 83780655-83780655 on Build GRCh38
rs563454822 Chr.9: 83780667-83780667 on Build GRCh38
rs559873758 Chr.9: 83780466-83780467 on Build GRCh38
rs764141580 Chr.9: 83780444-83780444 on Build GRCh38
rs770508211 Chr.9: 83780333-83780333 on Build GRCh38
rs753762886 Chr.9: 83780451-83780451 on Build GRCh38
rs961635320 Chr.9: 83780489-83780490 on Build GRCh38
rs761882263 Chr.9: 83780393-83780393 on Build GRCh38
rs935829851 Chr.9: 83780364-83780364 on Build GRCh38
rs751192563 Chr.9: 83780270-83780270 on Build GRCh38
rs748755928 Chr.9: 83780413-83780413 on Build GRCh38
rs752469252 Chr.9: 83780437-83780437 on Build GRCh38
rs946607258 Chr.9: 83780670-83780670 on Build GRCh38
rs72155123 Chr.9: 83780482-83780483 on Build GRCh38
rs181803158 Chr.9: 83780429-83780429 on Build GRCh38
rs867589665 Chr.9: 83780294-83780294 on Build GRCh38
rs779804701 Chr.9: 83780388-83780389 on Build GRCh38
rs575366200 Chr.9: 83780322-83780322 on Build GRCh38
rs980241113 Chr.9: 83780316-83780316 on Build GRCh38
rs538576821 Chr.9: 83780681-83780681 on Build GRCh38
rs1042074512 Chr.9: 83780457-83780457 on Build GRCh38

View Full Product Details