Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133459885-133460382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACFD1
Entrez Gene ID
11094
Gene Name
calcium channel flower domain containing 1
Gene Aliases
C9orf7, D9S2135, FLOWER
UniGene
Hs.62003
Gene Chromosome Location
Chr.9: 133459965-133470787 on Build GRCh38
Transcripts
NM_001135775.2 NM_001242369.1 NM_001242370.1 NM_017586.3 XM_011518182.1 XM_011518186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738202 c.94C>A p.R32S Chr.9: 133460160-133460160 on Build GRCh38
6432876 c.10T>C p.S4P Chr.9: 133460076-133460076 on Build GRCh38
4799259 c.67A>C p.M23L Chr.9: 133460133-133460133 on Build GRCh38
1738201 c.94C>A p.R32S Chr.9: 133460160-133460160 on Build GRCh38
6234571 c.99G>T p.L33L Chr.9: 133460165-133460165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587739306 Chr.9: 133459974-133459974 on Build GRCh38
rs1024390788 Chr.9: 133460318-133460318 on Build GRCh38
rs867642509 Chr.9: 133460144-133460144 on Build GRCh38
rs1043495070 Chr.9: 133459965-133459965 on Build GRCh38
rs992904111 Chr.9: 133460248-133460248 on Build GRCh38
rs587655555 Chr.9: 133459952-133459952 on Build GRCh38
rs376963405 Chr.9: 133460071-133460071 on Build GRCh38
rs781873440 Chr.9: 133460105-133460105 on Build GRCh38
rs148657560 Chr.9: 133460147-133460147 on Build GRCh38
rs900294544 Chr.9: 133460290-133460290 on Build GRCh38
rs782102769 Chr.9: 133460099-133460099 on Build GRCh38
rs966481282 Chr.9: 133460277-133460277 on Build GRCh38
rs1034233643 Chr.9: 133459983-133459983 on Build GRCh38
rs782387178 Chr.9: 133460202-133460202 on Build GRCh38
rs587693725 Chr.9: 133460006-133460006 on Build GRCh38
rs1048349408 Chr.9: 133460276-133460276 on Build GRCh38
rs879722510 Chr.9: 133460208-133460209 on Build GRCh38
rs782154334 Chr.9: 133460214-133460214 on Build GRCh38
rs781921467 Chr.9: 133460093-133460093 on Build GRCh38
rs985107296 Chr.9: 133460292-133460292 on Build GRCh38
rs782343711 Chr.9: 133460184-133460184 on Build GRCh38
rs782209844 Chr.9: 133460238-133460239 on Build GRCh38
rs376297486 Chr.9: 133460190-133460190 on Build GRCh38
rs28516935 Chr.9: 133460081-133460081 on Build GRCh38
rs899273746 Chr.9: 133460242-133460242 on Build GRCh38
rs369216620 Chr.9: 133460068-133460068 on Build GRCh38
rs781895307 Chr.9: 133460134-133460134 on Build GRCh38
rs943495146 Chr.9: 133460059-133460059 on Build GRCh38
rs782808554 Chr.9: 133460126-133460126 on Build GRCh38
rs587706782 Chr.9: 133460272-133460272 on Build GRCh38
rs1026449104 Chr.9: 133460041-133460041 on Build GRCh38
rs782117442 Chr.9: 133460117-133460117 on Build GRCh38
rs948668761 Chr.9: 133460253-133460253 on Build GRCh38
rs782435269 Chr.9: 133460089-133460097 on Build GRCh38
rs1013204816 Chr.9: 133460008-133460008 on Build GRCh38
rs960532582 Chr.9: 133460312-133460312 on Build GRCh38
rs782701202 Chr.9: 133460028-133460028 on Build GRCh38
rs2301615 Chr.9: 133460340-133460340 on Build GRCh38
rs886170253 Chr.9: 133460192-133460192 on Build GRCh38
rs372774526 Chr.9: 133460167-133460167 on Build GRCh38
rs995008715 Chr.9: 133460040-133460040 on Build GRCh38
rs782353386 Chr.9: 133460062-133460062 on Build GRCh38
rs1043395002 Chr.9: 133460297-133460297 on Build GRCh38
rs1018637042 Chr.9: 133460061-133460061 on Build GRCh38
rs182618302 Chr.9: 133460001-133460001 on Build GRCh38
rs917227246 Chr.9: 133460140-133460140 on Build GRCh38
rs587702555 Chr.9: 133460137-133460137 on Build GRCh38
rs782329177 Chr.9: 133460091-133460091 on Build GRCh38
rs982319006 Chr.9: 133459978-133459978 on Build GRCh38
rs782731993 Chr.9: 133460104-133460104 on Build GRCh38
rs980081424 Chr.9: 133460341-133460341 on Build GRCh38
rs967974377 Chr.9: 133460238-133460238 on Build GRCh38
rs782190340 Chr.9: 133460027-133460027 on Build GRCh38
rs977716551 Chr.9: 133460030-133460030 on Build GRCh38
rs866256519 Chr.9: 133460110-133460110 on Build GRCh38
rs1031356373 Chr.9: 133460310-133460310 on Build GRCh38
rs929415951 Chr.9: 133460335-133460335 on Build GRCh38
rs895415697 Chr.9: 133460357-133460357 on Build GRCh38
rs915045700 Chr.9: 133459942-133459942 on Build GRCh38
rs960837638 Chr.9: 133460247-133460247 on Build GRCh38
rs1055110365 Chr.9: 133460353-133460353 on Build GRCh38
rs934648702 Chr.9: 133459991-133459991 on Build GRCh38
rs1013846931 Chr.9: 133460313-133460313 on Build GRCh38
rs1003833795 Chr.9: 133460262-133460262 on Build GRCh38
rs866413610 Chr.9: 133460148-133460148 on Build GRCh38
rs1040066436 Chr.9: 133460289-133460289 on Build GRCh38
rs377184464 Chr.9: 133460057-133460057 on Build GRCh38
rs1049612155 Chr.9: 133460052-133460052 on Build GRCh38
rs946570137 Chr.9: 133459944-133459944 on Build GRCh38
rs978309573 Chr.9: 133460240-133460240 on Build GRCh38
rs911791844 Chr.9: 133459924-133459924 on Build GRCh38
rs370651118 Chr.9: 133460201-133460201 on Build GRCh38
rs144774636 Chr.9: 133460177-133460177 on Build GRCh38
rs867482128 Chr.9: 133460077-133460077 on Build GRCh38
rs781886424 Chr.9: 133460023-133460023 on Build GRCh38
rs587742091 Chr.9: 133460243-133460243 on Build GRCh38
rs140916557 Chr.9: 133460149-133460149 on Build GRCh38
rs782801031 Chr.9: 133460016-133460016 on Build GRCh38
rs985777177 Chr.9: 133460205-133460205 on Build GRCh38
rs782404041 Chr.9: 133460215-133460215 on Build GRCh38
rs964639440 Chr.9: 133459932-133459932 on Build GRCh38
rs903765094 Chr.9: 133460299-133460299 on Build GRCh38
rs199903701 Chr.9: 133460160-133460160 on Build GRCh38
rs587674078 Chr.9: 133459912-133459912 on Build GRCh38
rs782466906 Chr.9: 133460032-133460032 on Build GRCh38
rs1022170525 Chr.9: 133460084-133460084 on Build GRCh38
rs989228307 Chr.9: 133459992-133459992 on Build GRCh38
rs945316732 Chr.9: 133459908-133459908 on Build GRCh38
rs971345384 Chr.9: 133459956-133459956 on Build GRCh38
rs924685456 Chr.9: 133460121-133460121 on Build GRCh38
rs995792909 Chr.9: 133460291-133460291 on Build GRCh38
rs144044226 Chr.9: 133459938-133459938 on Build GRCh38
rs782030947 Chr.9: 133460085-133460085 on Build GRCh38
rs917342986 Chr.9: 133460249-133460249 on Build GRCh38
rs933949128 Chr.9: 133460204-133460204 on Build GRCh38
rs1004188059 Chr.9: 133460146-133460146 on Build GRCh38
rs587615035 Chr.9: 133460263-133460263 on Build GRCh38
rs782627382 Chr.9: 133460190-133460214 on Build GRCh38
rs3216889 Chr.9: 133460263-133460264 on Build GRCh38
rs998032334 Chr.9: 133459918-133459918 on Build GRCh38
rs587662076 Chr.9: 133460210-133460210 on Build GRCh38
rs934784916 Chr.9: 133460130-133460130 on Build GRCh38
rs1026085191 Chr.9: 133459910-133459910 on Build GRCh38
rs920624209 Chr.9: 133460343-133460343 on Build GRCh38
rs980133652 Chr.9: 133460264-133460264 on Build GRCh38
rs1044631391 Chr.9: 133460150-133460150 on Build GRCh38
rs374585220 Chr.9: 133460218-133460218 on Build GRCh38
rs587601948 Chr.9: 133460301-133460301 on Build GRCh38
rs781969766 Chr.9: 133460195-133460195 on Build GRCh38
rs587726063 Chr.9: 133459915-133459915 on Build GRCh38
rs906796322 Chr.9: 133460120-133460120 on Build GRCh38
rs782632641 Chr.9: 133460161-133460161 on Build GRCh38
rs1026247670 Chr.9: 133460088-133460088 on Build GRCh38
rs782194288 Chr.9: 133460178-133460178 on Build GRCh38
rs782366044 Chr.9: 133460082-133460082 on Build GRCh38
rs914946881 Chr.9: 133459994-133459994 on Build GRCh38
rs941203216 Chr.9: 133460224-133460224 on Build GRCh38
rs974606968 Chr.9: 133460078-133460078 on Build GRCh38
rs781841314 Chr.9: 133460029-133460029 on Build GRCh38
rs889220084 Chr.9: 133459938-133459939 on Build GRCh38
rs782455223 Chr.9: 133460143-133460143 on Build GRCh38
rs924724706 Chr.9: 133459984-133459984 on Build GRCh38
rs587665941 Chr.9: 133460271-133460271 on Build GRCh38
rs944872524 Chr.9: 133460286-133460286 on Build GRCh38
rs1044682733 Chr.9: 133460009-133460009 on Build GRCh38
rs1026531285 Chr.9: 133459955-133459955 on Build GRCh38
rs781958453 Chr.9: 133460222-133460222 on Build GRCh38
rs867364674 Chr.9: 133460135-133460135 on Build GRCh38
rs782142347 Chr.9: 133460090-133460090 on Build GRCh38
rs587624188 Chr.9: 133460044-133460044 on Build GRCh38
rs886584863 Chr.9: 133460282-133460282 on Build GRCh38
rs886224723 Chr.9: 133460053-133460053 on Build GRCh38
rs930791690 Chr.9: 133460164-133460164 on Build GRCh38
rs2301614 Chr.9: 133460038-133460038 on Build GRCh38
rs966429003 Chr.9: 133460196-133460196 on Build GRCh38
rs930934679 Chr.9: 133460275-133460275 on Build GRCh38
rs1057171467 Chr.9: 133459997-133459997 on Build GRCh38

View Full Product Details