Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133470717-133471212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACFD1
Entrez Gene ID
11094
Gene Name
calcium channel flower domain containing 1
Gene Aliases
C9orf7, D9S2135, FLOWER
UniGene
Hs.62003
Gene Chromosome Location
Chr.9: 133459965-133470787 on Build GRCh38
Transcripts
NM_001135775.2 NM_001242369.1 NM_001242370.1 NM_017586.3 XM_011518181.2 XM_011518182.1 XM_011518184.1 XM_011518185.2 XM_011518186.1 NM_001145099.1 NM_017585.3 XM_011518189.2 XM_017014237.1 XM_017014238.1

Gene Symbol
SLC2A6
Entrez Gene ID
11182
Gene Name
solute carrier family 2 member 6
Gene Aliases
GLUT6, GLUT9, HSA011372
UniGene
Hs.244378
Gene Chromosome Location
Chr.9: 133471094-133479154 on Build GRCh38
Transcripts
NM_001135775.2 NM_001242369.1 NM_001242370.1 NM_017586.3 XM_011518181.2 XM_011518182.1 XM_011518184.1 XM_011518185.2 XM_011518186.1 NM_001145099.1 NM_017585.3 XM_011518189.2 XM_017014237.1 XM_017014238.1

SNP Detail

SNP ID Chromosome Location
rs902424701 Chr.9: 133470803-133470803 on Build GRCh38
rs1043355659 Chr.9: 133470985-133470985 on Build GRCh38
rs1006701648 Chr.9: 133471057-133471057 on Build GRCh38
rs772296587 Chr.9: 133470763-133470763 on Build GRCh38
rs45489391 Chr.9: 133470753-133470753 on Build GRCh38
rs966132760 Chr.9: 133470772-133470772 on Build GRCh38
rs587762826 Chr.9: 133471053-133471053 on Build GRCh38
rs56739836 Chr.9: 133471097-133471097 on Build GRCh38
rs775447426 Chr.9: 133471086-133471086 on Build GRCh38
rs56409531 Chr.9: 133470797-133470797 on Build GRCh38
rs1001747463 Chr.9: 133470943-133470943 on Build GRCh38
rs782809295 Chr.9: 133470946-133470946 on Build GRCh38
rs950045229 Chr.9: 133471156-133471156 on Build GRCh38
rs373432181 Chr.9: 133470894-133470894 on Build GRCh38
rs967768298 Chr.9: 133471133-133471134 on Build GRCh38
rs999389706 Chr.9: 133470805-133470805 on Build GRCh38
rs994865756 Chr.9: 133471076-133471076 on Build GRCh38
rs979360743 Chr.9: 133471130-133471130 on Build GRCh38
rs34776812 Chr.9: 133471081-133471082 on Build GRCh38
rs887957895 Chr.9: 133470975-133470975 on Build GRCh38
rs950078424 Chr.9: 133470912-133470912 on Build GRCh38
rs782059107 Chr.9: 133471011-133471011 on Build GRCh38
rs939088737 Chr.9: 133470762-133470762 on Build GRCh38
rs961171550 Chr.9: 133471110-133471110 on Build GRCh38
rs983678971 Chr.9: 133470922-133470927 on Build GRCh38
rs920824127 Chr.9: 133470751-133470751 on Build GRCh38
rs187217836 Chr.9: 133471133-133471133 on Build GRCh38
rs927299389 Chr.9: 133471164-133471164 on Build GRCh38
rs988892286 Chr.9: 133471143-133471143 on Build GRCh38
rs370177237 Chr.9: 133471194-133471194 on Build GRCh38
rs1045956349 Chr.9: 133470913-133470913 on Build GRCh38
rs979226591 Chr.9: 133470823-133470823 on Build GRCh38
rs951794170 Chr.9: 133470761-133470761 on Build GRCh38
rs587599535 Chr.9: 133471032-133471032 on Build GRCh38
rs916300372 Chr.9: 133470973-133470973 on Build GRCh38
rs1050992017 Chr.9: 133471050-133471050 on Build GRCh38
rs369020312 Chr.9: 133470981-133470981 on Build GRCh38
rs113403699 Chr.9: 133471049-133471049 on Build GRCh38
rs375251371 Chr.9: 133471057-133471057 on Build GRCh38
rs374733854 Chr.9: 133470917-133470917 on Build GRCh38
rs921884737 Chr.9: 133470794-133470794 on Build GRCh38
rs973375534 Chr.9: 133470908-133470909 on Build GRCh38
rs782035414 Chr.9: 133470957-133470957 on Build GRCh38
rs587731728 Chr.9: 133470905-133470905 on Build GRCh38
rs587639449 Chr.9: 133470944-133470944 on Build GRCh38
rs373192527 Chr.9: 133470742-133470742 on Build GRCh38
rs146634648 Chr.9: 133471078-133471079 on Build GRCh38
rs587605127 Chr.9: 133471132-133471132 on Build GRCh38
rs1015113178 Chr.9: 133471074-133471074 on Build GRCh38
rs377314373 Chr.9: 133470748-133470748 on Build GRCh38
rs1009405436 Chr.9: 133470846-133470846 on Build GRCh38
rs902477082 Chr.9: 133471021-133471021 on Build GRCh38
rs369985651 Chr.9: 133470827-133470827 on Build GRCh38
rs372242658 Chr.9: 133470825-133470825 on Build GRCh38
rs936075718 Chr.9: 133470848-133470848 on Build GRCh38
rs1014565679 Chr.9: 133470766-133470766 on Build GRCh38
rs587723229 Chr.9: 133470951-133470951 on Build GRCh38
rs72619330 Chr.9: 133471085-133471085 on Build GRCh38
rs950928766 Chr.9: 133470921-133470922 on Build GRCh38
rs781881305 Chr.9: 133471159-133471159 on Build GRCh38
rs145943109 Chr.9: 133470922-133470922 on Build GRCh38
rs965740420 Chr.9: 133471075-133471075 on Build GRCh38
rs928129310 Chr.9: 133470933-133470933 on Build GRCh38
rs906156840 Chr.9: 133470926-133470926 on Build GRCh38
rs983626249 Chr.9: 133470765-133470765 on Build GRCh38
rs782106556 Chr.9: 133470797-133470798 on Build GRCh38
rs937567387 Chr.9: 133471193-133471193 on Build GRCh38
rs975980955 Chr.9: 133470790-133470790 on Build GRCh38
rs148282755 Chr.9: 133471147-133471147 on Build GRCh38
rs587650684 Chr.9: 133470760-133470760 on Build GRCh38

View Full Product Details