Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134031695-134032193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD3
Entrez Gene ID
8019
Gene Name
bromodomain containing 3
Gene Aliases
ORFX, RING3L
UniGene
Hs.522472
Gene Chromosome Location
Chr.9: 134030305-134068533 on Build GRCh38
Transcripts
NM_007371.3 XM_017015165.1 XM_017015166.1

SNP Detail

SNP ID Chromosome Location
rs932286817 Chr.9: 134032058-134032058 on Build GRCh38
rs1003510175 Chr.9: 134032072-134032072 on Build GRCh38
rs559894844 Chr.9: 134032054-134032054 on Build GRCh38
rs111890662 Chr.9: 134031888-134031888 on Build GRCh38
rs570970355 Chr.9: 134031907-134031907 on Build GRCh38
rs993555346 Chr.9: 134032048-134032048 on Build GRCh38
rs982037692 Chr.9: 134031805-134031805 on Build GRCh38
rs1033560912 Chr.9: 134031942-134031942 on Build GRCh38
rs982455353 Chr.9: 134031939-134031939 on Build GRCh38
rs375606332 Chr.9: 134032063-134032063 on Build GRCh38
rs112288060 Chr.9: 134031739-134031739 on Build GRCh38
rs889339606 Chr.9: 134032016-134032016 on Build GRCh38
rs945132918 Chr.9: 134031965-134031965 on Build GRCh38
rs966507008 Chr.9: 134032112-134032112 on Build GRCh38
rs989345984 Chr.9: 134031950-134031950 on Build GRCh38
rs906652685 Chr.9: 134032105-134032105 on Build GRCh38
rs936924893 Chr.9: 134032084-134032084 on Build GRCh38
rs143795991 Chr.9: 134031741-134031741 on Build GRCh38
rs550093493 Chr.9: 134031830-134031830 on Build GRCh38
rs943611971 Chr.9: 134032030-134032030 on Build GRCh38
rs548307040 Chr.9: 134031863-134031863 on Build GRCh38
rs556420800 Chr.9: 134031982-134031982 on Build GRCh38
rs749861087 Chr.9: 134031842-134031842 on Build GRCh38
rs572064417 Chr.9: 134032098-134032098 on Build GRCh38
rs115197915 Chr.9: 134031951-134031951 on Build GRCh38
rs778144461 Chr.9: 134031844-134031844 on Build GRCh38
rs1020062060 Chr.9: 134031820-134031832 on Build GRCh38
rs933599334 Chr.9: 134031883-134031883 on Build GRCh38
rs3802503 Chr.9: 134031850-134031850 on Build GRCh38
rs145262108 Chr.9: 134032011-134032011 on Build GRCh38
rs114367019 Chr.9: 134032099-134032099 on Build GRCh38
rs114433245 Chr.9: 134031922-134031922 on Build GRCh38
rs574829609 Chr.9: 134031989-134031989 on Build GRCh38
rs977824634 Chr.9: 134031968-134031968 on Build GRCh38
rs899326004 Chr.9: 134032039-134032039 on Build GRCh38
rs867017711 Chr.9: 134031740-134031740 on Build GRCh38
rs1026301510 Chr.9: 134031923-134031923 on Build GRCh38
rs552299738 Chr.9: 134031751-134031751 on Build GRCh38
rs1002158136 Chr.9: 134032108-134032108 on Build GRCh38
rs972005242 Chr.9: 134031768-134031768 on Build GRCh38
rs889361647 Chr.9: 134032064-134032064 on Build GRCh38
rs1053917627 Chr.9: 134032085-134032085 on Build GRCh38
rs772756982 Chr.9: 134032168-134032168 on Build GRCh38
rs566927065 Chr.9: 134031876-134031876 on Build GRCh38
rs75833842 Chr.9: 134031791-134031791 on Build GRCh38
rs747665555 Chr.9: 134031921-134031921 on Build GRCh38
rs3802502 Chr.9: 134031904-134031904 on Build GRCh38
rs959472477 Chr.9: 134031947-134031947 on Build GRCh38
rs963351446 Chr.9: 134031743-134031743 on Build GRCh38
rs970724090 Chr.9: 134031925-134031925 on Build GRCh38
rs1030941986 Chr.9: 134031731-134031731 on Build GRCh38
rs1016310295 Chr.9: 134031920-134031920 on Build GRCh38
rs933650745 Chr.9: 134031993-134031993 on Build GRCh38
rs1037874572 Chr.9: 134032031-134032031 on Build GRCh38
rs1042113898 Chr.9: 134031875-134031875 on Build GRCh38
rs531574762 Chr.9: 134031792-134031792 on Build GRCh38
rs1025003798 Chr.9: 134032104-134032104 on Build GRCh38
rs147284416 Chr.9: 134031849-134031849 on Build GRCh38
rs781644877 Chr.9: 134032141-134032141 on Build GRCh38
rs989636375 Chr.9: 134031843-134031843 on Build GRCh38

View Full Product Details