Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134034623-134035117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD3
Entrez Gene ID
8019
Gene Name
bromodomain containing 3
Gene Aliases
ORFX, RING3L
UniGene
Hs.522472
Gene Chromosome Location
Chr.9: 134030305-134068533 on Build GRCh38
Transcripts
NM_007371.3 XM_006717291.2 XM_011519052.2 XM_017015165.1 XM_017015166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612670 c.1976A>C p.E659A Chr.9: 134034790-134034790 on Build GRCh38
5663535 c.2019G>A p.Q673Q Chr.9: 134034747-134034747 on Build GRCh38
5620362 c.1966T>G p.S656A Chr.9: 134034800-134034800 on Build GRCh38
1674940 c.1984C>T p.Q662* Chr.9: 134034782-134034782 on Build GRCh38
6612656 c.2027G>A p.S676N Chr.9: 134034739-134034739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995072790 Chr.9: 134034816-134034816 on Build GRCh38
rs149395455 Chr.9: 134035065-134035065 on Build GRCh38
rs767562435 Chr.9: 134034881-134034881 on Build GRCh38
rs752878207 Chr.9: 134034768-134034770 on Build GRCh38
rs200658535 Chr.9: 134034866-134034866 on Build GRCh38
rs200445063 Chr.9: 134034834-134034834 on Build GRCh38
rs761605958 Chr.9: 134034654-134034654 on Build GRCh38
rs762577259 Chr.9: 134034685-134034685 on Build GRCh38
rs750698523 Chr.9: 134034660-134034660 on Build GRCh38
rs150679546 Chr.9: 134034770-134034770 on Build GRCh38
rs752803727 Chr.9: 134034876-134034876 on Build GRCh38
rs767879281 Chr.9: 134034690-134034690 on Build GRCh38
rs764475299 Chr.9: 134034652-134034652 on Build GRCh38
rs7024684 Chr.9: 134035092-134035092 on Build GRCh38
rs761021509 Chr.9: 134034862-134034862 on Build GRCh38
rs199544102 Chr.9: 134034663-134034663 on Build GRCh38
rs973284672 Chr.9: 134034647-134034647 on Build GRCh38
rs780107955 Chr.9: 134034662-134034662 on Build GRCh38
rs372535973 Chr.9: 134034835-134034835 on Build GRCh38
rs543423463 Chr.9: 134034664-134034664 on Build GRCh38
rs571041837 Chr.9: 134035010-134035010 on Build GRCh38
rs200178408 Chr.9: 134034697-134034697 on Build GRCh38
rs748745514 Chr.9: 134034754-134034754 on Build GRCh38
rs766556368 Chr.9: 134034798-134034798 on Build GRCh38
rs201095386 Chr.9: 134034776-134034776 on Build GRCh38
rs974415978 Chr.9: 134034829-134034829 on Build GRCh38
rs201878034 Chr.9: 134034686-134034686 on Build GRCh38
rs750574016 Chr.9: 134034693-134034693 on Build GRCh38
rs200522998 Chr.9: 134034799-134034799 on Build GRCh38
rs759836434 Chr.9: 134034870-134034870 on Build GRCh38
rs201409434 Chr.9: 134034867-134034867 on Build GRCh38
rs964710796 Chr.9: 134034885-134034885 on Build GRCh38
rs376499654 Chr.9: 134034661-134034661 on Build GRCh38
rs745469027 Chr.9: 134034713-134034713 on Build GRCh38
rs769551638 Chr.9: 134034738-134034738 on Build GRCh38
rs963367089 Chr.9: 134034813-134034813 on Build GRCh38
rs200751659 Chr.9: 134034656-134034656 on Build GRCh38
rs774660543 Chr.9: 134034759-134034759 on Build GRCh38
rs752179223 Chr.9: 134034688-134034688 on Build GRCh38
rs374242241 Chr.9: 134034857-134034857 on Build GRCh38
rs746980947 Chr.9: 134034709-134034709 on Build GRCh38
rs940145002 Chr.9: 134034875-134034875 on Build GRCh38
rs974472386 Chr.9: 134034964-134034964 on Build GRCh38
rs903243496 Chr.9: 134034722-134034722 on Build GRCh38
rs767825546 Chr.9: 134034764-134034764 on Build GRCh38
rs759266572 Chr.9: 134034673-134034673 on Build GRCh38
rs751413175 Chr.9: 134034805-134034805 on Build GRCh38
rs762309822 Chr.9: 134034762-134034762 on Build GRCh38
rs568764065 Chr.9: 134035047-134035047 on Build GRCh38
rs546651415 Chr.9: 134035009-134035009 on Build GRCh38
rs202110624 Chr.9: 134034659-134034659 on Build GRCh38
rs917355409 Chr.9: 134034975-134034975 on Build GRCh38
rs534363482 Chr.9: 134034972-134034972 on Build GRCh38
rs770784423 Chr.9: 134034710-134034710 on Build GRCh38
rs557797355 Chr.9: 134035011-134035011 on Build GRCh38
rs534893099 Chr.9: 134034853-134034854 on Build GRCh38
rs757284041 Chr.9: 134034808-134034808 on Build GRCh38
rs891632158 Chr.9: 134035026-134035026 on Build GRCh38
rs775166137 Chr.9: 134034676-134034676 on Build GRCh38
rs200692106 Chr.9: 134034666-134034666 on Build GRCh38
rs201491764 Chr.9: 134034837-134034837 on Build GRCh38
rs759838857 Chr.9: 134034667-134034667 on Build GRCh38
rs751851368 Chr.9: 134034654-134034654 on Build GRCh38
rs775252857 Chr.9: 134034741-134034741 on Build GRCh38
rs950058997 Chr.9: 134034677-134034678 on Build GRCh38
rs750685722 Chr.9: 134034661-134034661 on Build GRCh38
rs149702159 Chr.9: 134034802-134034802 on Build GRCh38
rs200539890 Chr.9: 134034695-134034695 on Build GRCh38
rs777566558 Chr.9: 134034707-134034707 on Build GRCh38
rs567265957 Chr.9: 134034910-134034910 on Build GRCh38
rs202198193 Chr.9: 134034737-134034737 on Build GRCh38
rs747140330 Chr.9: 134034997-134034997 on Build GRCh38
rs139511636 Chr.9: 134034780-134034780 on Build GRCh38
rs199789615 Chr.9: 134034871-134034871 on Build GRCh38
rs1044441911 Chr.9: 134034998-134034998 on Build GRCh38
rs368357181 Chr.9: 134034792-134034792 on Build GRCh38
rs556758211 Chr.9: 134035043-134035043 on Build GRCh38
rs202118381 Chr.9: 134034784-134034784 on Build GRCh38
rs773144321 Chr.9: 134034711-134034711 on Build GRCh38
rs768272924 Chr.9: 134034758-134034758 on Build GRCh38
rs758595679 Chr.9: 134034879-134034879 on Build GRCh38
rs199572673 Chr.9: 134034851-134034851 on Build GRCh38
rs756042834 Chr.9: 134034822-134034822 on Build GRCh38
rs200046701 Chr.9: 134034689-134034689 on Build GRCh38
rs200925930 Chr.9: 134034706-134034706 on Build GRCh38
rs866410787 Chr.9: 134035042-134035042 on Build GRCh38
rs201974693 Chr.9: 134034821-134034821 on Build GRCh38
rs781331762 Chr.9: 134034820-134034820 on Build GRCh38
rs535547843 Chr.9: 134034660-134034661 on Build GRCh38
rs190712968 Chr.9: 134034696-134034696 on Build GRCh38
rs770909070 Chr.9: 134034665-134034665 on Build GRCh38
rs765365622 Chr.9: 134034803-134034803 on Build GRCh38
rs1057133083 Chr.9: 134035093-134035093 on Build GRCh38
rs531256085 Chr.9: 134034841-134034841 on Build GRCh38
rs149036183 Chr.9: 134034697-134034698 on Build GRCh38
rs748601732 Chr.9: 134034845-134034845 on Build GRCh38
rs540744667 Chr.9: 134035089-134035090 on Build GRCh38
rs776652878 Chr.9: 134034868-134034868 on Build GRCh38
rs769389776 Chr.9: 134034674-134034674 on Build GRCh38

View Full Product Details