Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134909505-134910007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCN1
Entrez Gene ID
2219
Gene Name
ficolin 1
Gene Aliases
FCNM
UniGene
Hs.440898
Gene Chromosome Location
Chr.9: 134909585-134917960 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4421599 c.931G>A p.G311R Chr.9: 134909848-134909848 on Build GRCh38
3655504 c.820T>C p.F274L Chr.9: 134909959-134909959 on Build GRCh38
3655503 c.918G>C p.W306C Chr.9: 134909861-134909861 on Build GRCh38
1701940 c.882G>A p.M294I Chr.9: 134909897-134909897 on Build GRCh38
6662820 c.972G>A p.R324R Chr.9: 134909807-134909807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922182577 Chr.9: 134909789-134909789 on Build GRCh38
rs866409596 Chr.9: 134909896-134909896 on Build GRCh38
rs114390629 Chr.9: 134909743-134909743 on Build GRCh38
rs773870053 Chr.9: 134909739-134909739 on Build GRCh38
rs555801984 Chr.9: 134909774-134909774 on Build GRCh38
rs754815600 Chr.9: 134909882-134909882 on Build GRCh38
rs570055941 Chr.9: 134909742-134909742 on Build GRCh38
rs56151153 Chr.9: 134909872-134909872 on Build GRCh38
rs755088107 Chr.9: 134909908-134909908 on Build GRCh38
rs946845083 Chr.9: 134909639-134909639 on Build GRCh38
rs567267289 Chr.9: 134909686-134909686 on Build GRCh38
rs372242075 Chr.9: 134909938-134909938 on Build GRCh38
rs754067876 Chr.9: 134909745-134909745 on Build GRCh38
rs768037000 Chr.9: 134909558-134909558 on Build GRCh38
rs753319863 Chr.9: 134909624-134909624 on Build GRCh38
rs771679474 Chr.9: 134909910-134909910 on Build GRCh38
rs1128428 Chr.9: 134909834-134909834 on Build GRCh38
rs759627013 Chr.9: 134909750-134909750 on Build GRCh38
rs546732453 Chr.9: 134909698-134909698 on Build GRCh38
rs770688747 Chr.9: 134909613-134909613 on Build GRCh38
rs776360487 Chr.9: 134909634-134909634 on Build GRCh38
rs746958769 Chr.9: 134909601-134909601 on Build GRCh38
rs755909203 Chr.9: 134909779-134909779 on Build GRCh38
rs886559058 Chr.9: 134909901-134909901 on Build GRCh38
rs779764622 Chr.9: 134909788-134909788 on Build GRCh38
rs949724180 Chr.9: 134909843-134909843 on Build GRCh38
rs756846531 Chr.9: 134909981-134909981 on Build GRCh38
rs753513714 Chr.9: 134909864-134909864 on Build GRCh38
rs1128430 Chr.9: 134909829-134909829 on Build GRCh38
rs771767522 Chr.9: 134909799-134909799 on Build GRCh38
rs1048923998 Chr.9: 134909892-134909892 on Build GRCh38
rs749133890 Chr.9: 134909793-134909793 on Build GRCh38
rs201132846 Chr.9: 134909666-134909666 on Build GRCh38
rs576110383 Chr.9: 134909951-134909951 on Build GRCh38
rs780455054 Chr.9: 134909564-134909564 on Build GRCh38
rs745693633 Chr.9: 134909644-134909644 on Build GRCh38
rs150625869 Chr.9: 134909977-134909977 on Build GRCh38
rs752580161 Chr.9: 134909535-134909535 on Build GRCh38
rs968200258 Chr.9: 134909792-134909792 on Build GRCh38
rs747878485 Chr.9: 134909909-134909909 on Build GRCh38
rs750777696 Chr.9: 134909691-134909691 on Build GRCh38
rs548853763 Chr.9: 134909669-134909669 on Build GRCh38
rs777571891 Chr.9: 134909976-134909976 on Build GRCh38
rs148912895 Chr.9: 134909835-134909835 on Build GRCh38
rs187262304 Chr.9: 134909776-134909776 on Build GRCh38
rs975400818 Chr.9: 134909772-134909772 on Build GRCh38
rs769365850 Chr.9: 134909652-134909652 on Build GRCh38
rs765165815 Chr.9: 134909955-134909955 on Build GRCh38
rs374704410 Chr.9: 134909757-134909757 on Build GRCh38
rs371244932 Chr.9: 134909808-134909808 on Build GRCh38
rs780074993 Chr.9: 134909857-134909857 on Build GRCh38
rs762463985 Chr.9: 134909678-134909678 on Build GRCh38
rs775655624 Chr.9: 134909936-134909936 on Build GRCh38
rs754066535 Chr.9: 134909699-134909699 on Build GRCh38
rs982636078 Chr.9: 134909878-134909878 on Build GRCh38
rs777755090 Chr.9: 134909591-134909591 on Build GRCh38
rs757694931 Chr.9: 134909532-134909532 on Build GRCh38
rs199881741 Chr.9: 134909935-134909935 on Build GRCh38
rs375441016 Chr.9: 134909939-134909939 on Build GRCh38
rs775014502 Chr.9: 134909653-134909653 on Build GRCh38
rs373784301 Chr.9: 134909855-134909855 on Build GRCh38
rs539893739 Chr.9: 134909734-134909734 on Build GRCh38
rs769632447 Chr.9: 134909723-134909723 on Build GRCh38
rs775201645 Chr.9: 134909728-134909728 on Build GRCh38
rs888376734 Chr.9: 134909675-134909675 on Build GRCh38
rs749323278 Chr.9: 134909565-134909565 on Build GRCh38
rs530455123 Chr.9: 134909617-134909617 on Build GRCh38
rs368199008 Chr.9: 134909796-134909796 on Build GRCh38
rs535025459 Chr.9: 134909811-134909811 on Build GRCh38
rs994699702 Chr.9: 134909883-134909883 on Build GRCh38
rs746559178 Chr.9: 134909917-134909917 on Build GRCh38
rs761311523 Chr.9: 134909803-134909803 on Build GRCh38
rs755053481 Chr.9: 134909701-134909701 on Build GRCh38
rs748248387 Chr.9: 134909702-134909702 on Build GRCh38
rs1071583 Chr.9: 134909954-134909954 on Build GRCh38
rs771275627 Chr.9: 134909925-134909925 on Build GRCh38
rs909785632 Chr.9: 134909890-134909890 on Build GRCh38
rs1128429 Chr.9: 134909832-134909832 on Build GRCh38
rs2989735 Chr.9: 134909570-134909570 on Build GRCh38
rs768156833 Chr.9: 134909795-134909795 on Build GRCh38
rs368429986 Chr.9: 134909968-134909968 on Build GRCh38
rs553506777 Chr.9: 134909852-134909852 on Build GRCh38
rs757144814 Chr.9: 134909712-134909712 on Build GRCh38
rs752060354 Chr.9: 134909530-134909530 on Build GRCh38
rs750655186 Chr.9: 134909560-134909560 on Build GRCh38
rs752706989 Chr.9: 134909761-134909761 on Build GRCh38
rs138055828 Chr.9: 134909913-134909913 on Build GRCh38
rs761992771 Chr.9: 134909618-134909619 on Build GRCh38
rs189788577 Chr.9: 134909692-134909692 on Build GRCh38
rs765314208 Chr.9: 134909809-134909809 on Build GRCh38
rs761444480 Chr.9: 134909738-134909738 on Build GRCh38
rs762509396 Chr.9: 134909663-134909663 on Build GRCh38
rs145707204 Chr.9: 134909949-134909949 on Build GRCh38
rs56094122 Chr.9: 134909943-134909943 on Build GRCh38
rs951253557 Chr.9: 134909871-134909871 on Build GRCh38
rs141838661 Chr.9: 134909963-134909963 on Build GRCh38
rs757062307 Chr.9: 134909735-134909735 on Build GRCh38
rs758483750 Chr.9: 134909768-134909768 on Build GRCh38
rs773851990 Chr.9: 134909801-134909801 on Build GRCh38
rs565106056 Chr.9: 134909594-134909594 on Build GRCh38
rs571367422 Chr.9: 134909720-134909720 on Build GRCh38
rs778695287 Chr.9: 134909895-134909895 on Build GRCh38
rs756358865 Chr.9: 134909562-134909562 on Build GRCh38
rs754438411 Chr.9: 134909856-134909856 on Build GRCh38
rs776820698 Chr.9: 134909805-134909805 on Build GRCh38
rs180975999 Chr.9: 134909737-134909737 on Build GRCh38
rs745599820 Chr.9: 134909721-134909721 on Build GRCh38
rs777208442 Chr.9: 134909926-134909926 on Build GRCh38
rs758083254 Chr.9: 134909975-134909975 on Build GRCh38

View Full Product Details