Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136116706-136117208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf69
Entrez Gene ID
90120
Gene Name
chromosome 9 open reading frame 69
Gene Aliases
-
UniGene
Hs.287411
Gene Chromosome Location
Chr.9: 136111838-136118885 on Build GRCh38
Transcripts
NM_001256526.1 NM_152833.2 NR_134506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3375172 c.87C>T p.R29R Chr.9: 136116814-136116814 on Build GRCh38
3395667 c.186C>T p.C62C Chr.9: 136116715-136116715 on Build GRCh38
4163531 c.18A>C p.T6T Chr.9: 136116957-136116957 on Build GRCh38
7358802 c.256G>T p.A86S Chr.9: 136116719-136116719 on Build GRCh38
1155097 c.81C>T p.P27P Chr.9: 136116820-136116820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200364361 Chr.9: 136116730-136116730 on Build GRCh38
rs758362988 Chr.9: 136116844-136116844 on Build GRCh38
rs762421391 Chr.9: 136116963-136116963 on Build GRCh38
rs767967814 Chr.9: 136116866-136116885 on Build GRCh38
rs767981891 Chr.9: 136116822-136116822 on Build GRCh38
rs770678506 Chr.9: 136116791-136116791 on Build GRCh38
rs1011671300 Chr.9: 136116938-136116938 on Build GRCh38
rs887367517 Chr.9: 136117105-136117105 on Build GRCh38
rs371262861 Chr.9: 136116729-136116729 on Build GRCh38
rs372684658 Chr.9: 136116866-136116866 on Build GRCh38
rs755827958 Chr.9: 136116909-136116933 on Build GRCh38
rs919990116 Chr.9: 136117093-136117093 on Build GRCh38
rs776403766 Chr.9: 136116858-136116858 on Build GRCh38
rs568063226 Chr.9: 136117119-136117119 on Build GRCh38
rs371618276 Chr.9: 136116778-136116778 on Build GRCh38
rs78672817 Chr.9: 136117150-136117150 on Build GRCh38
rs778298587 Chr.9: 136116751-136116751 on Build GRCh38
rs773012855 Chr.9: 136116785-136116785 on Build GRCh38
rs770768308 Chr.9: 136116856-136116856 on Build GRCh38
rs369409953 Chr.9: 136116804-136116804 on Build GRCh38
rs980336868 Chr.9: 136117017-136117017 on Build GRCh38
rs780009833 Chr.9: 136116840-136116840 on Build GRCh38
rs753650432 Chr.9: 136117066-136117066 on Build GRCh38
rs35975639 Chr.9: 136117024-136117024 on Build GRCh38
rs904267227 Chr.9: 136117134-136117134 on Build GRCh38
rs745639465 Chr.9: 136116912-136116912 on Build GRCh38
rs117582746 Chr.9: 136116760-136116760 on Build GRCh38
rs13297832 Chr.9: 136117008-136117008 on Build GRCh38
rs769441947 Chr.9: 136116863-136116863 on Build GRCh38
rs367774660 Chr.9: 136116745-136116745 on Build GRCh38
rs948147801 Chr.9: 136117076-136117076 on Build GRCh38
rs531407777 Chr.9: 136117071-136117071 on Build GRCh38
rs189908080 Chr.9: 136116936-136116936 on Build GRCh38
rs370019472 Chr.9: 136116846-136116846 on Build GRCh38
rs773480177 Chr.9: 136117014-136117014 on Build GRCh38
rs754736865 Chr.9: 136117104-136117104 on Build GRCh38
rs1053145264 Chr.9: 136116921-136116921 on Build GRCh38
rs2985312 Chr.9: 136116942-136116942 on Build GRCh38
rs900320547 Chr.9: 136116948-136116948 on Build GRCh38
rs747013063 Chr.9: 136116900-136116900 on Build GRCh38
rs530294680 Chr.9: 136116937-136116937 on Build GRCh38
rs750154236 Chr.9: 136116907-136116908 on Build GRCh38
rs996047031 Chr.9: 136116952-136116952 on Build GRCh38
rs765966711 Chr.9: 136116732-136116732 on Build GRCh38
rs538274783 Chr.9: 136117082-136117082 on Build GRCh38
rs200259528 Chr.9: 136116805-136116805 on Build GRCh38
rs764661747 Chr.9: 136116801-136116801 on Build GRCh38
rs373479265 Chr.9: 136116813-136116813 on Build GRCh38
rs556439741 Chr.9: 136116733-136116733 on Build GRCh38
rs766583748 Chr.9: 136116892-136116892 on Build GRCh38
rs931352358 Chr.9: 136117102-136117102 on Build GRCh38
rs1053069885 Chr.9: 136117074-136117074 on Build GRCh38
rs954659867 Chr.9: 136116954-136116954 on Build GRCh38
rs552640540 Chr.9: 136117049-136117049 on Build GRCh38
rs753971897 Chr.9: 136116894-136116894 on Build GRCh38
rs938662171 Chr.9: 136116913-136116914 on Build GRCh38
rs1021424367 Chr.9: 136117133-136117133 on Build GRCh38
rs1030660324 Chr.9: 136116953-136116953 on Build GRCh38
rs758876273 Chr.9: 136116747-136116747 on Build GRCh38
rs762407047 Chr.9: 136116874-136116874 on Build GRCh38
rs754143695 Chr.9: 136117046-136117046 on Build GRCh38
rs944263255 Chr.9: 136117115-136117115 on Build GRCh38
rs752082199 Chr.9: 136116808-136116808 on Build GRCh38
rs571914564 Chr.9: 136117152-136117152 on Build GRCh38
rs974170615 Chr.9: 136116848-136116849 on Build GRCh38
rs1041622978 Chr.9: 136117118-136117118 on Build GRCh38
rs1014544823 Chr.9: 136116990-136116990 on Build GRCh38
rs1031630067 Chr.9: 136117034-136117034 on Build GRCh38
rs757130510 Chr.9: 136116905-136116905 on Build GRCh38
rs762274810 Chr.9: 136116858-136116859 on Build GRCh38
rs535262478 Chr.9: 136117130-136117130 on Build GRCh38
rs780874493 Chr.9: 136116909-136116909 on Build GRCh38
rs973576399 Chr.9: 136117006-136117006 on Build GRCh38
rs760418595 Chr.9: 136116790-136116790 on Build GRCh38
rs775257197 Chr.9: 136116932-136116932 on Build GRCh38
rs755095216 Chr.9: 136116895-136116895 on Build GRCh38
rs748038309 Chr.9: 136116781-136116781 on Build GRCh38
rs780175993 Chr.9: 136116775-136116775 on Build GRCh38
rs938923636 Chr.9: 136117038-136117038 on Build GRCh38
rs746923633 Chr.9: 136116849-136116849 on Build GRCh38
rs774897844 Chr.9: 136116867-136116867 on Build GRCh38
rs527743429 Chr.9: 136117019-136117019 on Build GRCh38
rs928865494 Chr.9: 136117035-136117035 on Build GRCh38
rs201211026 Chr.9: 136116757-136116757 on Build GRCh38
rs148372943 Chr.9: 136117092-136117092 on Build GRCh38
rs750679178 Chr.9: 136116829-136116829 on Build GRCh38
rs776310987 Chr.9: 136116797-136116797 on Build GRCh38
rs375770223 Chr.9: 136116836-136116836 on Build GRCh38
rs560557321 Chr.9: 136116995-136116995 on Build GRCh38
rs779718781 Chr.9: 136116931-136116932 on Build GRCh38
rs749339693 Chr.9: 136116841-136116841 on Build GRCh38
rs771987508 Chr.9: 136116782-136116782 on Build GRCh38
rs763675348 Chr.9: 136116872-136116872 on Build GRCh38
rs764610468 Chr.9: 136116748-136116768 on Build GRCh38
rs756312560 Chr.9: 136116878-136116878 on Build GRCh38
rs745524266 Chr.9: 136116862-136116862 on Build GRCh38
rs143889132 Chr.9: 136116798-136116798 on Build GRCh38
rs574533266 Chr.9: 136116738-136116738 on Build GRCh38
rs766626296 Chr.9: 136117018-136117018 on Build GRCh38
rs3739466 Chr.9: 136116957-136116957 on Build GRCh38
rs767996342 Chr.9: 136116875-136116875 on Build GRCh38
rs750739917 Chr.9: 136116876-136116876 on Build GRCh38
rs13292751 Chr.9: 136117007-136117007 on Build GRCh38
rs746067229 Chr.9: 136116766-136116766 on Build GRCh38
rs761140863 Chr.9: 136117015-136117015 on Build GRCh38
rs185051077 Chr.9: 136116768-136116768 on Build GRCh38
rs751898519 Chr.9: 136116781-136116783 on Build GRCh38
rs959951649 Chr.9: 136117047-136117047 on Build GRCh38
rs769448638 Chr.9: 136116916-136116916 on Build GRCh38
rs376959105 Chr.9: 136116868-136116868 on Build GRCh38
rs900028542 Chr.9: 136117124-136117124 on Build GRCh38

View Full Product Details