Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136724064-136724564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM69B
Entrez Gene ID
138311
Gene Name
family with sequence similarity 69 member B
Gene Aliases
C9orf136, pp6977
UniGene
Hs.741305
Gene Chromosome Location
Chr.9: 136712572-136724718 on Build GRCh38
Transcripts
NM_152421.3 XM_006716954.3 XM_011518238.2 XM_017014273.1

SNP Detail

SNP ID Chromosome Location
rs895508198 Chr.9: 136724455-136724455 on Build GRCh38
rs772769334 Chr.9: 136724128-136724128 on Build GRCh38
rs560017473 Chr.9: 136724452-136724452 on Build GRCh38
rs1014503424 Chr.9: 136724243-136724243 on Build GRCh38
rs936997352 Chr.9: 136724359-136724359 on Build GRCh38
rs532365111 Chr.9: 136724473-136724473 on Build GRCh38
rs1025231316 Chr.9: 136724464-136724464 on Build GRCh38
rs185818259 Chr.9: 136724407-136724407 on Build GRCh38
rs952843154 Chr.9: 136724507-136724507 on Build GRCh38
rs760747179 Chr.9: 136724252-136724252 on Build GRCh38
rs908618048 Chr.9: 136724522-136724522 on Build GRCh38
rs553260301 Chr.9: 136724335-136724335 on Build GRCh38
rs912024209 Chr.9: 136724091-136724091 on Build GRCh38
rs1021364634 Chr.9: 136724224-136724224 on Build GRCh38
rs1041577555 Chr.9: 136724254-136724254 on Build GRCh38
rs932494421 Chr.9: 136724405-136724405 on Build GRCh38
rs372644767 Chr.9: 136724353-136724353 on Build GRCh38
rs779808947 Chr.9: 136724281-136724281 on Build GRCh38
rs920009174 Chr.9: 136724339-136724339 on Build GRCh38
rs937049222 Chr.9: 136724200-136724200 on Build GRCh38
rs116901191 Chr.9: 136724238-136724238 on Build GRCh38
rs1025632513 Chr.9: 136724318-136724318 on Build GRCh38
rs562211951 Chr.9: 136724429-136724429 on Build GRCh38
rs970003938 Chr.9: 136724467-136724467 on Build GRCh38
rs531863121 Chr.9: 136724542-136724542 on Build GRCh38
rs982792480 Chr.9: 136724352-136724352 on Build GRCh38
rs943940768 Chr.9: 136724118-136724118 on Build GRCh38
rs1026459646 Chr.9: 136724489-136724489 on Build GRCh38
rs899688015 Chr.9: 136724151-136724151 on Build GRCh38
rs966617226 Chr.9: 136724094-136724094 on Build GRCh38
rs776871156 Chr.9: 136724309-136724309 on Build GRCh38
rs755692566 Chr.9: 136724096-136724096 on Build GRCh38
rs998534295 Chr.9: 136724451-136724451 on Build GRCh38
rs957193207 Chr.9: 136724293-136724293 on Build GRCh38
rs982887285 Chr.9: 136724518-136724518 on Build GRCh38
rs945383 Chr.9: 136724386-136724386 on Build GRCh38
rs562629408 Chr.9: 136724527-136724527 on Build GRCh38
rs889065415 Chr.9: 136724454-136724454 on Build GRCh38
rs930903923 Chr.9: 136724125-136724125 on Build GRCh38
rs1027125452 Chr.9: 136724474-136724474 on Build GRCh38
rs1019438911 Chr.9: 136724311-136724311 on Build GRCh38
rs78014636 Chr.9: 136724432-136724432 on Build GRCh38
rs770936521 Chr.9: 136724263-136724263 on Build GRCh38
rs953383510 Chr.9: 136724308-136724308 on Build GRCh38
rs34691592 Chr.9: 136724491-136724491 on Build GRCh38
rs1000082088 Chr.9: 136724284-136724284 on Build GRCh38
rs899739671 Chr.9: 136724402-136724402 on Build GRCh38
rs748647951 Chr.9: 136724110-136724110 on Build GRCh38
rs991212984 Chr.9: 136724360-136724360 on Build GRCh38
rs1055962235 Chr.9: 136724237-136724237 on Build GRCh38
rs952243273 Chr.9: 136724349-136724349 on Build GRCh38
rs1041346492 Chr.9: 136724132-136724132 on Build GRCh38
rs538943817 Chr.9: 136724357-136724357 on Build GRCh38
rs969615121 Chr.9: 136724334-136724334 on Build GRCh38
rs909561091 Chr.9: 136724189-136724189 on Build GRCh38
rs574559874 Chr.9: 136724412-136724412 on Build GRCh38
rs117283590 Chr.9: 136724340-136724340 on Build GRCh38
rs996632516 Chr.9: 136724188-136724188 on Build GRCh38
rs904300684 Chr.9: 136724442-136724442 on Build GRCh38
rs552190287 Chr.9: 136724513-136724513 on Build GRCh38
rs887041781 Chr.9: 136724095-136724095 on Build GRCh38
rs771810717 Chr.9: 136724456-136724456 on Build GRCh38

View Full Product Details