Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136807611-136808135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC183
Entrez Gene ID
84960
Gene Name
coiled-coil domain containing 183
Gene Aliases
KIAA1984, PARF, bA216L13.7
UniGene
Hs.728796
Gene Chromosome Location
Chr.9: 136796338-136807741 on Build GRCh38
Transcripts
NM_001039374.4 NR_024580.1 NM_001173988.1 NM_001173989.2 NM_024718.4

Gene Symbol
CCDC183-AS1
Entrez Gene ID
100131193
Gene Name
CCDC183 antisense RNA 1
Gene Aliases
KIAA1984-AS1
UniGene
Hs.734812
Gene Chromosome Location
Chr.9: 136803927-136808848 on Build GRCh38
Transcripts
NM_001039374.4 NR_024580.1 NM_001173988.1 NM_001173989.2 NM_024718.4

Gene Symbol
RABL6
Entrez Gene ID
55684
Gene Name
RAB, member RAS oncogene family-like 6
Gene Aliases
C9orf86, PARF, RBEL1, pp8875
UniGene
Hs.370555
Gene Chromosome Location
Chr.9: 136807922-136841187 on Build GRCh38
Transcripts
NM_001039374.4 NR_024580.1 NM_001173988.1 NM_001173989.2 NM_024718.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4952799 c.1545G>C p.K515N Chr.9: 136807630-136807630 on Build GRCh38
7204903 c.1526_1527insGTCC p.R511fs*>26 Chr.9: 136807611-136807612 on Build GRCh38
3216294 c.1594_1596delAAG p.K534delK Chr.9: 136807679-136807681 on Build GRCh38
6694511 c.1589A>C p.K530T Chr.9: 136807674-136807674 on Build GRCh38
5467646 c.1596G>T p.K532N Chr.9: 136807681-136807681 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs936537508 Chr.9: 136807839-136807839 on Build GRCh38
rs943913523 Chr.9: 136807958-136807958 on Build GRCh38
rs1052118041 Chr.9: 136807888-136807889 on Build GRCh38
rs920048810 Chr.9: 136807924-136807924 on Build GRCh38
rs965217110 Chr.9: 136808029-136808029 on Build GRCh38
rs747531950 Chr.9: 136807740-136807740 on Build GRCh38
rs1002148538 Chr.9: 136807893-136807893 on Build GRCh38
rs201531777 Chr.9: 136807643-136807643 on Build GRCh38
rs1055857801 Chr.9: 136807983-136807983 on Build GRCh38
rs748510317 Chr.9: 136807729-136807729 on Build GRCh38
rs1050117709 Chr.9: 136807685-136807685 on Build GRCh38
rs960307049 Chr.9: 136807908-136807908 on Build GRCh38
rs1034597188 Chr.9: 136807801-136807801 on Build GRCh38
rs542518620 Chr.9: 136807796-136807796 on Build GRCh38
rs929973107 Chr.9: 136808056-136808056 on Build GRCh38
rs549938078 Chr.9: 136807710-136807710 on Build GRCh38
rs755218301 Chr.9: 136807707-136807707 on Build GRCh38
rs374335664 Chr.9: 136807781-136807781 on Build GRCh38
rs1034546301 Chr.9: 136807904-136807904 on Build GRCh38
rs896004210 Chr.9: 136807997-136807997 on Build GRCh38
rs752162276 Chr.9: 136807813-136807813 on Build GRCh38
rs527512962 Chr.9: 136807802-136807802 on Build GRCh38
rs994673368 Chr.9: 136807792-136807792 on Build GRCh38
rs1027131325 Chr.9: 136807794-136807794 on Build GRCh38
rs952996334 Chr.9: 136807795-136807795 on Build GRCh38
rs549860502 Chr.9: 136807962-136807962 on Build GRCh38
rs889544483 Chr.9: 136807637-136807637 on Build GRCh38
rs957998391 Chr.9: 136808018-136808018 on Build GRCh38
rs751532733 Chr.9: 136807677-136807677 on Build GRCh38
rs11537630 Chr.9: 136808016-136808016 on Build GRCh38
rs1041135537 Chr.9: 136807749-136807749 on Build GRCh38
rs1014418283 Chr.9: 136808005-136808005 on Build GRCh38
rs911168040 Chr.9: 136807861-136807861 on Build GRCh38
rs533267449 Chr.9: 136807932-136807932 on Build GRCh38
rs909897700 Chr.9: 136808096-136808097 on Build GRCh38
rs759937715 Chr.9: 136807679-136807681 on Build GRCh38
rs765992841 Chr.9: 136807646-136807646 on Build GRCh38
rs957880762 Chr.9: 136807944-136807944 on Build GRCh38
rs1020500715 Chr.9: 136808010-136808010 on Build GRCh38
rs972801146 Chr.9: 136807919-136807919 on Build GRCh38
rs765077386 Chr.9: 136807668-136807668 on Build GRCh38
rs1049672424 Chr.9: 136807852-136807852 on Build GRCh38
rs561532790 Chr.9: 136807800-136807800 on Build GRCh38
rs202106813 Chr.9: 136807673-136807673 on Build GRCh38
rs564265514 Chr.9: 136807887-136807887 on Build GRCh38
rs776366172 Chr.9: 136807654-136807654 on Build GRCh38
rs976659680 Chr.9: 136808040-136808040 on Build GRCh38
rs917651181 Chr.9: 136807814-136807814 on Build GRCh38
rs999623269 Chr.9: 136808017-136808017 on Build GRCh38
rs562505462 Chr.9: 136807719-136807719 on Build GRCh38
rs1014470502 Chr.9: 136807914-136807914 on Build GRCh38
rs1008274013 Chr.9: 136807717-136807717 on Build GRCh38
rs1041038133 Chr.9: 136807868-136807868 on Build GRCh38
rs991810591 Chr.9: 136807803-136807803 on Build GRCh38
rs759168919 Chr.9: 136807659-136807659 on Build GRCh38
rs758288676 Chr.9: 136807828-136807828 on Build GRCh38
rs966779838 Chr.9: 136807931-136807931 on Build GRCh38
rs897179102 Chr.9: 136807875-136807876 on Build GRCh38
rs566272726 Chr.9: 136808043-136808043 on Build GRCh38
rs931304907 Chr.9: 136807929-136807929 on Build GRCh38
rs911071293 Chr.9: 136807957-136807957 on Build GRCh38
rs758337931 Chr.9: 136807675-136807675 on Build GRCh38
rs570088341 Chr.9: 136807963-136807963 on Build GRCh38
rs549527211 Chr.9: 136808027-136808027 on Build GRCh38
rs547668516 Chr.9: 136807821-136807821 on Build GRCh38
rs756117040 Chr.9: 136807684-136807684 on Build GRCh38
rs994226168 Chr.9: 136807876-136807876 on Build GRCh38
rs944009884 Chr.9: 136807865-136807865 on Build GRCh38
rs1021220996 Chr.9: 136807726-136807726 on Build GRCh38
rs925065199 Chr.9: 136807939-136807939 on Build GRCh38
rs1009863932 Chr.9: 136807699-136807699 on Build GRCh38
rs966455553 Chr.9: 136807820-136807820 on Build GRCh38
rs780118367 Chr.9: 136807704-136807704 on Build GRCh38
rs985322995 Chr.9: 136807954-136807954 on Build GRCh38
rs1024571906 Chr.9: 136807918-136807918 on Build GRCh38
rs756557466 Chr.9: 136807785-136807785 on Build GRCh38
rs985228219 Chr.9: 136808026-136808026 on Build GRCh38
rs984555337 Chr.9: 136808065-136808065 on Build GRCh38
rs773814791 Chr.9: 136807738-136807738 on Build GRCh38
rs576760799 Chr.9: 136807644-136807644 on Build GRCh38
rs749495333 Chr.9: 136807706-136807706 on Build GRCh38
rs772506479 Chr.9: 136807730-136807730 on Build GRCh38
rs937402399 Chr.9: 136808105-136808105 on Build GRCh38
rs918536091 Chr.9: 136808050-136808050 on Build GRCh38
rs976669508 Chr.9: 136807959-136807959 on Build GRCh38
rs925100620 Chr.9: 136807835-136807835 on Build GRCh38
rs1048470392 Chr.9: 136807965-136807965 on Build GRCh38

View Full Product Details