Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136829306-136829805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABL6
Entrez Gene ID
55684
Gene Name
RAB, member RAS oncogene family-like 6
Gene Aliases
C9orf86, PARF, RBEL1, pp8875
UniGene
Hs.370555
Gene Chromosome Location
Chr.9: 136807922-136841187 on Build GRCh38
Transcripts
NM_001173988.1 NM_001173989.2 NM_024718.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4547554 c.425G>A p.G142E Chr.9: 136829451-136829451 on Build GRCh38
4986077 c.630G>T p.E210D Chr.9: 136829422-136829422 on Build GRCh38
4986079 c.265G>T p.E89* Chr.9: 136829422-136829422 on Build GRCh38
4986075 c.396G>T p.E132D Chr.9: 136829422-136829422 on Build GRCh38
4986076 c.396G>T p.E132D Chr.9: 136829422-136829422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192580788 Chr.9: 136829777-136829777 on Build GRCh38
rs11145898 Chr.9: 136829705-136829705 on Build GRCh38
rs372992525 Chr.9: 136829368-136829368 on Build GRCh38
rs929759888 Chr.9: 136829411-136829411 on Build GRCh38
rs190079544 Chr.9: 136829501-136829501 on Build GRCh38
rs772746996 Chr.9: 136829529-136829529 on Build GRCh38
rs143101779 Chr.9: 136829689-136829689 on Build GRCh38
rs527809056 Chr.9: 136829670-136829670 on Build GRCh38
rs545821125 Chr.9: 136829598-136829598 on Build GRCh38
rs1000126799 Chr.9: 136829595-136829595 on Build GRCh38
rs1041660065 Chr.9: 136829697-136829697 on Build GRCh38
rs533701837 Chr.9: 136829548-136829548 on Build GRCh38
rs553715732 Chr.9: 136829582-136829582 on Build GRCh38
rs772741917 Chr.9: 136829361-136829361 on Build GRCh38
rs182623196 Chr.9: 136829752-136829752 on Build GRCh38
rs751102844 Chr.9: 136829500-136829501 on Build GRCh38
rs750555655 Chr.9: 136829500-136829500 on Build GRCh38
rs770586571 Chr.9: 136829462-136829462 on Build GRCh38
rs745918278 Chr.9: 136829386-136829386 on Build GRCh38
rs116599726 Chr.9: 136829364-136829364 on Build GRCh38
rs904413836 Chr.9: 136829594-136829594 on Build GRCh38
rs1008136003 Chr.9: 136829743-136829743 on Build GRCh38
rs945014817 Chr.9: 136829569-136829569 on Build GRCh38
rs1001393379 Chr.9: 136829463-136829463 on Build GRCh38
rs772923308 Chr.9: 136829426-136829426 on Build GRCh38
rs539772701 Chr.9: 136829431-136829431 on Build GRCh38
rs562899809 Chr.9: 136829614-136829614 on Build GRCh38
rs912110086 Chr.9: 136829546-136829546 on Build GRCh38
rs376553648 Chr.9: 136829449-136829449 on Build GRCh38
rs912162892 Chr.9: 136829358-136829358 on Build GRCh38
rs572222836 Chr.9: 136829742-136829742 on Build GRCh38
rs1034724171 Chr.9: 136829616-136829616 on Build GRCh38
rs577055223 Chr.9: 136829591-136829591 on Build GRCh38
rs953499630 Chr.9: 136829531-136829531 on Build GRCh38
rs904360523 Chr.9: 136829782-136829782 on Build GRCh38
rs370873785 Chr.9: 136829654-136829654 on Build GRCh38
rs542254097 Chr.9: 136829667-136829667 on Build GRCh38
rs983779622 Chr.9: 136829784-136829784 on Build GRCh38
rs754092358 Chr.9: 136829504-136829504 on Build GRCh38
rs780549419 Chr.9: 136829525-136829525 on Build GRCh38
rs780448034 Chr.9: 136829375-136829375 on Build GRCh38
rs945792488 Chr.9: 136829734-136829734 on Build GRCh38
rs781310246 Chr.9: 136829366-136829366 on Build GRCh38
rs200215294 Chr.9: 136829467-136829467 on Build GRCh38
rs201729834 Chr.9: 136829499-136829499 on Build GRCh38
rs762437766 Chr.9: 136829652-136829652 on Build GRCh38
rs574200970 Chr.9: 136829544-136829544 on Build GRCh38
rs770441857 Chr.9: 136829434-136829434 on Build GRCh38
rs771693782 Chr.9: 136829395-136829395 on Build GRCh38
rs780096508 Chr.9: 136829502-136829502 on Build GRCh38
rs756372491 Chr.9: 136829373-136829373 on Build GRCh38
rs550251522 Chr.9: 136829733-136829733 on Build GRCh38
rs567471206 Chr.9: 136829524-136829525 on Build GRCh38
rs562130146 Chr.9: 136829669-136829669 on Build GRCh38
rs754825561 Chr.9: 136829354-136829354 on Build GRCh38
rs910830434 Chr.9: 136829698-136829698 on Build GRCh38
rs571096327 Chr.9: 136829379-136829379 on Build GRCh38
rs774650717 Chr.9: 136829394-136829394 on Build GRCh38
rs113350986 Chr.9: 136829730-136829730 on Build GRCh38
rs1014139042 Chr.9: 136829693-136829693 on Build GRCh38
rs138527911 Chr.9: 136829785-136829785 on Build GRCh38
rs564557027 Chr.9: 136829690-136829690 on Build GRCh38
rs114156024 Chr.9: 136829514-136829514 on Build GRCh38
rs754103109 Chr.9: 136829349-136829349 on Build GRCh38
rs900954173 Chr.9: 136829545-136829545 on Build GRCh38
rs545002882 Chr.9: 136829666-136829666 on Build GRCh38
rs779390521 Chr.9: 136829528-136829528 on Build GRCh38
rs972301642 Chr.9: 136829763-136829763 on Build GRCh38
rs746477116 Chr.9: 136829534-136829534 on Build GRCh38
rs917359021 Chr.9: 136829468-136829468 on Build GRCh38
rs763449455 Chr.9: 136829541-136829541 on Build GRCh38
rs760132315 Chr.9: 136829407-136829407 on Build GRCh38
rs1027790509 Chr.9: 136829683-136829683 on Build GRCh38
rs776377340 Chr.9: 136829448-136829448 on Build GRCh38
rs1014704631 Chr.9: 136829648-136829648 on Build GRCh38
rs769092443 Chr.9: 136829382-136829382 on Build GRCh38
rs765095792 Chr.9: 136829452-136829452 on Build GRCh38
rs752619444 Chr.9: 136829455-136829455 on Build GRCh38
rs565017708 Chr.9: 136829711-136829711 on Build GRCh38
rs920825130 Chr.9: 136829506-136829506 on Build GRCh38
rs894472858 Chr.9: 136829645-136829645 on Build GRCh38
rs1019143280 Chr.9: 136829745-136829745 on Build GRCh38
rs186601495 Chr.9: 136829772-136829772 on Build GRCh38
rs868845823 Chr.9: 136829450-136829450 on Build GRCh38
rs1000211335 Chr.9: 136829712-136829712 on Build GRCh38
rs1048503632 Chr.9: 136829526-136829526 on Build GRCh38
rs542896415 Chr.9: 136829369-136829369 on Build GRCh38

View Full Product Details