Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136845533-136846039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf172
Entrez Gene ID
389813
Gene Name
chromosome 9 open reading frame 172
Gene Aliases
-
UniGene
Hs.495485
Gene Chromosome Location
Chr.9: 136844415-136847345 on Build GRCh38
Transcripts
NM_001080482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317472 c.1456G>A p.E486K Chr.9: 136845870-136845870 on Build GRCh38
4924690 c.1517T>C p.L506P Chr.9: 136845931-136845931 on Build GRCh38
5428997 c.1499C>G p.P500R Chr.9: 136845913-136845913 on Build GRCh38
4423744 c.1364_1365insG p.P457fs*81 Chr.9: 136845778-136845779 on Build GRCh38
7327174 c.1127C>A p.T376K Chr.9: 136845541-136845541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760566982 Chr.9: 136845579-136845579 on Build GRCh38
rs770970200 Chr.9: 136845588-136845588 on Build GRCh38
rs780780682 Chr.9: 136845803-136845803 on Build GRCh38
rs765630338 Chr.9: 136845865-136845865 on Build GRCh38
rs773101030 Chr.9: 136845566-136845566 on Build GRCh38
rs371230590 Chr.9: 136845905-136845905 on Build GRCh38
rs761429011 Chr.9: 136845968-136845968 on Build GRCh38
rs756007351 Chr.9: 136845886-136845886 on Build GRCh38
rs866823298 Chr.9: 136845973-136845973 on Build GRCh38
rs565217311 Chr.9: 136845722-136845722 on Build GRCh38
rs772481691 Chr.9: 136845773-136845773 on Build GRCh38
rs749166836 Chr.9: 136845890-136845890 on Build GRCh38
rs754235633 Chr.9: 136845767-136845767 on Build GRCh38
rs958064310 Chr.9: 136845675-136845675 on Build GRCh38
rs763483715 Chr.9: 136845961-136845961 on Build GRCh38
rs748225899 Chr.9: 136845812-136845812 on Build GRCh38
rs1032336512 Chr.9: 136845674-136845674 on Build GRCh38
rs765793499 Chr.9: 136845775-136845775 on Build GRCh38
rs751014060 Chr.9: 136845966-136845966 on Build GRCh38
rs778221852 Chr.9: 136845690-136845690 on Build GRCh38
rs756944857 Chr.9: 136845797-136845797 on Build GRCh38
rs764134799 Chr.9: 136845619-136845619 on Build GRCh38
rs755735358 Chr.9: 136845990-136845990 on Build GRCh38
rs773294771 Chr.9: 136845820-136845820 on Build GRCh38
rs1018741444 Chr.9: 136845727-136845727 on Build GRCh38
rs753665822 Chr.9: 136846009-136846009 on Build GRCh38
rs755524596 Chr.9: 136845683-136845683 on Build GRCh38
rs188700594 Chr.9: 136846010-136846010 on Build GRCh38
rs778869237 Chr.9: 136846015-136846015 on Build GRCh38
rs753322469 Chr.9: 136845866-136845866 on Build GRCh38
rs375030600 Chr.9: 136845984-136845984 on Build GRCh38
rs745700697 Chr.9: 136845805-136845805 on Build GRCh38
rs764345831 Chr.9: 136845796-136845797 on Build GRCh38
rs755402093 Chr.9: 136845774-136845774 on Build GRCh38
rs745731145 Chr.9: 136845719-136845719 on Build GRCh38
rs768586780 Chr.9: 136845900-136845900 on Build GRCh38
rs977302415 Chr.9: 136845754-136845754 on Build GRCh38
rs767752100 Chr.9: 136845638-136845638 on Build GRCh38
rs949181550 Chr.9: 136845855-136845855 on Build GRCh38
rs201654604 Chr.9: 136845558-136845558 on Build GRCh38
rs908877783 Chr.9: 136845831-136845831 on Build GRCh38
rs764726457 Chr.9: 136845962-136845962 on Build GRCh38
rs544474087 Chr.9: 136845806-136845806 on Build GRCh38
rs761986244 Chr.9: 136845632-136845632 on Build GRCh38
rs751115612 Chr.9: 136845876-136845876 on Build GRCh38
rs774248919 Chr.9: 136845720-136845720 on Build GRCh38
rs776469636 Chr.9: 136845589-136845589 on Build GRCh38
rs761015315 Chr.9: 136845745-136845745 on Build GRCh38
rs547283300 Chr.9: 136845959-136845959 on Build GRCh38
rs767075175 Chr.9: 136845980-136845980 on Build GRCh38
rs777616846 Chr.9: 136845699-136845699 on Build GRCh38
rs750767950 Chr.9: 136845643-136845643 on Build GRCh38
rs759814625 Chr.9: 136845938-136845938 on Build GRCh38
rs776006669 Chr.9: 136845953-136845953 on Build GRCh38
rs779758813 Chr.9: 136846004-136846004 on Build GRCh38
rs775451001 Chr.9: 136845810-136845810 on Build GRCh38
rs575731810 Chr.9: 136845743-136845743 on Build GRCh38
rs766595592 Chr.9: 136845746-136845746 on Build GRCh38
rs771800176 Chr.9: 136845696-136845696 on Build GRCh38
rs746769333 Chr.9: 136845704-136845704 on Build GRCh38
rs752231389 Chr.9: 136845778-136845778 on Build GRCh38
rs1054805512 Chr.9: 136845848-136845848 on Build GRCh38
rs759993819 Chr.9: 136845860-136845860 on Build GRCh38
rs776810419 Chr.9: 136845843-136845843 on Build GRCh38
rs561417352 Chr.9: 136845926-136845926 on Build GRCh38
rs746713977 Chr.9: 136845795-136845795 on Build GRCh38
rs747747733 Chr.9: 136845561-136845561 on Build GRCh38
rs770693489 Chr.9: 136845712-136845712 on Build GRCh38
rs780950397 Chr.9: 136845879-136845879 on Build GRCh38
rs779957428 Chr.9: 136845889-136845889 on Build GRCh38
rs776614631 Chr.9: 136845716-136845716 on Build GRCh38
rs763636539 Chr.9: 136845873-136845873 on Build GRCh38
rs756515568 Chr.9: 136845662-136845662 on Build GRCh38
rs763080457 Chr.9: 136845610-136845610 on Build GRCh38
rs756817203 Chr.9: 136845878-136845878 on Build GRCh38
rs751670859 Chr.9: 136845629-136845629 on Build GRCh38
rs766657390 Chr.9: 136845832-136845832 on Build GRCh38
rs771857283 Chr.9: 136845564-136845564 on Build GRCh38
rs771034683 Chr.9: 136845918-136845918 on Build GRCh38
rs530324864 Chr.9: 136845949-136845949 on Build GRCh38
rs999467034 Chr.9: 136845673-136845673 on Build GRCh38
rs780607139 Chr.9: 136845664-136845664 on Build GRCh38
rs747136344 Chr.9: 136845912-136845912 on Build GRCh38
rs777563591 Chr.9: 136845784-136845784 on Build GRCh38
rs941778698 Chr.9: 136845557-136845557 on Build GRCh38
rs767698977 Chr.9: 136845735-136845735 on Build GRCh38
rs778614881 Chr.9: 136845560-136845560 on Build GRCh38
rs762091883 Chr.9: 136845700-136845701 on Build GRCh38
rs941650741 Chr.9: 136845839-136845839 on Build GRCh38
rs184306980 Chr.9: 136845671-136845671 on Build GRCh38
rs757964656 Chr.9: 136845869-136845869 on Build GRCh38
rs758021856 Chr.9: 136845781-136845781 on Build GRCh38
rs894857922 Chr.9: 136845849-136845849 on Build GRCh38

View Full Product Details