Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136940348-136940854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW5
Entrez Gene ID
54461
Gene Name
F-box and WD repeat domain containing 5
Gene Aliases
Fbw5
UniGene
Hs.522507
Gene Chromosome Location
Chr.9: 136940433-136944774 on Build GRCh38
Transcripts
NM_018998.3 XM_005266089.2 XM_011518789.1 XM_017014812.1

SNP Detail

SNP ID Chromosome Location
rs925937656 Chr.9: 136940828-136940828 on Build GRCh38
rs1042178777 Chr.9: 136940578-136940578 on Build GRCh38
rs554800624 Chr.9: 136940469-136940469 on Build GRCh38
rs974905738 Chr.9: 136940431-136940433 on Build GRCh38
rs1000391706 Chr.9: 136940491-136940491 on Build GRCh38
rs531412773 Chr.9: 136940714-136940714 on Build GRCh38
rs374020833 Chr.9: 136940778-136940778 on Build GRCh38
rs764939890 Chr.9: 136940460-136940460 on Build GRCh38
rs1028978380 Chr.9: 136940813-136940813 on Build GRCh38
rs988733879 Chr.9: 136940408-136940408 on Build GRCh38
rs538077811 Chr.9: 136940403-136940403 on Build GRCh38
rs1018002292 Chr.9: 136940799-136940799 on Build GRCh38
rs922004870 Chr.9: 136940570-136940570 on Build GRCh38
rs1009990053 Chr.9: 136940796-136940796 on Build GRCh38
rs1025270620 Chr.9: 136940784-136940784 on Build GRCh38
rs954807418 Chr.9: 136940712-136940712 on Build GRCh38
rs547623993 Chr.9: 136940793-136940793 on Build GRCh38
rs115270904 Chr.9: 136940581-136940581 on Build GRCh38
rs889730219 Chr.9: 136940424-136940424 on Build GRCh38
rs983892355 Chr.9: 136940537-136940537 on Build GRCh38
rs545986506 Chr.9: 136940538-136940538 on Build GRCh38
rs1055932383 Chr.9: 136940601-136940601 on Build GRCh38
rs1020407000 Chr.9: 136940478-136940478 on Build GRCh38
rs1040998765 Chr.9: 136940552-136940552 on Build GRCh38
rs867354282 Chr.9: 136940556-136940556 on Build GRCh38
rs918410301 Chr.9: 136940416-136940416 on Build GRCh38
rs1046392190 Chr.9: 136940770-136940770 on Build GRCh38
rs111631993 Chr.9: 136940670-136940670 on Build GRCh38
rs1056958608 Chr.9: 136940662-136940662 on Build GRCh38
rs939718935 Chr.9: 136940489-136940490 on Build GRCh38
rs936039291 Chr.9: 136940726-136940726 on Build GRCh38
rs1003159558 Chr.9: 136940647-136940647 on Build GRCh38
rs759743273 Chr.9: 136940791-136940791 on Build GRCh38
rs1014509408 Chr.9: 136940606-136940606 on Build GRCh38
rs930969597 Chr.9: 136940749-136940749 on Build GRCh38
rs968666916 Chr.9: 136940543-136940543 on Build GRCh38
rs60742160 Chr.9: 136940394-136940394 on Build GRCh38
rs1051014 Chr.9: 136940475-136940475 on Build GRCh38
rs554224094 Chr.9: 136940521-136940521 on Build GRCh38
rs886653369 Chr.9: 136940488-136940488 on Build GRCh38
rs113874081 Chr.9: 136940412-136940412 on Build GRCh38
rs577142047 Chr.9: 136940533-136940533 on Build GRCh38
rs1024602128 Chr.9: 136940608-136940608 on Build GRCh38
rs566931327 Chr.9: 136940445-136940446 on Build GRCh38
rs1000297928 Chr.9: 136940594-136940594 on Build GRCh38
rs576452073 Chr.9: 136940561-136940561 on Build GRCh38
rs952441635 Chr.9: 136940622-136940622 on Build GRCh38
rs1009004185 Chr.9: 136940825-136940826 on Build GRCh38
rs1048282529 Chr.9: 136940419-136940419 on Build GRCh38
rs762212778 Chr.9: 136940534-136940534 on Build GRCh38
rs1010728747 Chr.9: 136940477-136940477 on Build GRCh38
rs911046400 Chr.9: 136940653-136940653 on Build GRCh38
rs534063619 Chr.9: 136940516-136940516 on Build GRCh38
rs767316721 Chr.9: 136940380-136940380 on Build GRCh38
rs756238878 Chr.9: 136940685-136940685 on Build GRCh38
rs568980362 Chr.9: 136940713-136940713 on Build GRCh38
rs891730427 Chr.9: 136940664-136940666 on Build GRCh38
rs958918959 Chr.9: 136940510-136940510 on Build GRCh38
rs904669811 Chr.9: 136940483-136940483 on Build GRCh38
rs904445148 Chr.9: 136940801-136940801 on Build GRCh38
rs946091597 Chr.9: 136940567-136940567 on Build GRCh38
rs1034529004 Chr.9: 136940496-136940496 on Build GRCh38
rs931188150 Chr.9: 136940417-136940417 on Build GRCh38
rs761443954 Chr.9: 136940448-136940449 on Build GRCh38
rs1044177977 Chr.9: 136940620-136940620 on Build GRCh38
rs977747429 Chr.9: 136940826-136940826 on Build GRCh38
rs570767934 Chr.9: 136940797-136940797 on Build GRCh38
rs548438411 Chr.9: 136940716-136940716 on Build GRCh38
rs1040983356 Chr.9: 136940453-136940453 on Build GRCh38
rs922443910 Chr.9: 136940823-136940823 on Build GRCh38
rs776899900 Chr.9: 136940766-136940766 on Build GRCh38
rs914698702 Chr.9: 136940417-136940418 on Build GRCh38
rs942635039 Chr.9: 136940438-136940438 on Build GRCh38
rs1014450084 Chr.9: 136940769-136940769 on Build GRCh38
rs972894288 Chr.9: 136940520-136940520 on Build GRCh38
rs968696678 Chr.9: 136940681-136940681 on Build GRCh38
rs1025746911 Chr.9: 136940621-136940621 on Build GRCh38
rs12344250 Chr.9: 136940722-136940722 on Build GRCh38
rs990454465 Chr.9: 136940389-136940389 on Build GRCh38
rs901180193 Chr.9: 136940468-136940468 on Build GRCh38
rs947462297 Chr.9: 136940425-136940425 on Build GRCh38
rs1029529437 Chr.9: 136940692-136940692 on Build GRCh38
rs147091042 Chr.9: 136940551-136940551 on Build GRCh38
rs539863955 Chr.9: 136940812-136940812 on Build GRCh38
rs568392765 Chr.9: 136940511-136940511 on Build GRCh38
rs1041693784 Chr.9: 136940719-136940719 on Build GRCh38
rs1044065589 Chr.9: 136940750-136940750 on Build GRCh38
rs898049041 Chr.9: 136940775-136940775 on Build GRCh38
rs963885823 Chr.9: 136940658-136940658 on Build GRCh38
rs987652128 Chr.9: 136940583-136940583 on Build GRCh38
rs867287130 Chr.9: 136940688-136940688 on Build GRCh38
rs1005255360 Chr.9: 136940792-136940792 on Build GRCh38
rs3750516 Chr.9: 136940617-136940617 on Build GRCh38
rs866710413 Chr.9: 136940680-136940680 on Build GRCh38
rs866445033 Chr.9: 136940731-136940731 on Build GRCh38
rs949119214 Chr.9: 136940396-136940396 on Build GRCh38
rs150230588 Chr.9: 136940676-136940676 on Build GRCh38
rs967280784 Chr.9: 136940825-136940825 on Build GRCh38
rs908021169 Chr.9: 136940717-136940717 on Build GRCh38
rs551874741 Chr.9: 136940704-136940704 on Build GRCh38
rs566973231 Chr.9: 136940718-136940718 on Build GRCh38
rs138442305 Chr.9: 136940605-136940605 on Build GRCh38
rs983843206 Chr.9: 136940646-136940646 on Build GRCh38
rs766419131 Chr.9: 136940691-136940691 on Build GRCh38
rs970456019 Chr.9: 136940517-136940517 on Build GRCh38
rs528015538 Chr.9: 136940616-136940616 on Build GRCh38
rs894646996 Chr.9: 136940767-136940767 on Build GRCh38
rs976580993 Chr.9: 136940819-136940819 on Build GRCh38
rs1055924027 Chr.9: 136940754-136940754 on Build GRCh38
rs151332880 Chr.9: 136940384-136940384 on Build GRCh38
rs995239606 Chr.9: 136940509-136940509 on Build GRCh38
rs534102241 Chr.9: 136940772-136940772 on Build GRCh38

View Full Product Details