Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136941129-136941622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW5
Entrez Gene ID
54461
Gene Name
F-box and WD repeat domain containing 5
Gene Aliases
Fbw5
UniGene
Hs.522507
Gene Chromosome Location
Chr.9: 136940433-136944774 on Build GRCh38
Transcripts
NM_018998.3 XM_005266089.2 XM_011518789.1 XM_017014812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6662473 c.1255G>A p.V419M Chr.9: 136941453-136941453 on Build GRCh38
4651669 c.1181C>T p.A394V Chr.9: 136941600-136941600 on Build GRCh38
5821326 c.697G>A p.V233I Chr.9: 136941589-136941589 on Build GRCh38
6662470 c.1346A>G p.D449G Chr.9: 136941362-136941362 on Build GRCh38
5652700 c.1362G>T p.R454R Chr.9: 136941346-136941346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777973791 Chr.9: 136941370-136941370 on Build GRCh38
rs745525167 Chr.9: 136941342-136941342 on Build GRCh38
rs765978255 Chr.9: 136941298-136941298 on Build GRCh38
rs762147478 Chr.9: 136941546-136941546 on Build GRCh38
rs753006959 Chr.9: 136941408-136941408 on Build GRCh38
rs760331772 Chr.9: 136941297-136941297 on Build GRCh38
rs745792984 Chr.9: 136941544-136941544 on Build GRCh38
rs138273782 Chr.9: 136941350-136941350 on Build GRCh38
rs144092114 Chr.9: 136941521-136941521 on Build GRCh38
rs751445990 Chr.9: 136941308-136941308 on Build GRCh38
rs773355737 Chr.9: 136941590-136941590 on Build GRCh38
rs537752014 Chr.9: 136941431-136941431 on Build GRCh38
rs770751292 Chr.9: 136941538-136941538 on Build GRCh38
rs144664027 Chr.9: 136941200-136941200 on Build GRCh38
rs569722013 Chr.9: 136941384-136941384 on Build GRCh38
rs572141771 Chr.9: 136941489-136941489 on Build GRCh38
rs376577916 Chr.9: 136941488-136941488 on Build GRCh38
rs747777692 Chr.9: 136941478-136941478 on Build GRCh38
rs367827758 Chr.9: 136941274-136941274 on Build GRCh38
rs768895155 Chr.9: 136941545-136941545 on Build GRCh38
rs767969786 Chr.9: 136941459-136941459 on Build GRCh38
rs111761422 Chr.9: 136941336-136941336 on Build GRCh38
rs201094330 Chr.9: 136941476-136941476 on Build GRCh38
rs370227114 Chr.9: 136941239-136941239 on Build GRCh38
rs779965747 Chr.9: 136941181-136941181 on Build GRCh38
rs759237517 Chr.9: 136941301-136941301 on Build GRCh38
rs374722743 Chr.9: 136941501-136941501 on Build GRCh38
rs764752014 Chr.9: 136941562-136941562 on Build GRCh38
rs777588724 Chr.9: 136941565-136941565 on Build GRCh38
rs763005819 Chr.9: 136941496-136941496 on Build GRCh38
rs201429106 Chr.9: 136941364-136941364 on Build GRCh38
rs370826295 Chr.9: 136941224-136941224 on Build GRCh38
rs377369299 Chr.9: 136941201-136941201 on Build GRCh38
rs376123602 Chr.9: 136941352-136941352 on Build GRCh38
rs200276837 Chr.9: 136941300-136941300 on Build GRCh38
rs372226218 Chr.9: 136941574-136941574 on Build GRCh38
rs768666071 Chr.9: 136941183-136941183 on Build GRCh38
rs926633709 Chr.9: 136941460-136941460 on Build GRCh38
rs775269844 Chr.9: 136941385-136941385 on Build GRCh38
rs368257696 Chr.9: 136941469-136941469 on Build GRCh38
rs777063269 Chr.9: 136941155-136941155 on Build GRCh38
rs778768955 Chr.9: 136941533-136941533 on Build GRCh38
rs761706594 Chr.9: 136941353-136941353 on Build GRCh38
rs143868121 Chr.9: 136941388-136941388 on Build GRCh38
rs760052894 Chr.9: 136941602-136941602 on Build GRCh38
rs372957829 Chr.9: 136941415-136941415 on Build GRCh38
rs765513807 Chr.9: 136941382-136941384 on Build GRCh38
rs370063089 Chr.9: 136941600-136941600 on Build GRCh38
rs1031286437 Chr.9: 136941487-136941487 on Build GRCh38
rs535554815 Chr.9: 136941204-136941204 on Build GRCh38
rs368712198 Chr.9: 136941474-136941474 on Build GRCh38
rs777351828 Chr.9: 136941568-136941568 on Build GRCh38
rs945452936 Chr.9: 136941592-136941592 on Build GRCh38
rs868572748 Chr.9: 136941168-136941168 on Build GRCh38
rs757637555 Chr.9: 136941316-136941316 on Build GRCh38
rs772731220 Chr.9: 136941246-136941246 on Build GRCh38
rs34849272 Chr.9: 136941584-136941585 on Build GRCh38
rs548960687 Chr.9: 136941393-136941393 on Build GRCh38
rs555399084 Chr.9: 136941208-136941208 on Build GRCh38
rs541956434 Chr.9: 136941344-136941344 on Build GRCh38
rs746711546 Chr.9: 136941537-136941537 on Build GRCh38
rs200545656 Chr.9: 136941601-136941601 on Build GRCh38
rs773186295 Chr.9: 136941442-136941442 on Build GRCh38
rs201352991 Chr.9: 136941184-136941184 on Build GRCh38
rs759720702 Chr.9: 136941401-136941401 on Build GRCh38
rs771898527 Chr.9: 136941480-136941480 on Build GRCh38
rs377761216 Chr.9: 136941182-136941182 on Build GRCh38
rs376136024 Chr.9: 136941355-136941355 on Build GRCh38
rs775386923 Chr.9: 136941452-136941452 on Build GRCh38
rs776963435 Chr.9: 136941286-136941288 on Build GRCh38
rs752483874 Chr.9: 136941303-136941303 on Build GRCh38
rs771754193 Chr.9: 136941252-136941257 on Build GRCh38
rs766968739 Chr.9: 136941245-136941245 on Build GRCh38
rs781297662 Chr.9: 136941232-136941232 on Build GRCh38
rs745830013 Chr.9: 136941577-136941577 on Build GRCh38
rs112814747 Chr.9: 136941454-136941454 on Build GRCh38
rs781006039 Chr.9: 136941576-136941576 on Build GRCh38
rs759999433 Chr.9: 136941561-136941561 on Build GRCh38
rs769055293 Chr.9: 136941283-136941283 on Build GRCh38
rs148314608 Chr.9: 136941326-136941326 on Build GRCh38
rs144039568 Chr.9: 136941463-136941463 on Build GRCh38
rs752108537 Chr.9: 136941574-136941575 on Build GRCh38
rs199873362 Chr.9: 136941363-136941363 on Build GRCh38
rs367994518 Chr.9: 136941494-136941494 on Build GRCh38
rs763621568 Chr.9: 136941166-136941166 on Build GRCh38
rs749499384 Chr.9: 136941280-136941280 on Build GRCh38
rs769425591 Chr.9: 136941348-136941348 on Build GRCh38
rs564578256 Chr.9: 136941445-136941445 on Build GRCh38
rs139353750 Chr.9: 136941498-136941498 on Build GRCh38
rs374042401 Chr.9: 136941207-136941207 on Build GRCh38
rs745899081 Chr.9: 136941275-136941275 on Build GRCh38
rs561808530 Chr.9: 136941347-136941347 on Build GRCh38
rs375317859 Chr.9: 136941240-136941240 on Build GRCh38
rs780017206 Chr.9: 136941212-136941212 on Build GRCh38
rs377703274 Chr.9: 136941358-136941358 on Build GRCh38
rs934354953 Chr.9: 136941391-136941391 on Build GRCh38
rs200879994 Chr.9: 136941557-136941557 on Build GRCh38
rs754344421 Chr.9: 136941558-136941558 on Build GRCh38
rs769804182 Chr.9: 136941579-136941579 on Build GRCh38
rs201834079 Chr.9: 136941457-136941457 on Build GRCh38
rs761753439 Chr.9: 136941397-136941397 on Build GRCh38
rs753630793 Chr.9: 136941213-136941213 on Build GRCh38
rs773478949 Chr.9: 136941549-136941549 on Build GRCh38
rs201822750 Chr.9: 136941335-136941335 on Build GRCh38
rs868130570 Chr.9: 136941416-136941416 on Build GRCh38
rs371147879 Chr.9: 136941318-136941318 on Build GRCh38
rs760258773 Chr.9: 136941258-136941258 on Build GRCh38
rs750358020 Chr.9: 136941400-136941400 on Build GRCh38
rs760921049 Chr.9: 136941593-136941593 on Build GRCh38
rs746105556 Chr.9: 136941236-136941236 on Build GRCh38
rs145980462 Chr.9: 136941548-136941548 on Build GRCh38
rs772320395 Chr.9: 136941330-136941330 on Build GRCh38
rs750083255 Chr.9: 136941179-136941179 on Build GRCh38
rs779146764 Chr.9: 136941580-136941580 on Build GRCh38
rs369668241 Chr.9: 136941165-136941165 on Build GRCh38
rs372873658 Chr.9: 136941317-136941317 on Build GRCh38
rs760501718 Chr.9: 136941443-136941443 on Build GRCh38
rs748171605 Chr.9: 136941586-136941586 on Build GRCh38
rs367798532 Chr.9: 136941241-136941241 on Build GRCh38
rs764697970 Chr.9: 136941542-136941544 on Build GRCh38
rs534791664 Chr.9: 136941414-136941414 on Build GRCh38
rs752381964 Chr.9: 136941523-136941523 on Build GRCh38
rs150544333 Chr.9: 136941522-136941522 on Build GRCh38
rs748748915 Chr.9: 136941193-136941194 on Build GRCh38
rs762358306 Chr.9: 136941242-136941242 on Build GRCh38
rs771841749 Chr.9: 136941441-136941441 on Build GRCh38
rs750085450 Chr.9: 136941209-136941209 on Build GRCh38
rs374757784 Chr.9: 136941273-136941273 on Build GRCh38
rs999669605 Chr.9: 136941575-136941575 on Build GRCh38
rs549760300 Chr.9: 136941375-136941375 on Build GRCh38
rs756947005 Chr.9: 136941170-136941170 on Build GRCh38
rs755144335 Chr.9: 136941322-136941322 on Build GRCh38
rs759785463 Chr.9: 136941190-136941190 on Build GRCh38
rs753248943 Chr.9: 136941159-136941159 on Build GRCh38
rs748480629 Chr.9: 136941327-136941327 on Build GRCh38
rs763064833 Chr.9: 136941528-136941529 on Build GRCh38
rs768595180 Chr.9: 136941198-136941199 on Build GRCh38
rs200014282 Chr.9: 136941510-136941510 on Build GRCh38
rs761000831 Chr.9: 136941152-136941152 on Build GRCh38
rs755753376 Chr.9: 136941211-136941211 on Build GRCh38
rs780859496 Chr.9: 136941376-136941376 on Build GRCh38
rs748047155 Chr.9: 136941224-136941240 on Build GRCh38
rs781780796 Chr.9: 136941425-136941425 on Build GRCh38
rs780321428 Chr.9: 136941279-136941279 on Build GRCh38
rs938551061 Chr.9: 136941530-136941530 on Build GRCh38
rs764654690 Chr.9: 136941262-136941262 on Build GRCh38
rs375915640 Chr.9: 136941563-136941563 on Build GRCh38
rs768536340 Chr.9: 136941390-136941390 on Build GRCh38
rs372912376 Chr.9: 136941153-136941153 on Build GRCh38
rs1057018926 Chr.9: 136941250-136941250 on Build GRCh38
rs992172616 Chr.9: 136941583-136941583 on Build GRCh38
rs769950338 Chr.9: 136941237-136941237 on Build GRCh38
rs527442645 Chr.9: 136941157-136941157 on Build GRCh38
rs755697963 Chr.9: 136941426-136941426 on Build GRCh38
rs376744591 Chr.9: 136941531-136941531 on Build GRCh38
rs765104564 Chr.9: 136941266-136941266 on Build GRCh38
rs770610292 Chr.9: 136941341-136941341 on Build GRCh38
rs373284803 Chr.9: 136941540-136941540 on Build GRCh38
rs754882485 Chr.9: 136941264-136941264 on Build GRCh38
rs774208303 Chr.9: 136941223-136941224 on Build GRCh38
rs758849693 Chr.9: 136941368-136941368 on Build GRCh38
rs565863002 Chr.9: 136941409-136941409 on Build GRCh38
rs373520014 Chr.9: 136941171-136941171 on Build GRCh38
rs370273990 Chr.9: 136941205-136941205 on Build GRCh38
rs766063269 Chr.9: 136941259-136941259 on Build GRCh38
rs376036392 Chr.9: 136941158-136941158 on Build GRCh38
rs751206571 Chr.9: 136941569-136941569 on Build GRCh38
rs747554111 Chr.9: 136941373-136941373 on Build GRCh38
rs540737570 Chr.9: 136941495-136941495 on Build GRCh38
rs138768587 Chr.9: 136941307-136941307 on Build GRCh38
rs930596842 Chr.9: 136941222-136941222 on Build GRCh38
rs149358178 Chr.9: 136941197-136941197 on Build GRCh38
rs754356376 Chr.9: 136941513-136941513 on Build GRCh38
rs750705294 Chr.9: 136941505-136941505 on Build GRCh38
rs372563353 Chr.9: 136941289-136941289 on Build GRCh38
rs886330878 Chr.9: 136941532-136941532 on Build GRCh38
rs768022966 Chr.9: 136941504-136941504 on Build GRCh38
rs143554180 Chr.9: 136941288-136941288 on Build GRCh38
rs752620281 Chr.9: 136941269-136941269 on Build GRCh38
rs978254715 Chr.9: 136941597-136941597 on Build GRCh38
rs140009447 Chr.9: 136941424-136941424 on Build GRCh38
rs964886549 Chr.9: 136941265-136941265 on Build GRCh38
rs375877399 Chr.9: 136941589-136941589 on Build GRCh38
rs368964340 Chr.9: 136941511-136941511 on Build GRCh38
rs375678798 Chr.9: 136941225-136941225 on Build GRCh38
rs759649346 Chr.9: 136941302-136941303 on Build GRCh38
rs776777980 Chr.9: 136941186-136941186 on Build GRCh38
rs756537723 Chr.9: 136941464-136941464 on Build GRCh38
rs779572531 Chr.9: 136941180-136941181 on Build GRCh38
rs765394156 Chr.9: 136941444-136941444 on Build GRCh38
rs370733758 Chr.9: 136941437-136941437 on Build GRCh38
rs772603230 Chr.9: 136941292-136941292 on Build GRCh38
rs758054443 Chr.9: 136941567-136941567 on Build GRCh38
rs757333173 Chr.9: 136941231-136941231 on Build GRCh38
rs775730930 Chr.9: 136941479-136941480 on Build GRCh38
rs1006209018 Chr.9: 136941451-136941451 on Build GRCh38
rs139795863 Chr.9: 136941553-136941553 on Build GRCh38
rs149391188 Chr.9: 136941429-136941429 on Build GRCh38
rs755917999 Chr.9: 136941180-136941180 on Build GRCh38
rs774739687 Chr.9: 136941285-136941285 on Build GRCh38
rs767375918 Chr.9: 136941398-136941398 on Build GRCh38
rs762094350 Chr.9: 136941502-136941502 on Build GRCh38
rs989885389 Chr.9: 136941404-136941404 on Build GRCh38
rs150683637 Chr.9: 136941407-136941407 on Build GRCh38
rs146243977 Chr.9: 136941430-136941430 on Build GRCh38
rs756329824 Chr.9: 136941278-136941278 on Build GRCh38
rs752769641 Chr.9: 136941196-136941196 on Build GRCh38
rs376477610 Chr.9: 136941470-136941470 on Build GRCh38
rs763246115 Chr.9: 136941453-136941453 on Build GRCh38
rs774137088 Chr.9: 136941435-136941435 on Build GRCh38
rs751018678 Chr.9: 136941169-136941169 on Build GRCh38
rs367852952 Chr.9: 136941217-136941217 on Build GRCh38

View Full Product Details